Assay Details
Gene Symbol: TRIM14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_033219.2 | NP_150088.1 | 6 - 7 | 1521 | 145 | |
GenBank mRNA | AF220130.1 | - | 6 - 7 | 1365 | 145 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC09001403.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0900010968.hg.1 | ENST00000341469.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010968.hg.1 | ENST00000341469.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010968.hg.1 | ENST00000341469.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010968.hg.1 | ENST00000341469.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010968.hg.1 | ENST00000341469.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010968.hg.1 | ENST00000341469.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010968.hg.1 | ENST00000341469.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010968.hg.1 | ENST00000341469.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010968.hg.1 | ENST00000341469.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010968.hg.1 | ENST00000341469.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010968.hg.1 | ENST00000341469.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010968.hg.1 | ENST00000341469.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010968.hg.1 | ENST00000342043.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010968.hg.1 | ENST00000342043.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010968.hg.1 | ENST00000342043.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010968.hg.1 | ENST00000342043.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010968.hg.1 | ENST00000342043.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010968.hg.1 | ENST00000342043.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010968.hg.1 | ENST00000342043.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010968.hg.1 | ENST00000342043.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010968.hg.1 | ENST00000342043.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010968.hg.1 | ENST00000342043.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010968.hg.1 | ENST00000342043.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010968.hg.1 | ENST00000375098.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010968.hg.1 | ENST00000375098.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010968.hg.1 | ENST00000375098.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010968.hg.1 | ENST00000375098.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010968.hg.1 | ENST00000375098.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010968.hg.1 | ENST00000375098.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010968.hg.1 | ENST00000375098.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010968.hg.1 | ENST00000375098.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010968.hg.1 | ENST00000375098.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010968.hg.1 | ENST00000375098.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010968.hg.1 | ENST00000375098.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010968.hg.1 | ENST00000475147.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010968.hg.1 | ENST00000475147.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010968.hg.1 | ENST00000475147.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010968.hg.1 | ENST00000475147.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010968.hg.1 | ENST00000475147.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010968.hg.1 | ENST00000475147.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010968.hg.1 | ENST00000478401.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010968.hg.1 | ENST00000478401.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010968.hg.1 | ENST00000478401.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010968.hg.1 | ENST00000478401.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010968.hg.1 | ENST00000478401.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010968.hg.1 | ENST00000478401.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010968.hg.1 | ENST00000478530.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010968.hg.1 | ENST00000478530.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010968.hg.1 | ENST00000478530.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010968.hg.1 | ENST00000478530.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010968.hg.1 | NM_014788.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010968.hg.1 | NM_014788.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010968.hg.1 | NM_014788.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010968.hg.1 | NM_014788.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010968.hg.1 | NM_014788.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010968.hg.1 | NM_014788.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010968.hg.1 | NM_014788.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010968.hg.1 | NM_014788.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010968.hg.1 | NM_014788.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010968.hg.1 | NM_014788.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010968.hg.1 | NM_014788.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010968.hg.1 | NM_014788.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010968.hg.1 | NM_014788.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010968.hg.1 | NM_033219.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010968.hg.1 | NM_033219.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010968.hg.1 | NM_033219.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010968.hg.1 | NM_033219.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010968.hg.1 | NM_033219.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010968.hg.1 | NM_033219.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010968.hg.1 | NM_033219.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010968.hg.1 | NM_033219.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010968.hg.1 | NM_033219.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010968.hg.1 | NM_033219.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010968.hg.1 | NM_033219.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010968.hg.1 | NM_033219.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR09017445.hg.1 | TC09001403.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR09017445.hg.1 | ENST00000341469.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR09017445.hg.1 | ENST00000342043.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR09017445.hg.1 | ENST00000375084.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR09017445.hg.1 | NM_014788.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR09017445.hg.1 | NM_033219.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900174715.hg.1 | ENST00000341469.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900174715.hg.1 | ENST00000342043.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900174715.hg.1 | NM_014788.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900174715.hg.1 | NM_033219.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900174715.hg.1 | ENST00000341469.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900174715.hg.1 | ENST00000342043.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900174715.hg.1 | NM_014788.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900174715.hg.1 | NM_033219.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900174715.hg.1 | TC0900010968.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0900010968.hg.1 | ENST00000341469.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010968.hg.1 | ENST00000341469.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010968.hg.1 | ENST00000341469.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010968.hg.1 | ENST00000341469.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010968.hg.1 | ENST00000341469.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010968.hg.1 | ENST00000341469.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010968.hg.1 | ENST00000341469.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010968.hg.1 | ENST00000341469.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010968.hg.1 | ENST00000341469.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010968.hg.1 | ENST00000341469.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010968.hg.1 | ENST00000341469.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010968.hg.1 | ENST00000341469.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010968.hg.1 | ENST00000342043.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010968.hg.1 | ENST00000342043.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010968.hg.1 | ENST00000342043.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010968.hg.1 | ENST00000342043.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010968.hg.1 | ENST00000342043.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010968.hg.1 | ENST00000342043.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010968.hg.1 | ENST00000342043.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010968.hg.1 | ENST00000342043.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010968.hg.1 | ENST00000342043.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010968.hg.1 | ENST00000342043.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010968.hg.1 | ENST00000342043.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010968.hg.1 | ENST00000375098.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010968.hg.1 | ENST00000375098.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010968.hg.1 | ENST00000375098.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010968.hg.1 | ENST00000375098.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010968.hg.1 | ENST00000375098.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010968.hg.1 | ENST00000375098.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010968.hg.1 | ENST00000375098.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010968.hg.1 | ENST00000375098.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010968.hg.1 | ENST00000375098.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010968.hg.1 | ENST00000375098.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010968.hg.1 | ENST00000375098.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010968.hg.1 | ENST00000475147.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010968.hg.1 | ENST00000475147.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010968.hg.1 | ENST00000475147.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010968.hg.1 | ENST00000475147.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010968.hg.1 | ENST00000475147.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010968.hg.1 | ENST00000475147.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010968.hg.1 | ENST00000478401.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010968.hg.1 | ENST00000478401.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010968.hg.1 | ENST00000478401.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010968.hg.1 | ENST00000478401.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010968.hg.1 | ENST00000478401.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010968.hg.1 | ENST00000478401.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010968.hg.1 | ENST00000478530.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010968.hg.1 | ENST00000478530.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010968.hg.1 | ENST00000478530.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010968.hg.1 | ENST00000478530.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010968.hg.1 | NM_014788.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010968.hg.1 | NM_014788.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010968.hg.1 | NM_014788.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010968.hg.1 | NM_014788.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010968.hg.1 | NM_014788.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010968.hg.1 | NM_014788.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010968.hg.1 | NM_014788.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010968.hg.1 | NM_014788.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010968.hg.1 | NM_014788.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010968.hg.1 | NM_014788.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010968.hg.1 | NM_014788.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010968.hg.1 | NM_014788.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010968.hg.1 | NM_014788.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010968.hg.1 | NM_033219.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010968.hg.1 | NM_033219.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010968.hg.1 | NM_033219.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010968.hg.1 | NM_033219.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010968.hg.1 | NM_033219.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010968.hg.1 | NM_033219.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010968.hg.1 | NM_033219.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010968.hg.1 | NM_033219.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010968.hg.1 | NM_033219.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010968.hg.1 | NM_033219.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010968.hg.1 | NM_033219.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010968.hg.1 | NM_033219.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900174717.hg.1 | ENST00000341469.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900174717.hg.1 | ENST00000342043.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900174717.hg.1 | ENST00000375098.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900174717.hg.1 | ENST00000478530.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900174717.hg.1 | NM_014788.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900174717.hg.1 | NM_033219.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900174717.hg.1 | ENST00000341469.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900174717.hg.1 | ENST00000342043.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900174717.hg.1 | ENST00000375098.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900174717.hg.1 | ENST00000478530.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900174717.hg.1 | NM_014788.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900174717.hg.1 | NM_033219.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900174717.hg.1 | TC0900010968.hg.1 | Clariom D TC |
