Assay Details
Gene Symbol: ARNTL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030272.2 | NP_001025443.1 | 4 - 5 | 538 | 67 | |
NM_001178.5 | NP_001169.3 | 5 - 6 | 593 | 67 | ||
NM_001297719.1 | NP_001284648.1 | 5 - 6 | 593 | 67 | ||
NM_001297722.1 | NP_001284651.1 | 4 - 5 | 538 | 67 | ||
XM_006718233.3 | XP_006718296.1 | 4 - 5 | 480 | 67 | ||
XM_011520105.2 | XP_011518407.2 | 3 - 4 | 644 | 67 | ||
XM_011520107.2 | XP_011518409.2 | 3 - 4 | 639 | 67 | ||
XM_011520108.1 | XP_011518410.1 | 5 - 6 | 586 | 67 | ||
XM_011520109.1 | XP_011518411.1 | 3 - 4 | 341 | 67 | ||
XM_017017738.1 | XP_016873227.1 | 3 - 4 | 642 | 67 | ||
XM_017017739.1 | XP_016873228.1 | 3 - 4 | 638 | 67 | ||
XM_017017740.1 | XP_016873229.1 | 4 - 5 | 480 | 67 | ||
XM_017017741.1 | XP_016873230.1 | 3 - 4 | 341 | 67 | ||
XM_017017742.1 | XP_016873231.1 | 3 - 4 | 341 | 67 | ||
XM_017017743.1 | XP_016873232.1 | 3 - 4 | 341 | 67 | ||
GenBank mRNA | AB000812.1 | - | 2 - 3 | 174 | 67 | |
AF044288.1 | - | 5 - 6 | 503 | 67 | ||
AK291510.1 | - | 4 - 5 | 451 | 67 | ||
AK315174.1 | - | 5 - 6 | 531 | 67 | ||
BC041129.1 | - | 5 - 6 | 517 | 5753529 | 67 | |
U60415.1 | - | 4 - 5 | 437 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
