| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03000775.hg.1 |
TC03000040.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03000775.hg.1 |
ENST00000383831.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03000775.hg.1 |
ENST00000383832.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03000775.hg.1 |
NM_153635.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300145106.hg.1 |
ENST00000273027.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300145106.hg.1 |
ENST00000383831.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300145106.hg.1 |
ENST00000383832.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300145106.hg.1 |
ENST00000613455.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300145106.hg.1 |
NM_001308388.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300145106.hg.1 |
NM_153635.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300145106.hg.1 |
ENST00000273027.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300145106.hg.1 |
ENST00000383831.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300145106.hg.1 |
ENST00000383832.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300145106.hg.1 |
ENST00000613455.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300145106.hg.1 |
NM_001308388.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300145106.hg.1 |
NM_153635.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300145106.hg.1 |
TC0300006554.hg.1 |
Clariom D TC |