Assay Details
| Gene Symbol: UFC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016406.3 | NP_057490.2 | 2 - 3 | 444 | 64 | |
| XM_005245254.1 | XP_005245311.1 | 2 - 3 | 396 | 64 | ||
| XM_017001450.1 | XP_016856939.1 | 3 - 4 | 289 | 64 | ||
| XM_017001451.1 | XP_016856940.1 | 3 - 4 | 553 | 64 | ||
| GenBank mRNA | AB154405.1 | - | 2 - 3 | 190 | 64 | |
| AF151884.1 | - | 2 - 3 | 411 | 64 | ||
| AF161504.1 | - | 2 - 3 | 430 | 64 | ||
| AK292776.1 | - | 2 - 3 | 396 | 64 | ||
| BC005187.1 | - | 2 - 3 | 436 | 3961569 | 64 | |
| CF128121.1 | - | 2 - 3 | 443 | 64 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01001378.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0100016161.hg.1 | ENST00000420498.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016161.hg.1 | ENST00000420498.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01021671.hg.1 | TC01001378.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01021671.hg.1 | ENST00000368003.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01021671.hg.1 | ENST00000473766.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01021671.hg.1 | NM_016406.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01021669.hg.1 | TC01001378.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01021669.hg.1 | ENST00000368003.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01021669.hg.1 | ENST00000473766.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01021669.hg.1 | NM_016406.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01021674.hg.1 | TC01001378.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01021674.hg.1 | ENST00000368003.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01021674.hg.1 | ENST00000473766.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01021674.hg.1 | NM_016406.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010334.hg.1 | ENST00000368003.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010334.hg.1 | ENST00000368003.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010334.hg.1 | ENST00000368003.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010334.hg.1 | ENST00000368003.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010334.hg.1 | ENST00000368003.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010334.hg.1 | ENST00000368003.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010334.hg.1 | ENST00000368003.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010334.hg.1 | ENST00000368003.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010334.hg.1 | ENST00000368003.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010334.hg.1 | ENST00000368003.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010334.hg.1 | ENST00000368003.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010334.hg.1 | ENST00000463735.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010334.hg.1 | ENST00000463735.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010334.hg.1 | ENST00000463735.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010334.hg.1 | ENST00000463735.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010334.hg.1 | ENST00000463735.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010334.hg.1 | ENST00000467540.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010334.hg.1 | ENST00000467540.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010334.hg.1 | ENST00000467540.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010334.hg.1 | ENST00000467540.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010334.hg.1 | ENST00000467540.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010334.hg.1 | ENST00000467540.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010334.hg.1 | ENST00000467540.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010334.hg.1 | ENST00000467540.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010334.hg.1 | ENST00000467540.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010334.hg.1 | ENST00000473766.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010334.hg.1 | ENST00000473766.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010334.hg.1 | ENST00000473766.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010334.hg.1 | ENST00000473766.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010334.hg.1 | ENST00000473766.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010334.hg.1 | ENST00000473766.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010334.hg.1 | ENST00000473766.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010334.hg.1 | ENST00000473766.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010334.hg.1 | ENST00000482672.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010334.hg.1 | ENST00000482672.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010334.hg.1 | ENST00000482672.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010334.hg.1 | ENST00000482672.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010334.hg.1 | ENST00000482672.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010334.hg.1 | ENST00000483191.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010334.hg.1 | ENST00000483191.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010334.hg.1 | ENST00000483191.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010334.hg.1 | ENST00000483191.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010334.hg.1 | ENST00000483191.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010334.hg.1 | ENST00000483191.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010334.hg.1 | ENST00000483191.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010334.hg.1 | NM_016406.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010334.hg.1 | NM_016406.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010334.hg.1 | NM_016406.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010334.hg.1 | NM_016406.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010334.hg.1 | NM_016406.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010334.hg.1 | NM_016406.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010334.hg.1 | NM_016406.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010334.hg.1 | NM_016406.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010334.hg.1 | NM_016406.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010334.hg.1 | NM_016406.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010334.hg.1 | NM_016406.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008160.hg.1 | ENST00000442074.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01021666.hg.1 | TC01001378.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01021666.hg.1 | ENST00000368003.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01021666.hg.1 | NM_016406.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01021672.hg.1 | TC01001378.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01021672.hg.1 | ENST00000368003.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01021672.hg.1 | ENST00000473766.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01021672.hg.1 | NM_016406.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01021665.hg.1 | TC01001378.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01021665.hg.1 | ENST00000368003.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01021665.hg.1 | NM_016406.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010334.hg.1 | ENST00000368003.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010334.hg.1 | ENST00000368003.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010334.hg.1 | ENST00000368003.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010334.hg.1 | ENST00000368003.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010334.hg.1 | ENST00000368003.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010334.hg.1 | ENST00000368003.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010334.hg.1 | ENST00000368003.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010334.hg.1 | ENST00000368003.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010334.hg.1 | ENST00000368003.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010334.hg.1 | ENST00000368003.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010334.hg.1 | ENST00000368003.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010334.hg.1 | ENST00000463735.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010334.hg.1 | ENST00000463735.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010334.hg.1 | ENST00000463735.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010334.hg.1 | ENST00000463735.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010334.hg.1 | ENST00000463735.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010334.hg.1 | ENST00000467540.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010334.hg.1 | ENST00000467540.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010334.hg.1 | ENST00000467540.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010334.hg.1 | ENST00000467540.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010334.hg.1 | ENST00000467540.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010334.hg.1 | ENST00000467540.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010334.hg.1 | ENST00000467540.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010334.hg.1 | ENST00000467540.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010334.hg.1 | ENST00000467540.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010334.hg.1 | ENST00000473766.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010334.hg.1 | ENST00000473766.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010334.hg.1 | ENST00000473766.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010334.hg.1 | ENST00000473766.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010334.hg.1 | ENST00000473766.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010334.hg.1 | ENST00000473766.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010334.hg.1 | ENST00000473766.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010334.hg.1 | ENST00000473766.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010334.hg.1 | ENST00000482672.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010334.hg.1 | ENST00000482672.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010334.hg.1 | ENST00000482672.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010334.hg.1 | ENST00000482672.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010334.hg.1 | ENST00000482672.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010334.hg.1 | ENST00000483191.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010334.hg.1 | ENST00000483191.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010334.hg.1 | ENST00000483191.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010334.hg.1 | ENST00000483191.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010334.hg.1 | ENST00000483191.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010334.hg.1 | ENST00000483191.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010334.hg.1 | ENST00000483191.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010334.hg.1 | NM_016406.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010334.hg.1 | NM_016406.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010334.hg.1 | NM_016406.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010334.hg.1 | NM_016406.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010334.hg.1 | NM_016406.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010334.hg.1 | NM_016406.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010334.hg.1 | NM_016406.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010334.hg.1 | NM_016406.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010334.hg.1 | NM_016406.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010334.hg.1 | NM_016406.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010334.hg.1 | NM_016406.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016161.hg.1 | ENST00000420498.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016161.hg.1 | ENST00000420498.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100175894.hg.1 | ENST00000368003.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100175894.hg.1 | ENST00000467540.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100175894.hg.1 | ENST00000473766.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100175894.hg.1 | ENST00000482672.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100175894.hg.1 | ENST00000483191.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100175894.hg.1 | NM_016406.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100175894.hg.1 | ENST00000368003.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100175894.hg.1 | ENST00000467540.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100175894.hg.1 | ENST00000473766.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100175894.hg.1 | ENST00000482672.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100175894.hg.1 | ENST00000483191.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100175894.hg.1 | NM_016406.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100175894.hg.1 | TC0100010334.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100175889.hg.1 | ENST00000368003.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100175889.hg.1 | NM_016406.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100175889.hg.1 | ENST00000368003.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100175889.hg.1 | NM_016406.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100175889.hg.1 | TC0100010334.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0700008160.hg.1 | ENST00000442074.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100175890.hg.1 | ENST00000368003.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100175890.hg.1 | ENST00000467540.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100175890.hg.1 | NM_016406.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100175890.hg.1 | ENST00000368003.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100175890.hg.1 | ENST00000467540.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100175890.hg.1 | NM_016406.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100175890.hg.1 | TC0100010334.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100175896.hg.1 | ENST00000368003.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100175896.hg.1 | ENST00000463735.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100175896.hg.1 | ENST00000467540.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100175896.hg.1 | ENST00000473766.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100175896.hg.1 | ENST00000483191.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100175896.hg.1 | NM_016406.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100175896.hg.1 | ENST00000368003.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100175896.hg.1 | ENST00000463735.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100175896.hg.1 | ENST00000467540.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100175896.hg.1 | ENST00000473766.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100175896.hg.1 | ENST00000483191.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100175896.hg.1 | NM_016406.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100175896.hg.1 | TC0100010334.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100175897.hg.1 | ENST00000368003.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100175897.hg.1 | ENST00000463735.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100175897.hg.1 | ENST00000467540.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100175897.hg.1 | ENST00000473766.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100175897.hg.1 | ENST00000482672.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100175897.hg.1 | ENST00000483191.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100175897.hg.1 | NM_016406.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100175897.hg.1 | ENST00000368003.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100175897.hg.1 | ENST00000463735.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100175897.hg.1 | ENST00000467540.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100175897.hg.1 | ENST00000473766.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100175897.hg.1 | ENST00000482672.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100175897.hg.1 | ENST00000483191.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100175897.hg.1 | NM_016406.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100175897.hg.1 | TC0100010334.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100175899.hg.1 | ENST00000368003.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100175899.hg.1 | ENST00000463735.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100175899.hg.1 | ENST00000467540.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100175899.hg.1 | ENST00000473766.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100175899.hg.1 | ENST00000482672.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100175899.hg.1 | ENST00000483191.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100175899.hg.1 | NM_016406.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100175899.hg.1 | ENST00000368003.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100175899.hg.1 | ENST00000463735.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100175899.hg.1 | ENST00000467540.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100175899.hg.1 | ENST00000473766.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100175899.hg.1 | ENST00000482672.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100175899.hg.1 | ENST00000483191.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100175899.hg.1 | NM_016406.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100175899.hg.1 | TC0100010334.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 186251 | - | - |

UFC1
Genomic Map