Assay Details
| Gene Symbol: TOB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_005749.3 | NP_005740.1 | 1 - 2 | 296 | 117 | |
| GenBank mRNA | BC070493.1 | - | 1 - 2 | 286 | 6470511 | 117 |
| BC098415.1 | - | 1 - 2 | 287 | 6480318 | 117 | |
| DA039934.1 | - | 1 - 2 | 296 | 117 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC17001698.hg.1
Additional Information:
This assay is in the 5'UTR region of the BC070493.1,BC098415.1,NM_005749.3 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
SMAD protein import into nucleus
negative regulation of cell proliferation
positive regulation of signal transduction
negative regulation of translation
negative regulation of BMP signaling pathway
negative regulation of osteoblast differentiation
negative regulation of nuclear-transcribed mRNA poly(A) tail shortening
positive regulation of nuclear-transcribed mRNA poly(A) tail shortening
positive regulation of nuclear-transcribed mRNA catabolic process, deadenylation-dependent decay
negative regulation of nucleic acid-templated transcription
transcription corepressor activity
SH3/SH2 adaptor activity
protein binding
receptor tyrosine kinase binding
SMAD binding
SMAD protein import into nucleus
negative regulation of cell proliferation
positive regulation of signal transduction
negative regulation of translation
negative regulation of BMP signaling pathway
negative regulation of osteoblast differentiation
negative regulation of nuclear-transcribed mRNA poly(A) tail shortening
positive regulation of nuclear-transcribed mRNA poly(A) tail shortening
positive regulation of nuclear-transcribed mRNA catabolic process, deadenylation-dependent decay
negative regulation of nucleic acid-templated transcription
transcription corepressor activity
SH3/SH2 adaptor activity
protein binding
receptor tyrosine kinase binding
SMAD binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR17022786.hg.1 | ENST00000268957.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17022786.hg.1 | ENST00000499247.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17022786.hg.1 | ENST00000509385.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17022786.hg.1 | NM_001243877.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17022786.hg.1 | NM_001243885.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17022786.hg.1 | NM_005749.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17022786.hg.1 | TC17001698.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR17022789.hg.1 | ENST00000268957.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17022789.hg.1 | ENST00000499247.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17022789.hg.1 | NM_001243877.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17022789.hg.1 | NM_005749.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17022789.hg.1 | TC17001698.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR17022792.hg.1 | ENST00000268957.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17022792.hg.1 | ENST00000499247.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17022792.hg.1 | NM_001243877.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17022792.hg.1 | NM_005749.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17022792.hg.1 | TC17001698.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR17022787.hg.1 | ENST00000268957.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17022787.hg.1 | ENST00000499247.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17022787.hg.1 | NM_001243877.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17022787.hg.1 | NM_005749.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17022787.hg.1 | TC17001698.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR17022793.hg.1 | ENST00000499247.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17022793.hg.1 | ENST00000509385.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17022793.hg.1 | NM_001243885.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17022793.hg.1 | NM_005749.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17022793.hg.1 | TC17001698.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011121.hg.1 | ENST00000268957.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011121.hg.1 | ENST00000268957.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011121.hg.1 | ENST00000268957.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011121.hg.1 | ENST00000268957.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011121.hg.1 | ENST00000268957.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011121.hg.1 | ENST00000268957.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011121.hg.1 | ENST00000499247.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011121.hg.1 | ENST00000499247.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011121.hg.1 | ENST00000499247.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011121.hg.1 | ENST00000499247.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011121.hg.1 | ENST00000499247.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011121.hg.1 | ENST00000499247.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011121.hg.1 | ENST00000509385.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011121.hg.1 | ENST00000509385.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011121.hg.1 | NM_001243877.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011121.hg.1 | NM_001243877.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011121.hg.1 | NM_001243877.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011121.hg.1 | NM_001243877.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011121.hg.1 | NM_001243877.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011121.hg.1 | NM_001243877.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011121.hg.1 | NM_001243877.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011121.hg.1 | NM_001243885.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011121.hg.1 | NM_001243885.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011121.hg.1 | NM_001243885.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011121.hg.1 | NM_001243885.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011121.hg.1 | NM_005749.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011121.hg.1 | NM_005749.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011121.hg.1 | NM_005749.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011121.hg.1 | NM_005749.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011121.hg.1 | NM_005749.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011121.hg.1 | NM_005749.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011121.hg.1 | ENST00000268957.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011121.hg.1 | ENST00000268957.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011121.hg.1 | ENST00000268957.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011121.hg.1 | ENST00000268957.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011121.hg.1 | ENST00000268957.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011121.hg.1 | ENST00000268957.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011121.hg.1 | ENST00000499247.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011121.hg.1 | ENST00000499247.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011121.hg.1 | ENST00000499247.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011121.hg.1 | ENST00000499247.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011121.hg.1 | ENST00000499247.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011121.hg.1 | ENST00000499247.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011121.hg.1 | ENST00000509385.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011121.hg.1 | ENST00000509385.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011121.hg.1 | NM_001243877.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011121.hg.1 | NM_001243877.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011121.hg.1 | NM_001243877.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011121.hg.1 | NM_001243877.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011121.hg.1 | NM_001243877.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011121.hg.1 | NM_001243877.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011121.hg.1 | NM_001243877.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011121.hg.1 | NM_001243885.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011121.hg.1 | NM_001243885.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011121.hg.1 | NM_001243885.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011121.hg.1 | NM_001243885.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011121.hg.1 | NM_005749.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011121.hg.1 | NM_005749.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011121.hg.1 | NM_005749.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011121.hg.1 | NM_005749.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011121.hg.1 | NM_005749.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011121.hg.1 | NM_005749.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700189503.hg.1 | ENST00000268957.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700189503.hg.1 | ENST00000499247.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700189503.hg.1 | NM_001243877.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700189503.hg.1 | NM_005749.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700189503.hg.1 | ENST00000268957.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700189503.hg.1 | ENST00000499247.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700189503.hg.1 | NM_001243877.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700189503.hg.1 | NM_005749.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700189503.hg.1 | TC1700011121.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700189506.hg.1 | ENST00000499247.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700189506.hg.1 | ENST00000509385.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700189506.hg.1 | NM_001243885.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700189506.hg.1 | NM_005749.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700189506.hg.1 | ENST00000499247.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700189506.hg.1 | ENST00000509385.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700189506.hg.1 | NM_001243885.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700189506.hg.1 | NM_005749.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700189506.hg.1 | TC1700011121.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700189502.hg.1 | ENST00000268957.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700189502.hg.1 | ENST00000499247.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700189502.hg.1 | ENST00000509385.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700189502.hg.1 | NM_001243877.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700189502.hg.1 | NM_001243885.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700189502.hg.1 | NM_005749.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700189502.hg.1 | ENST00000268957.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700189502.hg.1 | ENST00000499247.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700189502.hg.1 | ENST00000509385.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700189502.hg.1 | NM_001243877.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700189502.hg.1 | NM_001243885.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700189502.hg.1 | NM_005749.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700189502.hg.1 | TC1700011121.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700189505.hg.1 | ENST00000268957.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700189505.hg.1 | ENST00000499247.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700189505.hg.1 | NM_001243877.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700189505.hg.1 | NM_005749.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700189505.hg.1 | ENST00000268957.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700189505.hg.1 | ENST00000499247.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700189505.hg.1 | NM_001243877.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700189505.hg.1 | NM_005749.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700189505.hg.1 | TC1700011121.hg.1 | Clariom D TC |

TOB1
Genomic Map