Assay Details
Gene Symbol: BTNL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001304561.1 | NP_001291490.1 | 3 - 4 | 717 | 82 | |
XM_017011057.1 | XP_016866546.1 | 3 - 4 | 2466 | 82 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
