Assay Details
| Gene Symbol: TRMT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017026947.1 | XP_016882436.1 | 7 - 8 | 1318 | 69 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017026947.1 transcripts.
Set Membership: |
|

TRMT1
Genomic Map