Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR19015512.hg.1 |
ENST00000301458.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19015512.hg.1 |
ENST00000323539.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19015512.hg.1 |
ENST00000537716.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19015512.hg.1 |
NM_001165895.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19015512.hg.1 |
NM_016579.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19015512.hg.1 |
TC19001127.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011856.hg.1 |
ENST00000301458.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011856.hg.1 |
ENST00000301458.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011856.hg.1 |
ENST00000301458.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011856.hg.1 |
ENST00000301458.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011856.hg.1 |
ENST00000301458.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011856.hg.1 |
ENST00000301458.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011856.hg.1 |
ENST00000301458.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011856.hg.1 |
ENST00000301458.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011856.hg.1 |
ENST00000537716.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011856.hg.1 |
ENST00000537716.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011856.hg.1 |
ENST00000537716.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011856.hg.1 |
ENST00000537716.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011856.hg.1 |
ENST00000537716.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011856.hg.1 |
ENST00000537716.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011856.hg.1 |
ENST00000537716.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011856.hg.1 |
ENST00000596002.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011856.hg.1 |
ENST00000596002.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011856.hg.1 |
ENST00000596002.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011856.hg.1 |
ENST00000596002.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011856.hg.1 |
ENST00000596002.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011856.hg.1 |
ENST00000596002.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011856.hg.1 |
ENST00000596002.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011856.hg.1 |
ENST00000596002.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011856.hg.1 |
ENST00000596002.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011856.hg.1 |
ENST00000596246.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011856.hg.1 |
ENST00000596246.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011856.hg.1 |
ENST00000596246.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011856.hg.1 |
ENST00000598299.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011856.hg.1 |
ENST00000598299.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011856.hg.1 |
ENST00000598299.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011856.hg.1 |
ENST00000599573.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011856.hg.1 |
ENST00000599573.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011856.hg.1 |
ENST00000599573.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011856.hg.1 |
ENST00000599573.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011856.hg.1 |
ENST00000599573.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011856.hg.1 |
ENST00000599573.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011856.hg.1 |
NM_001165895.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011856.hg.1 |
NM_001165895.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011856.hg.1 |
NM_001165895.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011856.hg.1 |
NM_001165895.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011856.hg.1 |
NM_001165895.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011856.hg.1 |
NM_001165895.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011856.hg.1 |
NM_001165895.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011856.hg.1 |
NM_016579.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011856.hg.1 |
NM_016579.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011856.hg.1 |
NM_016579.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011856.hg.1 |
NM_016579.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011856.hg.1 |
NM_016579.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011856.hg.1 |
NM_016579.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011856.hg.1 |
NM_016579.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011856.hg.1 |
NM_016579.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900203857.hg.1 |
ENST00000301458.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900203857.hg.1 |
ENST00000537716.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900203857.hg.1 |
ENST00000596002.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900203857.hg.1 |
ENST00000599573.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900203857.hg.1 |
NM_001165895.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900203857.hg.1 |
NM_016579.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900203857.hg.1 |
TC1900011856.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900203857.hg.1 |
ENST00000301458.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900203857.hg.1 |
ENST00000537716.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900203857.hg.1 |
ENST00000596002.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900203857.hg.1 |
ENST00000599573.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900203857.hg.1 |
NM_001165895.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900203857.hg.1 |
NM_016579.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011856.hg.1 |
ENST00000301458.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011856.hg.1 |
ENST00000301458.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011856.hg.1 |
ENST00000301458.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011856.hg.1 |
ENST00000301458.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011856.hg.1 |
ENST00000301458.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011856.hg.1 |
ENST00000301458.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011856.hg.1 |
ENST00000301458.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011856.hg.1 |
ENST00000301458.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011856.hg.1 |
ENST00000537716.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011856.hg.1 |
ENST00000537716.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011856.hg.1 |
ENST00000537716.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011856.hg.1 |
ENST00000537716.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011856.hg.1 |
ENST00000537716.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011856.hg.1 |
ENST00000537716.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011856.hg.1 |
ENST00000537716.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011856.hg.1 |
ENST00000596002.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011856.hg.1 |
ENST00000596002.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011856.hg.1 |
ENST00000596002.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011856.hg.1 |
ENST00000596002.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011856.hg.1 |
ENST00000596002.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011856.hg.1 |
ENST00000596002.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011856.hg.1 |
ENST00000596002.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011856.hg.1 |
ENST00000596002.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011856.hg.1 |
ENST00000596002.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011856.hg.1 |
ENST00000596246.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011856.hg.1 |
ENST00000596246.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011856.hg.1 |
ENST00000596246.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011856.hg.1 |
ENST00000598299.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011856.hg.1 |
ENST00000598299.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011856.hg.1 |
ENST00000598299.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011856.hg.1 |
ENST00000599573.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011856.hg.1 |
ENST00000599573.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011856.hg.1 |
ENST00000599573.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011856.hg.1 |
ENST00000599573.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011856.hg.1 |
ENST00000599573.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011856.hg.1 |
ENST00000599573.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011856.hg.1 |
NM_001165895.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011856.hg.1 |
NM_001165895.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011856.hg.1 |
NM_001165895.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011856.hg.1 |
NM_001165895.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011856.hg.1 |
NM_001165895.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011856.hg.1 |
NM_001165895.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011856.hg.1 |
NM_001165895.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011856.hg.1 |
NM_016579.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011856.hg.1 |
NM_016579.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011856.hg.1 |
NM_016579.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011856.hg.1 |
NM_016579.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011856.hg.1 |
NM_016579.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011856.hg.1 |
NM_016579.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011856.hg.1 |
NM_016579.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011856.hg.1 |
NM_016579.3 |
Refseq |