Assay Details
Gene Symbol: DDX18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_006773.3 | NP_006764.3 | 1 - 2 | 181 | 131 | |
GenBank mRNA | AK001467.1 | - | 1 - 2 | 153 | 131 | |
BC001238.1 | - | 1 - 2 | 110 | 3452935 | 131 | |
BC003360.1 | - | 1 - 2 | 140 | 2900252 | 131 | |
BC024739.1 | - | 1 - 2 | 155 | 4995871 | 131 | |
BU599536.1 | - | 1 - 2 | 199 | 131 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC02000742.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR02011558.hg.1 | TC02000742.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR02011558.hg.1 | ENST00000263239.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02011558.hg.1 | NM_006773.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600010968.hg.1 | ENST00000404856.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008174.hg.1 | ENST00000511690.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009628.hg.1 | ENST00000558620.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014747.hg.1 | ENST00000417893.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02011555.hg.1 | TC02000742.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR02011555.hg.1 | ENST00000263239.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02011555.hg.1 | NM_006773.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | ENST00000263239.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | ENST00000263239.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | ENST00000263239.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | ENST00000263239.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | ENST00000263239.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | ENST00000263239.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | ENST00000263239.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | ENST00000263239.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | ENST00000263239.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | ENST00000263239.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | ENST00000263239.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | ENST00000263239.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | ENST00000263239.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | ENST00000263239.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | ENST00000263239.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | ENST00000263239.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | ENST00000263239.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | ENST00000263239.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | ENST00000263239.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | ENST00000263239.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | ENST00000263239.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | ENST00000263239.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | ENST00000263239.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | ENST00000415038.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | ENST00000415038.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | ENST00000415038.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | ENST00000415038.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | ENST00000415038.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | ENST00000415038.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | ENST00000461443.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | ENST00000461443.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | ENST00000461443.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | ENST00000474694.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | ENST00000474694.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | ENST00000474694.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | ENST00000474694.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | ENST00000474694.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | ENST00000474694.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | ENST00000474694.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | ENST00000474694.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | ENST00000474694.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | ENST00000474694.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | ENST00000476149.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | ENST00000476149.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | ENST00000476149.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | ENST00000476149.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | ENST00000476149.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | ENST00000476149.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | ENST00000476149.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | ENST00000476149.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | ENST00000476149.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | ENST00000476149.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | ENST00000476149.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | ENST00000476149.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | ENST00000476149.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | ENST00000476149.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | ENST00000489933.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | ENST00000489933.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | ENST00000489933.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | NM_006773.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | NM_006773.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | NM_006773.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | NM_006773.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | NM_006773.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | NM_006773.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | NM_006773.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | NM_006773.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | NM_006773.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | NM_006773.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | NM_006773.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | NM_006773.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | NM_006773.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | NM_006773.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | NM_006773.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | NM_006773.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | NM_006773.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | NM_006773.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | NM_006773.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | NM_006773.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | NM_006773.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | NM_006773.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400007545.hg.1 | ENST00000556558.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008429.hg.1 | ENST00000450119.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02011561.hg.1 | TC02000742.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR02011561.hg.1 | ENST00000263239.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02011561.hg.1 | ENST00000474694.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02011561.hg.1 | NM_006773.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200163211.hg.1 | ENST00000263239.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200163211.hg.1 | NM_006773.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200163211.hg.1 | ENST00000263239.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200163211.hg.1 | NM_006773.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200163211.hg.1 | TC0200009041.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0200014747.hg.1 | ENST00000417893.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200163207.hg.1 | ENST00000263239.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200163207.hg.1 | NM_006773.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200163207.hg.1 | ENST00000263239.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200163207.hg.1 | NM_006773.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200163207.hg.1 | TC0200009041.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0200163214.hg.1 | ENST00000263239.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200163214.hg.1 | ENST00000474694.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200163214.hg.1 | NM_006773.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200163214.hg.1 | ENST00000263239.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200163214.hg.1 | ENST00000474694.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200163214.hg.1 | NM_006773.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200163214.hg.1 | TC0200009041.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | ENST00000263239.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | ENST00000263239.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | ENST00000263239.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | ENST00000263239.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | ENST00000263239.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | ENST00000263239.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | ENST00000263239.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | ENST00000263239.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | ENST00000263239.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | ENST00000263239.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | ENST00000263239.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | ENST00000263239.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | ENST00000263239.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | ENST00000263239.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | ENST00000263239.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | ENST00000263239.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | ENST00000263239.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | ENST00000263239.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | ENST00000263239.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | ENST00000263239.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | ENST00000263239.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | ENST00000263239.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | ENST00000263239.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | ENST00000415038.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | ENST00000415038.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | ENST00000415038.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | ENST00000415038.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | ENST00000415038.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | ENST00000415038.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | ENST00000461443.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | ENST00000461443.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | ENST00000461443.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | ENST00000474694.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | ENST00000474694.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | ENST00000474694.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | ENST00000474694.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | ENST00000474694.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | ENST00000474694.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | ENST00000474694.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | ENST00000474694.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | ENST00000474694.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | ENST00000474694.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | ENST00000476149.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | ENST00000476149.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | ENST00000476149.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | ENST00000476149.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | ENST00000476149.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | ENST00000476149.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | ENST00000476149.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | ENST00000476149.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | ENST00000476149.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | ENST00000476149.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | ENST00000476149.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | ENST00000476149.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | ENST00000476149.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | ENST00000476149.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | ENST00000489933.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | ENST00000489933.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | ENST00000489933.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | NM_006773.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | NM_006773.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | NM_006773.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | NM_006773.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | NM_006773.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | NM_006773.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | NM_006773.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | NM_006773.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | NM_006773.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | NM_006773.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | NM_006773.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | NM_006773.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | NM_006773.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | NM_006773.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | NM_006773.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | NM_006773.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | NM_006773.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | NM_006773.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | NM_006773.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | NM_006773.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | NM_006773.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | NM_006773.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400007545.hg.1 | ENST00000556558.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200163209.hg.1 | ENST00000263239.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200163209.hg.1 | NM_006773.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200163209.hg.1 | ENST00000263239.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200163209.hg.1 | NM_006773.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200163209.hg.1 | TC0200009041.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0200163210.hg.1 | ENST00000263239.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200163210.hg.1 | NM_006773.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200163210.hg.1 | ENST00000263239.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200163210.hg.1 | NM_006773.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200163210.hg.1 | TC0200009041.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0500008174.hg.1 | ENST00000511690.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010968.hg.1 | ENST00000404856.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008429.hg.1 | ENST00000450119.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009628.hg.1 | ENST00000558620.2 | Ensembl |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 211972 | - | - |
