Assay Details
Gene Symbol: DDX18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_006773.3 | NP_006764.3 | 2 - 3 | 466 | 130 | |
GenBank mRNA | AK001467.1 | - | 2 - 3 | 438 | 130 | |
BC001238.1 | - | 2 - 3 | 395 | 3452935 | 130 | |
BC003360.1 | - | 2 - 3 | 425 | 2900252 | 130 | |
BC024739.1 | - | 2 - 3 | 440 | 4995871 | 130 | |
CR457060.1 | - | 1 - 2 | 187 | 130 | ||
X98743.1 | - | 1 - 2 | 204 | 130 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC02000742.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | ENST00000263239.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | ENST00000263239.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | ENST00000263239.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | ENST00000263239.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | ENST00000263239.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | ENST00000263239.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | ENST00000263239.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | ENST00000263239.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | ENST00000263239.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | ENST00000263239.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | ENST00000263239.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | ENST00000263239.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | ENST00000263239.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | ENST00000263239.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | ENST00000263239.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | ENST00000263239.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | ENST00000263239.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | ENST00000263239.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | ENST00000263239.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | ENST00000263239.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | ENST00000263239.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | ENST00000263239.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | ENST00000263239.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | ENST00000415038.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | ENST00000415038.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | ENST00000415038.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | ENST00000415038.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | ENST00000415038.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | ENST00000415038.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | ENST00000461443.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | ENST00000461443.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | ENST00000461443.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | ENST00000474694.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | ENST00000474694.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | ENST00000474694.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | ENST00000474694.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | ENST00000474694.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | ENST00000474694.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | ENST00000474694.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | ENST00000474694.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | ENST00000474694.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | ENST00000474694.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | ENST00000476149.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | ENST00000476149.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | ENST00000476149.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | ENST00000476149.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | ENST00000476149.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | ENST00000476149.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | ENST00000476149.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | ENST00000476149.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | ENST00000476149.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | ENST00000476149.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | ENST00000476149.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | ENST00000476149.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | ENST00000476149.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | ENST00000476149.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | ENST00000489933.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | ENST00000489933.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | ENST00000489933.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | NM_006773.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | NM_006773.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | NM_006773.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | NM_006773.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | NM_006773.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | NM_006773.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | NM_006773.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | NM_006773.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | NM_006773.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | NM_006773.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | NM_006773.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | NM_006773.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | NM_006773.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | NM_006773.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | NM_006773.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | NM_006773.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | NM_006773.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | NM_006773.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | NM_006773.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | NM_006773.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | NM_006773.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | NM_006773.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR02011562.hg.1 | TC02000742.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR02011562.hg.1 | ENST00000263239.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02011562.hg.1 | ENST00000474694.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02011562.hg.1 | NM_006773.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200163215.hg.1 | ENST00000263239.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200163215.hg.1 | ENST00000474694.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200163215.hg.1 | NM_006773.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200163215.hg.1 | ENST00000263239.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200163215.hg.1 | ENST00000474694.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200163215.hg.1 | NM_006773.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200163215.hg.1 | TC0200009041.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | ENST00000263239.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | ENST00000263239.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | ENST00000263239.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | ENST00000263239.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | ENST00000263239.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | ENST00000263239.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | ENST00000263239.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | ENST00000263239.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | ENST00000263239.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | ENST00000263239.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | ENST00000263239.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | ENST00000263239.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | ENST00000263239.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | ENST00000263239.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | ENST00000263239.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | ENST00000263239.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | ENST00000263239.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | ENST00000263239.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | ENST00000263239.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | ENST00000263239.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | ENST00000263239.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | ENST00000263239.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | ENST00000263239.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | ENST00000415038.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | ENST00000415038.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | ENST00000415038.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | ENST00000415038.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | ENST00000415038.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | ENST00000415038.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | ENST00000461443.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | ENST00000461443.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | ENST00000461443.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | ENST00000474694.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | ENST00000474694.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | ENST00000474694.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | ENST00000474694.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | ENST00000474694.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | ENST00000474694.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | ENST00000474694.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | ENST00000474694.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | ENST00000474694.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | ENST00000474694.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | ENST00000476149.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | ENST00000476149.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | ENST00000476149.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | ENST00000476149.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | ENST00000476149.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | ENST00000476149.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | ENST00000476149.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | ENST00000476149.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | ENST00000476149.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | ENST00000476149.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | ENST00000476149.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | ENST00000476149.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | ENST00000476149.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | ENST00000476149.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | ENST00000489933.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | ENST00000489933.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | ENST00000489933.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | NM_006773.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | NM_006773.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | NM_006773.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | NM_006773.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | NM_006773.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | NM_006773.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | NM_006773.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | NM_006773.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | NM_006773.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | NM_006773.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | NM_006773.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | NM_006773.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | NM_006773.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | NM_006773.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | NM_006773.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | NM_006773.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | NM_006773.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | NM_006773.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | NM_006773.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | NM_006773.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | NM_006773.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | NM_006773.3 | Refseq |
