Assay Details
| Gene Symbol: TNS3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AL137468.1 | - | 1 | 2053 | 139 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC730338 | LOC730338 | -682.4kb | Sense | Chr. 7 - 46687879 - 46697122 on NCBI Build GRCh38 | |
| Downstream | long intergenic non-protein coding RNA 1447 | LINC01447 | 240.2kb | Anti-sense | Chr. 7 - 47621939 - 47629893 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

TNS3
Genomic Map