Assay Details
Gene Symbol: ERG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243429.1 | NP_001230358.1 | 5 - 6 | 617 | 83 | |
NM_004449.4 | NP_004440.1 | 7 - 8 | 958 | 83 | ||
XM_017028288.1 | XP_016883777.1 | 5 - 6 | 743 | 83 | ||
GenBank mRNA | KP100545.1 | - | 5 - 6 | 665 | 83 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
