Assay Details
| Gene Symbol: NCAM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_004540.3 | NP_004531.2 | 16 - 17 | 2533 | 89 | |
| XM_011529575.2 | XP_011527877.1 | 17 - 18 | 3008 | 89 | ||
| XM_011529576.2 | XP_011527878.1 | 17 - 18 | 2585 | 89 | ||
| XM_011529579.2 | XP_011527881.1 | 16 - 17 | 2509 | 89 | ||
| XM_011529585.2 | XP_011527887.1 | 13 - 14 | 1875 | 89 | ||
| XM_017028353.1 | XP_016883842.1 | 17 - 18 | 2673 | 89 | ||
| XM_017028356.1 | XP_016883845.1 | 16 - 17 | 2408 | 89 | ||
| XM_017028357.1 | XP_016883846.1 | 16 - 17 | 2408 | 89 | ||
| GenBank mRNA | AK299054.1 | - | 13 - 14 | 2046 | 89 | |
| BC052946.1 | - | 16 - 17 | 2533 | 5315109 | 89 | |
| U75330.1 | - | 16 - 17 | 2353 | 89 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC21000060.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | ENST00000284894.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | ENST00000284894.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | ENST00000284894.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | ENST00000284894.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | ENST00000284894.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | ENST00000284894.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | ENST00000284894.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | ENST00000284894.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | ENST00000284894.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | ENST00000284894.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | ENST00000284894.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | ENST00000284894.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | ENST00000284894.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | ENST00000284894.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | ENST00000284894.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | ENST00000284894.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | ENST00000284894.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | ENST00000284894.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | ENST00000284894.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | ENST00000284894.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | ENST00000284894.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | ENST00000400546.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | ENST00000400546.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | ENST00000400546.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | ENST00000400546.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | ENST00000400546.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | ENST00000400546.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | ENST00000400546.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | ENST00000400546.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | ENST00000400546.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | ENST00000400546.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | ENST00000400546.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | ENST00000400546.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | ENST00000400546.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | ENST00000400546.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | ENST00000400546.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | ENST00000400546.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | ENST00000400546.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | ENST00000400546.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | ENST00000400546.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | ENST00000400546.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | ENST00000400546.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | ENST00000400546.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | ENST00000400546.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | ENST00000400546.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | ENST00000461281.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | ENST00000461281.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | ENST00000461281.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | ENST00000461281.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | ENST00000461281.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | ENST00000461281.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | ENST00000484983.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | ENST00000484983.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | ENST00000484983.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | ENST00000486367.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | ENST00000486367.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | ENST00000486367.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | ENST00000486367.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | ENST00000486367.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | NM_004540.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | NM_004540.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | NM_004540.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | NM_004540.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | NM_004540.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | NM_004540.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | NM_004540.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | NM_004540.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | NM_004540.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | NM_004540.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | NM_004540.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | NM_004540.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | NM_004540.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | NM_004540.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | NM_004540.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | NM_004540.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | NM_004540.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | NM_004540.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | NM_004540.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | NM_004540.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | NM_004540.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | NM_004540.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | NM_004540.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR21000307.hg.1 | ENST00000284894.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR21000307.hg.1 | ENST00000400546.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR21000307.hg.1 | NM_004540.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR21000307.hg.1 | TC21000060.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | ENST00000284894.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | ENST00000284894.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | ENST00000284894.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | ENST00000284894.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | ENST00000284894.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | ENST00000284894.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | ENST00000284894.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | ENST00000284894.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | ENST00000284894.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | ENST00000284894.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | ENST00000284894.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | ENST00000284894.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | ENST00000284894.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | ENST00000284894.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | ENST00000284894.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | ENST00000284894.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | ENST00000284894.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | ENST00000284894.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | ENST00000284894.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | ENST00000284894.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | ENST00000284894.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | ENST00000400546.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | ENST00000400546.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | ENST00000400546.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | ENST00000400546.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | ENST00000400546.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | ENST00000400546.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | ENST00000400546.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | ENST00000400546.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | ENST00000400546.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | ENST00000400546.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | ENST00000400546.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | ENST00000400546.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | ENST00000400546.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | ENST00000400546.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | ENST00000400546.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | ENST00000400546.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | ENST00000400546.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | ENST00000400546.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | ENST00000400546.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | ENST00000400546.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | ENST00000400546.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | ENST00000400546.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | ENST00000400546.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | ENST00000400546.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | ENST00000461281.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | ENST00000461281.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | ENST00000461281.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | ENST00000461281.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | ENST00000461281.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | ENST00000461281.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | ENST00000484983.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | ENST00000484983.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | ENST00000484983.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | ENST00000486367.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | ENST00000486367.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | ENST00000486367.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | ENST00000486367.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | ENST00000486367.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | NM_004540.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | NM_004540.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | NM_004540.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | NM_004540.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | NM_004540.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | NM_004540.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | NM_004540.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | NM_004540.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | NM_004540.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | NM_004540.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | NM_004540.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | NM_004540.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | NM_004540.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | NM_004540.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | NM_004540.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | NM_004540.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | NM_004540.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | NM_004540.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | NM_004540.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | NM_004540.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | NM_004540.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | NM_004540.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | NM_004540.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100145422.hg.1 | ENST00000284894.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100145422.hg.1 | ENST00000400546.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100145422.hg.1 | NM_004540.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100145422.hg.1 | TC2100006714.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2100145422.hg.1 | ENST00000284894.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100145422.hg.1 | ENST00000400546.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100145422.hg.1 | NM_004540.4 | Refseq |

NCAM2
Genomic Map