Assay Details
| Gene Symbol: FTCD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001320412.1 | NP_001307341.1 | 14 - 15 | 1717 | 117 | |
| XM_011529436.2 | XP_011527738.1 | 14 - 15 | 2745 | 117 | ||
| GenBank mRNA | AF289022.1 | - | 14 - 15 | 1717 | 117 | |
| BC032037.1 | - | 4 - 5 | 1024 | 117 | ||
More Information
Set Membership: |
|

FTCD
Genomic Map