Assay Details
Gene Symbol: CPQ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_016134.3 | NP_057218.1 | 1 - 2 | 174 | 116 | |
XM_017012953.1 | XP_016868442.1 | 1 - 2 | 174 | 116 | ||
XM_017012954.1 | XP_016868443.1 | 1 - 2 | 174 | 116 | ||
XM_017012955.1 | XP_016868444.1 | 1 - 2 | 174 | 116 | ||
GenBank mRNA | AK075132.1 | - | 1 - 2 | 92 | 116 | |
AK296013.1 | - | 1 - 2 | 37 | 116 | ||
AK304008.1 | - | 1 - 2 | 91 | 116 | ||
AK315447.1 | - | 1 - 2 | 91 | 116 | ||
BC020689.1 | - | 1 - 2 | 130 | 4251802 | 116 | |
DA699663.1 | - | 1 - 2 | 174 | 116 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC08000591.hg
Additional Information:
This assay is in the 5'UTR region of the AK075132.1,AK296013.1,AK304008.1,AK315447.1,BC020689.1,NM_016134.3,XM_017012953.1,XM_017012954.1,XM_017012955.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR08008194.hg.1 | TC08000591.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR08008194.hg.1 | ENST00000220763.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR08008194.hg.1 | NM_016134.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR08008192.hg.1 | TC08000591.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR08008192.hg.1 | ENST00000220763.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR08008192.hg.1 | NM_016134.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR08008199.hg.1 | TC08000591.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR08008199.hg.1 | ENST00000220763.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR08008199.hg.1 | NM_016134.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008300.hg.1 | ENST00000220763.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008300.hg.1 | ENST00000220763.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008300.hg.1 | ENST00000220763.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008300.hg.1 | ENST00000220763.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008300.hg.1 | ENST00000220763.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008300.hg.1 | ENST00000220763.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008300.hg.1 | ENST00000220763.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008300.hg.1 | ENST00000220763.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008300.hg.1 | ENST00000220763.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008300.hg.1 | ENST00000220763.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008300.hg.1 | ENST00000220763.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008300.hg.1 | ENST00000220763.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008300.hg.1 | ENST00000220763.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008300.hg.1 | ENST00000220763.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008300.hg.1 | ENST00000220763.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008300.hg.1 | ENST00000517742.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008300.hg.1 | ENST00000517742.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008300.hg.1 | ENST00000517742.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008300.hg.1 | ENST00000517742.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008300.hg.1 | ENST00000517742.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008300.hg.1 | ENST00000517742.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008300.hg.1 | ENST00000519484.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008300.hg.1 | ENST00000519484.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008300.hg.1 | ENST00000519484.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008300.hg.1 | ENST00000519484.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008300.hg.1 | ENST00000519900.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008300.hg.1 | ENST00000519900.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008300.hg.1 | ENST00000519900.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008300.hg.1 | ENST00000519900.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008300.hg.1 | ENST00000519900.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008300.hg.1 | ENST00000521142.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008300.hg.1 | ENST00000521142.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008300.hg.1 | ENST00000521142.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008300.hg.1 | ENST00000521142.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008300.hg.1 | ENST00000522617.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008300.hg.1 | ENST00000522617.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008300.hg.1 | ENST00000522617.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008300.hg.1 | ENST00000522617.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008300.hg.1 | ENST00000525310.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008300.hg.1 | ENST00000525310.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008300.hg.1 | ENST00000525310.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008300.hg.1 | ENST00000525310.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008300.hg.1 | ENST00000529551.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008300.hg.1 | ENST00000529551.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008300.hg.1 | ENST00000529551.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008300.hg.1 | ENST00000529551.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008300.hg.1 | ENST00000532528.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008300.hg.1 | ENST00000532528.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008300.hg.1 | ENST00000532528.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008300.hg.1 | ENST00000532528.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008300.hg.1 | NM_016134.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008300.hg.1 | NM_016134.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008300.hg.1 | NM_016134.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008300.hg.1 | NM_016134.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008300.hg.1 | NM_016134.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008300.hg.1 | NM_016134.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008300.hg.1 | NM_016134.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008300.hg.1 | NM_016134.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008300.hg.1 | NM_016134.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008300.hg.1 | NM_016134.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008300.hg.1 | NM_016134.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008300.hg.1 | NM_016134.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008300.hg.1 | NM_016134.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008300.hg.1 | NM_016134.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008300.hg.1 | NM_016134.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800157189.hg.1 | ENST00000220763.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800157189.hg.1 | ENST00000517742.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800157189.hg.1 | ENST00000519484.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800157189.hg.1 | ENST00000519900.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800157189.hg.1 | ENST00000521142.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800157189.hg.1 | ENST00000525310.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800157189.hg.1 | NM_016134.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800157189.hg.1 | ENST00000220763.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800157189.hg.1 | ENST00000517742.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800157189.hg.1 | ENST00000519484.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800157189.hg.1 | ENST00000519900.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800157189.hg.1 | ENST00000521142.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800157189.hg.1 | ENST00000525310.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800157189.hg.1 | NM_016134.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800157189.hg.1 | TC0800008300.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0800157188.hg.1 | ENST00000220763.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800157188.hg.1 | ENST00000517742.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800157188.hg.1 | ENST00000519484.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800157188.hg.1 | ENST00000519900.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800157188.hg.1 | ENST00000521142.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800157188.hg.1 | ENST00000525310.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800157188.hg.1 | NM_016134.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800157188.hg.1 | ENST00000220763.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800157188.hg.1 | ENST00000517742.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800157188.hg.1 | ENST00000519484.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800157188.hg.1 | ENST00000519900.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800157188.hg.1 | ENST00000521142.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800157188.hg.1 | ENST00000525310.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800157188.hg.1 | NM_016134.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800157188.hg.1 | TC0800008300.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0800008300.hg.1 | ENST00000220763.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008300.hg.1 | ENST00000220763.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008300.hg.1 | ENST00000220763.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008300.hg.1 | ENST00000220763.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008300.hg.1 | ENST00000220763.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008300.hg.1 | ENST00000220763.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008300.hg.1 | ENST00000220763.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008300.hg.1 | ENST00000220763.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008300.hg.1 | ENST00000220763.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008300.hg.1 | ENST00000220763.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008300.hg.1 | ENST00000220763.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008300.hg.1 | ENST00000220763.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008300.hg.1 | ENST00000220763.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008300.hg.1 | ENST00000220763.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008300.hg.1 | ENST00000220763.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008300.hg.1 | ENST00000517742.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008300.hg.1 | ENST00000517742.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008300.hg.1 | ENST00000517742.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008300.hg.1 | ENST00000517742.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008300.hg.1 | ENST00000517742.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008300.hg.1 | ENST00000517742.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008300.hg.1 | ENST00000519484.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008300.hg.1 | ENST00000519484.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008300.hg.1 | ENST00000519484.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008300.hg.1 | ENST00000519484.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008300.hg.1 | ENST00000519900.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008300.hg.1 | ENST00000519900.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008300.hg.1 | ENST00000519900.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008300.hg.1 | ENST00000519900.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008300.hg.1 | ENST00000519900.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008300.hg.1 | ENST00000521142.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008300.hg.1 | ENST00000521142.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008300.hg.1 | ENST00000521142.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008300.hg.1 | ENST00000521142.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008300.hg.1 | ENST00000522617.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008300.hg.1 | ENST00000522617.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008300.hg.1 | ENST00000522617.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008300.hg.1 | ENST00000522617.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008300.hg.1 | ENST00000525310.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008300.hg.1 | ENST00000525310.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008300.hg.1 | ENST00000525310.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008300.hg.1 | ENST00000525310.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008300.hg.1 | ENST00000529551.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008300.hg.1 | ENST00000529551.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008300.hg.1 | ENST00000529551.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008300.hg.1 | ENST00000529551.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008300.hg.1 | ENST00000532528.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008300.hg.1 | ENST00000532528.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008300.hg.1 | ENST00000532528.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008300.hg.1 | ENST00000532528.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008300.hg.1 | NM_016134.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008300.hg.1 | NM_016134.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008300.hg.1 | NM_016134.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008300.hg.1 | NM_016134.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008300.hg.1 | NM_016134.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008300.hg.1 | NM_016134.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008300.hg.1 | NM_016134.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008300.hg.1 | NM_016134.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008300.hg.1 | NM_016134.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008300.hg.1 | NM_016134.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008300.hg.1 | NM_016134.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008300.hg.1 | NM_016134.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008300.hg.1 | NM_016134.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008300.hg.1 | NM_016134.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008300.hg.1 | NM_016134.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800157179.hg.1 | ENST00000220763.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800157179.hg.1 | NM_016134.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800157179.hg.1 | ENST00000220763.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800157179.hg.1 | NM_016134.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800157179.hg.1 | TC0800008300.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0800157180.hg.1 | ENST00000220763.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800157180.hg.1 | NM_016134.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800157180.hg.1 | ENST00000220763.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800157180.hg.1 | NM_016134.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800157180.hg.1 | TC0800008300.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0800157182.hg.1 | ENST00000220763.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800157182.hg.1 | ENST00000517742.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800157182.hg.1 | ENST00000519900.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800157182.hg.1 | NM_016134.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800157182.hg.1 | ENST00000220763.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800157182.hg.1 | ENST00000517742.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800157182.hg.1 | ENST00000519900.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800157182.hg.1 | NM_016134.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800157182.hg.1 | TC0800008300.hg.1 | Clariom D TC |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 211282 | - | - |
