Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR08008519.hg.1 |
TC08000612.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08008519.hg.1 |
ENST00000353107.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08008519.hg.1 |
ENST00000519765.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08008519.hg.1 |
ENST00000522439.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08008519.hg.1 |
NM_005034.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008370.hg.1 |
ENST00000353107.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008370.hg.1 |
ENST00000353107.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008370.hg.1 |
ENST00000353107.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008370.hg.1 |
ENST00000353107.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008370.hg.1 |
ENST00000353107.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008370.hg.1 |
ENST00000353107.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008370.hg.1 |
ENST00000353107.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008370.hg.1 |
ENST00000353107.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008370.hg.1 |
ENST00000519765.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008370.hg.1 |
ENST00000519765.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008370.hg.1 |
ENST00000519765.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008370.hg.1 |
ENST00000519765.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008370.hg.1 |
ENST00000519765.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008370.hg.1 |
ENST00000519765.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008370.hg.1 |
ENST00000519765.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008370.hg.1 |
ENST00000522439.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008370.hg.1 |
ENST00000522439.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008370.hg.1 |
ENST00000522439.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008370.hg.1 |
ENST00000522439.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008370.hg.1 |
ENST00000522439.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008370.hg.1 |
NM_005034.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008370.hg.1 |
NM_005034.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008370.hg.1 |
NM_005034.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008370.hg.1 |
NM_005034.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008370.hg.1 |
NM_005034.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008370.hg.1 |
NM_005034.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008370.hg.1 |
NM_005034.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800157643.hg.1 |
ENST00000353107.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800157643.hg.1 |
ENST00000519765.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800157643.hg.1 |
ENST00000522439.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800157643.hg.1 |
NM_005034.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800157643.hg.1 |
ENST00000353107.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800157643.hg.1 |
ENST00000519765.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800157643.hg.1 |
ENST00000522439.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800157643.hg.1 |
NM_005034.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800157643.hg.1 |
TC0800008370.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0800008370.hg.1 |
ENST00000353107.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008370.hg.1 |
ENST00000353107.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008370.hg.1 |
ENST00000353107.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008370.hg.1 |
ENST00000353107.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008370.hg.1 |
ENST00000353107.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008370.hg.1 |
ENST00000353107.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008370.hg.1 |
ENST00000353107.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008370.hg.1 |
ENST00000353107.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008370.hg.1 |
ENST00000519765.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008370.hg.1 |
ENST00000519765.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008370.hg.1 |
ENST00000519765.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008370.hg.1 |
ENST00000519765.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008370.hg.1 |
ENST00000519765.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008370.hg.1 |
ENST00000519765.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008370.hg.1 |
ENST00000519765.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008370.hg.1 |
ENST00000522439.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008370.hg.1 |
ENST00000522439.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008370.hg.1 |
ENST00000522439.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008370.hg.1 |
ENST00000522439.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008370.hg.1 |
ENST00000522439.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008370.hg.1 |
NM_005034.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008370.hg.1 |
NM_005034.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008370.hg.1 |
NM_005034.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008370.hg.1 |
NM_005034.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008370.hg.1 |
NM_005034.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008370.hg.1 |
NM_005034.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008370.hg.1 |
NM_005034.3 |
Refseq |