Assay Details
Gene Symbol: GNL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001323623.1 | NP_001310552.1 | 2 - 3 | 311 | 87 | |
NM_001323624.1 | NP_001310553.1 | 2 - 3 | 311 | 87 | ||
NM_013285.2 | NP_037417.1 | 2 - 3 | 311 | 87 | ||
GenBank mRNA | AK298358.1 | - | 2 - 3 | 265 | 87 | |
AK307352.1 | - | 2 - 3 | 265 | 87 | ||
AK312915.1 | - | 2 - 3 | 265 | 87 | ||
BC000107.2 | - | 2 - 3 | 247 | 3506131 | 87 | |
BC009250.2 | - | 2 - 3 | 247 | 2960125 | 87 | |
L05425.1 | - | 2 - 3 | 227 | 87 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC01002519.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK298358.1,NM_001323624.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0100013748.hg.1 | ENST00000373062.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013748.hg.1 | ENST00000373062.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013748.hg.1 | ENST00000373062.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013748.hg.1 | ENST00000373062.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013748.hg.1 | ENST00000373062.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013748.hg.1 | ENST00000373062.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013748.hg.1 | ENST00000373062.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013748.hg.1 | ENST00000373062.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013748.hg.1 | ENST00000373062.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013748.hg.1 | ENST00000373062.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013748.hg.1 | ENST00000373062.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013748.hg.1 | ENST00000373062.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013748.hg.1 | ENST00000373062.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013748.hg.1 | ENST00000373062.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013748.hg.1 | ENST00000373062.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013748.hg.1 | ENST00000373062.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013748.hg.1 | ENST00000373062.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013748.hg.1 | ENST00000373062.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013748.hg.1 | ENST00000373062.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013748.hg.1 | ENST00000373062.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013748.hg.1 | ENST00000373062.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013748.hg.1 | ENST00000462812.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013748.hg.1 | ENST00000462812.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013748.hg.1 | ENST00000462812.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013748.hg.1 | ENST00000462812.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013748.hg.1 | ENST00000462812.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013748.hg.1 | ENST00000462812.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013748.hg.1 | ENST00000463351.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013748.hg.1 | ENST00000463351.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013748.hg.1 | ENST00000463351.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013748.hg.1 | ENST00000463351.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013748.hg.1 | ENST00000469191.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013748.hg.1 | ENST00000469191.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013748.hg.1 | ENST00000469191.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013748.hg.1 | ENST00000469191.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013748.hg.1 | ENST00000469191.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013748.hg.1 | ENST00000479255.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013748.hg.1 | ENST00000479255.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013748.hg.1 | ENST00000479255.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013748.hg.1 | ENST00000479255.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013748.hg.1 | ENST00000479255.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013748.hg.1 | ENST00000488496.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013748.hg.1 | ENST00000488496.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013748.hg.1 | ENST00000488496.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013748.hg.1 | ENST00000488496.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013748.hg.1 | ENST00000489146.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013748.hg.1 | ENST00000489146.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013748.hg.1 | ENST00000489146.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013748.hg.1 | ENST00000489146.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013748.hg.1 | ENST00000490029.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013748.hg.1 | ENST00000490029.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013748.hg.1 | ENST00000490029.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013748.hg.1 | ENST00000490029.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013748.hg.1 | ENST00000538069.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013748.hg.1 | ENST00000538069.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013748.hg.1 | ENST00000538069.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013748.hg.1 | ENST00000538069.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013748.hg.1 | ENST00000538069.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013748.hg.1 | ENST00000538069.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013748.hg.1 | ENST00000538069.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013748.hg.1 | NM_013285.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013748.hg.1 | NM_013285.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013748.hg.1 | NM_013285.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013748.hg.1 | NM_013285.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013748.hg.1 | NM_013285.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013748.hg.1 | NM_013285.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013748.hg.1 | NM_013285.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013748.hg.1 | NM_013285.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013748.hg.1 | NM_013285.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013748.hg.1 | NM_013285.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013748.hg.1 | NM_013285.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013748.hg.1 | NM_013285.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013748.hg.1 | NM_013285.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013748.hg.1 | NM_013285.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013748.hg.1 | NM_013285.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013748.hg.1 | NM_013285.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013748.hg.1 | NM_013285.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013748.hg.1 | NM_013285.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013748.hg.1 | NM_013285.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013748.hg.1 | NM_013285.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013748.hg.1 | NM_013285.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013748.hg.1 | NM_013285.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01039156.hg.1 | TC01002519.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01039156.hg.1 | ENST00000373062.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01039156.hg.1 | ENST00000545489.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01039156.hg.1 | NM_013285.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013748.hg.1 | ENST00000373062.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013748.hg.1 | ENST00000373062.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013748.hg.1 | ENST00000373062.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013748.hg.1 | ENST00000373062.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013748.hg.1 | ENST00000373062.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013748.hg.1 | ENST00000373062.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013748.hg.1 | ENST00000373062.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013748.hg.1 | ENST00000373062.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013748.hg.1 | ENST00000373062.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013748.hg.1 | ENST00000373062.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013748.hg.1 | ENST00000373062.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013748.hg.1 | ENST00000373062.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013748.hg.1 | ENST00000373062.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013748.hg.1 | ENST00000373062.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013748.hg.1 | ENST00000373062.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013748.hg.1 | ENST00000373062.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013748.hg.1 | ENST00000373062.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013748.hg.1 | ENST00000373062.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013748.hg.1 | ENST00000373062.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013748.hg.1 | ENST00000373062.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013748.hg.1 | ENST00000373062.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013748.hg.1 | ENST00000462812.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013748.hg.1 | ENST00000462812.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013748.hg.1 | ENST00000462812.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013748.hg.1 | ENST00000462812.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013748.hg.1 | ENST00000462812.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013748.hg.1 | ENST00000462812.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013748.hg.1 | ENST00000463351.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013748.hg.1 | ENST00000463351.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013748.hg.1 | ENST00000463351.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013748.hg.1 | ENST00000463351.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013748.hg.1 | ENST00000469191.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013748.hg.1 | ENST00000469191.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013748.hg.1 | ENST00000469191.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013748.hg.1 | ENST00000469191.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013748.hg.1 | ENST00000469191.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013748.hg.1 | ENST00000479255.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013748.hg.1 | ENST00000479255.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013748.hg.1 | ENST00000479255.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013748.hg.1 | ENST00000479255.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013748.hg.1 | ENST00000479255.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013748.hg.1 | ENST00000488496.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013748.hg.1 | ENST00000488496.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013748.hg.1 | ENST00000488496.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013748.hg.1 | ENST00000488496.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013748.hg.1 | ENST00000489146.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013748.hg.1 | ENST00000489146.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013748.hg.1 | ENST00000489146.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013748.hg.1 | ENST00000489146.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013748.hg.1 | ENST00000490029.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013748.hg.1 | ENST00000490029.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013748.hg.1 | ENST00000490029.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013748.hg.1 | ENST00000490029.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013748.hg.1 | ENST00000538069.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013748.hg.1 | ENST00000538069.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013748.hg.1 | ENST00000538069.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013748.hg.1 | ENST00000538069.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013748.hg.1 | ENST00000538069.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013748.hg.1 | ENST00000538069.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013748.hg.1 | ENST00000538069.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013748.hg.1 | NM_013285.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013748.hg.1 | NM_013285.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013748.hg.1 | NM_013285.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013748.hg.1 | NM_013285.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013748.hg.1 | NM_013285.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013748.hg.1 | NM_013285.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013748.hg.1 | NM_013285.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013748.hg.1 | NM_013285.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013748.hg.1 | NM_013285.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013748.hg.1 | NM_013285.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013748.hg.1 | NM_013285.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013748.hg.1 | NM_013285.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013748.hg.1 | NM_013285.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013748.hg.1 | NM_013285.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013748.hg.1 | NM_013285.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013748.hg.1 | NM_013285.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013748.hg.1 | NM_013285.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013748.hg.1 | NM_013285.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013748.hg.1 | NM_013285.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013748.hg.1 | NM_013285.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013748.hg.1 | NM_013285.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013748.hg.1 | NM_013285.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100202543.hg.1 | ENST00000373062.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100202543.hg.1 | ENST00000463351.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100202543.hg.1 | NM_013285.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100202543.hg.1 | ENST00000373062.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100202543.hg.1 | ENST00000463351.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100202543.hg.1 | NM_013285.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100202543.hg.1 | TC0100013748.hg.1 | Clariom D TC |
