Assay Details
| Gene Symbol: GNL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001323623.1 | NP_001310552.1 | 6 - 7 | 793 | 140 | |
| NM_013285.2 | NP_037417.1 | 6 - 7 | 793 | 140 | ||
| GenBank mRNA | AK298358.1 | - | 6 - 7 | 703 | 140 | |
| AK312915.1 | - | 6 - 7 | 747 | 140 | ||
| BC000107.2 | - | 6 - 7 | 729 | 3506131 | 140 | |
| BC009250.2 | - | 6 - 7 | 729 | 2960125 | 140 | |
| L05425.1 | - | 6 - 7 | 709 | 140 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01002519.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR01039147.hg.1 | TC01002519.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01039147.hg.1 | ENST00000373062.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01039147.hg.1 | ENST00000545489.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01039147.hg.1 | NM_013285.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013748.hg.1 | ENST00000373062.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013748.hg.1 | ENST00000373062.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013748.hg.1 | ENST00000373062.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013748.hg.1 | ENST00000373062.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013748.hg.1 | ENST00000373062.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013748.hg.1 | ENST00000373062.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013748.hg.1 | ENST00000373062.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013748.hg.1 | ENST00000373062.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013748.hg.1 | ENST00000373062.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013748.hg.1 | ENST00000373062.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013748.hg.1 | ENST00000373062.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013748.hg.1 | ENST00000373062.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013748.hg.1 | ENST00000373062.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013748.hg.1 | ENST00000373062.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013748.hg.1 | ENST00000373062.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013748.hg.1 | ENST00000373062.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013748.hg.1 | ENST00000373062.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013748.hg.1 | ENST00000373062.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013748.hg.1 | ENST00000373062.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013748.hg.1 | ENST00000373062.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013748.hg.1 | ENST00000373062.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013748.hg.1 | ENST00000462812.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013748.hg.1 | ENST00000462812.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013748.hg.1 | ENST00000462812.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013748.hg.1 | ENST00000462812.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013748.hg.1 | ENST00000462812.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013748.hg.1 | ENST00000462812.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013748.hg.1 | ENST00000463351.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013748.hg.1 | ENST00000463351.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013748.hg.1 | ENST00000463351.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013748.hg.1 | ENST00000463351.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013748.hg.1 | ENST00000469191.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013748.hg.1 | ENST00000469191.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013748.hg.1 | ENST00000469191.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013748.hg.1 | ENST00000469191.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013748.hg.1 | ENST00000469191.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013748.hg.1 | ENST00000479255.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013748.hg.1 | ENST00000479255.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013748.hg.1 | ENST00000479255.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013748.hg.1 | ENST00000479255.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013748.hg.1 | ENST00000479255.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013748.hg.1 | ENST00000488496.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013748.hg.1 | ENST00000488496.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013748.hg.1 | ENST00000488496.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013748.hg.1 | ENST00000488496.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013748.hg.1 | ENST00000489146.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013748.hg.1 | ENST00000489146.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013748.hg.1 | ENST00000489146.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013748.hg.1 | ENST00000489146.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013748.hg.1 | ENST00000490029.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013748.hg.1 | ENST00000490029.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013748.hg.1 | ENST00000490029.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013748.hg.1 | ENST00000490029.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013748.hg.1 | ENST00000538069.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013748.hg.1 | ENST00000538069.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013748.hg.1 | ENST00000538069.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013748.hg.1 | ENST00000538069.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013748.hg.1 | ENST00000538069.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013748.hg.1 | ENST00000538069.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013748.hg.1 | ENST00000538069.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013748.hg.1 | NM_013285.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013748.hg.1 | NM_013285.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013748.hg.1 | NM_013285.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013748.hg.1 | NM_013285.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013748.hg.1 | NM_013285.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013748.hg.1 | NM_013285.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013748.hg.1 | NM_013285.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013748.hg.1 | NM_013285.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013748.hg.1 | NM_013285.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013748.hg.1 | NM_013285.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013748.hg.1 | NM_013285.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013748.hg.1 | NM_013285.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013748.hg.1 | NM_013285.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013748.hg.1 | NM_013285.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013748.hg.1 | NM_013285.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013748.hg.1 | NM_013285.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013748.hg.1 | NM_013285.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013748.hg.1 | NM_013285.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013748.hg.1 | NM_013285.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013748.hg.1 | NM_013285.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013748.hg.1 | NM_013285.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013748.hg.1 | NM_013285.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013748.hg.1 | ENST00000373062.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013748.hg.1 | ENST00000373062.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013748.hg.1 | ENST00000373062.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013748.hg.1 | ENST00000373062.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013748.hg.1 | ENST00000373062.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013748.hg.1 | ENST00000373062.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013748.hg.1 | ENST00000373062.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013748.hg.1 | ENST00000373062.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013748.hg.1 | ENST00000373062.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013748.hg.1 | ENST00000373062.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013748.hg.1 | ENST00000373062.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013748.hg.1 | ENST00000373062.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013748.hg.1 | ENST00000373062.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013748.hg.1 | ENST00000373062.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013748.hg.1 | ENST00000373062.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013748.hg.1 | ENST00000373062.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013748.hg.1 | ENST00000373062.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013748.hg.1 | ENST00000373062.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013748.hg.1 | ENST00000373062.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013748.hg.1 | ENST00000373062.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013748.hg.1 | ENST00000373062.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013748.hg.1 | ENST00000462812.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013748.hg.1 | ENST00000462812.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013748.hg.1 | ENST00000462812.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013748.hg.1 | ENST00000462812.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013748.hg.1 | ENST00000462812.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013748.hg.1 | ENST00000462812.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013748.hg.1 | ENST00000463351.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013748.hg.1 | ENST00000463351.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013748.hg.1 | ENST00000463351.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013748.hg.1 | ENST00000463351.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013748.hg.1 | ENST00000469191.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013748.hg.1 | ENST00000469191.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013748.hg.1 | ENST00000469191.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013748.hg.1 | ENST00000469191.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013748.hg.1 | ENST00000469191.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013748.hg.1 | ENST00000479255.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013748.hg.1 | ENST00000479255.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013748.hg.1 | ENST00000479255.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013748.hg.1 | ENST00000479255.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013748.hg.1 | ENST00000479255.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013748.hg.1 | ENST00000488496.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013748.hg.1 | ENST00000488496.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013748.hg.1 | ENST00000488496.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013748.hg.1 | ENST00000488496.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013748.hg.1 | ENST00000489146.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013748.hg.1 | ENST00000489146.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013748.hg.1 | ENST00000489146.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013748.hg.1 | ENST00000489146.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013748.hg.1 | ENST00000490029.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013748.hg.1 | ENST00000490029.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013748.hg.1 | ENST00000490029.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013748.hg.1 | ENST00000490029.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013748.hg.1 | ENST00000538069.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013748.hg.1 | ENST00000538069.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013748.hg.1 | ENST00000538069.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013748.hg.1 | ENST00000538069.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013748.hg.1 | ENST00000538069.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013748.hg.1 | ENST00000538069.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013748.hg.1 | ENST00000538069.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013748.hg.1 | NM_013285.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013748.hg.1 | NM_013285.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013748.hg.1 | NM_013285.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013748.hg.1 | NM_013285.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013748.hg.1 | NM_013285.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013748.hg.1 | NM_013285.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013748.hg.1 | NM_013285.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013748.hg.1 | NM_013285.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013748.hg.1 | NM_013285.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013748.hg.1 | NM_013285.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013748.hg.1 | NM_013285.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013748.hg.1 | NM_013285.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013748.hg.1 | NM_013285.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013748.hg.1 | NM_013285.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013748.hg.1 | NM_013285.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013748.hg.1 | NM_013285.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013748.hg.1 | NM_013285.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013748.hg.1 | NM_013285.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013748.hg.1 | NM_013285.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013748.hg.1 | NM_013285.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013748.hg.1 | NM_013285.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013748.hg.1 | NM_013285.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100202536.hg.1 | ENST00000373062.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100202536.hg.1 | ENST00000489146.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100202536.hg.1 | ENST00000538069.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100202536.hg.1 | NM_013285.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100202536.hg.1 | ENST00000373062.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100202536.hg.1 | ENST00000489146.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100202536.hg.1 | ENST00000538069.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100202536.hg.1 | NM_013285.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100202536.hg.1 | TC0100013748.hg.1 | Clariom D TC |

GNL2
Genomic Map