Assay Details
Gene Symbol: TMEM201 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001130924.2 | NP_001124396.2 | 7 - 8 | 1448 | 61 | |
XM_017000549.1 | XP_016856038.1 | 7 - 8 | 1479 | 61 | ||
XM_017000551.1 | XP_016856040.1 | 7 - 8 | 1689 | 61 | ||
XM_017000552.1 | XP_016856041.1 | 7 - 8 | 1517 | 61 | ||
GenBank mRNA | BG828926.1 | - | 2 - 3 | 317 | 61 | |
DA603558.1 | - | 2 - 3 | 416 | 61 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC01000117.hg.1
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR01001791.hg.1 | TC01000117.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01001791.hg.1 | ENST00000340381.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01001791.hg.1 | ENST00000377376.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01001791.hg.1 | NM_001130924.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01001790.hg.1 | TC01000117.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01001790.hg.1 | ENST00000340381.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01001790.hg.1 | ENST00000377376.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01001790.hg.1 | ENST00000510900.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01001790.hg.1 | NM_001130924.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01001792.hg.1 | TC01000117.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01001792.hg.1 | ENST00000340381.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01001792.hg.1 | ENST00000377376.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01001792.hg.1 | NM_001130924.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01001793.hg.1 | TC01000117.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01001793.hg.1 | ENST00000340381.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01001793.hg.1 | NM_001130924.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100006784.hg.1 | ENST00000340305.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100006784.hg.1 | ENST00000340305.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100006784.hg.1 | ENST00000340305.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100006784.hg.1 | ENST00000340305.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100006784.hg.1 | ENST00000340305.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100006784.hg.1 | ENST00000340305.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100006784.hg.1 | ENST00000340305.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100006784.hg.1 | ENST00000340305.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100006784.hg.1 | ENST00000340305.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100006784.hg.1 | ENST00000340381.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100006784.hg.1 | ENST00000340381.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100006784.hg.1 | ENST00000340381.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100006784.hg.1 | ENST00000340381.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100006784.hg.1 | ENST00000340381.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100006784.hg.1 | ENST00000340381.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100006784.hg.1 | ENST00000340381.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100006784.hg.1 | ENST00000340381.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100006784.hg.1 | ENST00000340381.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100006784.hg.1 | ENST00000340381.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100006784.hg.1 | ENST00000340381.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100006784.hg.1 | ENST00000340381.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100006784.hg.1 | ENST00000340381.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100006784.hg.1 | ENST00000340381.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100006784.hg.1 | ENST00000340381.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100006784.hg.1 | ENST00000340381.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100006784.hg.1 | ENST00000416541.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100006784.hg.1 | ENST00000416541.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100006784.hg.1 | ENST00000416541.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100006784.hg.1 | ENST00000416541.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100006784.hg.1 | ENST00000416541.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100006784.hg.1 | ENST00000416541.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100006784.hg.1 | ENST00000416541.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100006784.hg.1 | ENST00000416541.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100006784.hg.1 | ENST00000416541.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100006784.hg.1 | ENST00000416541.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100006784.hg.1 | ENST00000416541.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100006784.hg.1 | ENST00000416541.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100006784.hg.1 | ENST00000508400.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100006784.hg.1 | ENST00000508400.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100006784.hg.1 | ENST00000508400.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100006784.hg.1 | ENST00000508400.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100006784.hg.1 | ENST00000508400.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100006784.hg.1 | ENST00000508400.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100006784.hg.1 | ENST00000508400.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100006784.hg.1 | ENST00000510900.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100006784.hg.1 | ENST00000510900.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100006784.hg.1 | NM_001010866.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100006784.hg.1 | NM_001010866.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100006784.hg.1 | NM_001010866.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100006784.hg.1 | NM_001010866.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100006784.hg.1 | NM_001010866.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100006784.hg.1 | NM_001010866.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100006784.hg.1 | NM_001010866.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100006784.hg.1 | NM_001010866.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100006784.hg.1 | NM_001010866.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100006784.hg.1 | NM_001010866.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100006784.hg.1 | NM_001130924.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100006784.hg.1 | NM_001130924.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100006784.hg.1 | NM_001130924.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100006784.hg.1 | NM_001130924.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100006784.hg.1 | NM_001130924.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100006784.hg.1 | NM_001130924.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100006784.hg.1 | NM_001130924.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100006784.hg.1 | NM_001130924.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100006784.hg.1 | NM_001130924.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100006784.hg.1 | NM_001130924.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100006784.hg.1 | NM_001130924.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100006784.hg.1 | NM_001130924.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100006784.hg.1 | NM_001130924.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100006784.hg.1 | NM_001130924.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100006784.hg.1 | NM_001130924.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100006784.hg.1 | NM_001130924.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100006784.hg.1 | NM_001130924.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100006784.hg.1 | ENST00000340305.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100006784.hg.1 | ENST00000340305.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100006784.hg.1 | ENST00000340305.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100006784.hg.1 | ENST00000340305.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100006784.hg.1 | ENST00000340305.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100006784.hg.1 | ENST00000340305.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100006784.hg.1 | ENST00000340305.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100006784.hg.1 | ENST00000340305.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100006784.hg.1 | ENST00000340305.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100006784.hg.1 | ENST00000340381.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100006784.hg.1 | ENST00000340381.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100006784.hg.1 | ENST00000340381.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100006784.hg.1 | ENST00000340381.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100006784.hg.1 | ENST00000340381.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100006784.hg.1 | ENST00000340381.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100006784.hg.1 | ENST00000340381.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100006784.hg.1 | ENST00000340381.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100006784.hg.1 | ENST00000340381.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100006784.hg.1 | ENST00000340381.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100006784.hg.1 | ENST00000340381.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100006784.hg.1 | ENST00000340381.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100006784.hg.1 | ENST00000340381.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100006784.hg.1 | ENST00000340381.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100006784.hg.1 | ENST00000340381.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100006784.hg.1 | ENST00000340381.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100006784.hg.1 | ENST00000416541.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100006784.hg.1 | ENST00000416541.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100006784.hg.1 | ENST00000416541.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100006784.hg.1 | ENST00000416541.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100006784.hg.1 | ENST00000416541.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100006784.hg.1 | ENST00000416541.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100006784.hg.1 | ENST00000416541.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100006784.hg.1 | ENST00000416541.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100006784.hg.1 | ENST00000416541.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100006784.hg.1 | ENST00000416541.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100006784.hg.1 | ENST00000416541.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100006784.hg.1 | ENST00000416541.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100006784.hg.1 | ENST00000508400.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100006784.hg.1 | ENST00000508400.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100006784.hg.1 | ENST00000508400.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100006784.hg.1 | ENST00000508400.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100006784.hg.1 | ENST00000508400.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100006784.hg.1 | ENST00000508400.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100006784.hg.1 | ENST00000508400.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100006784.hg.1 | ENST00000510900.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100006784.hg.1 | ENST00000510900.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100006784.hg.1 | NM_001010866.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100006784.hg.1 | NM_001010866.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100006784.hg.1 | NM_001010866.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100006784.hg.1 | NM_001010866.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100006784.hg.1 | NM_001010866.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100006784.hg.1 | NM_001010866.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100006784.hg.1 | NM_001010866.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100006784.hg.1 | NM_001010866.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100006784.hg.1 | NM_001010866.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100006784.hg.1 | NM_001010866.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100006784.hg.1 | NM_001130924.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100006784.hg.1 | NM_001130924.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100006784.hg.1 | NM_001130924.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100006784.hg.1 | NM_001130924.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100006784.hg.1 | NM_001130924.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100006784.hg.1 | NM_001130924.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100006784.hg.1 | NM_001130924.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100006784.hg.1 | NM_001130924.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100006784.hg.1 | NM_001130924.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100006784.hg.1 | NM_001130924.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100006784.hg.1 | NM_001130924.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100006784.hg.1 | NM_001130924.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100006784.hg.1 | NM_001130924.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100006784.hg.1 | NM_001130924.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100006784.hg.1 | NM_001130924.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100006784.hg.1 | NM_001130924.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100006784.hg.1 | NM_001130924.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100146933.hg.1 | ENST00000340381.10 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100146933.hg.1 | ENST00000416541.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100146933.hg.1 | ENST00000510900.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100146933.hg.1 | NM_001130924.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100146933.hg.1 | ENST00000340381.10 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100146933.hg.1 | ENST00000416541.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100146933.hg.1 | ENST00000510900.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100146933.hg.1 | NM_001130924.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100146933.hg.1 | TC0100006784.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100146934.hg.1 | ENST00000340381.10 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100146934.hg.1 | ENST00000416541.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100146934.hg.1 | NM_001130924.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100146934.hg.1 | ENST00000340381.10 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100146934.hg.1 | ENST00000416541.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100146934.hg.1 | NM_001130924.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100146934.hg.1 | TC0100006784.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100146936.hg.1 | ENST00000340381.10 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100146936.hg.1 | ENST00000508400.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100146936.hg.1 | NM_001130924.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100146936.hg.1 | ENST00000340381.10 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100146936.hg.1 | ENST00000508400.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100146936.hg.1 | NM_001130924.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100146936.hg.1 | TC0100006784.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100146935.hg.1 | ENST00000340381.10 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100146935.hg.1 | ENST00000416541.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100146935.hg.1 | ENST00000508400.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100146935.hg.1 | NM_001130924.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100146935.hg.1 | ENST00000340381.10 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100146935.hg.1 | ENST00000416541.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100146935.hg.1 | ENST00000508400.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100146935.hg.1 | NM_001130924.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100146935.hg.1 | TC0100006784.hg.1 | Clariom D TC |
