Assay Details
| Gene Symbol: BMPER | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133468.4 | NP_597725.1 | 13 - 14 | 1787 | 84 | |
| XM_017011800.1 | XP_016867289.1 | 11 - 12 | 1347 | 84 | ||
| GenBank mRNA | AB075845.1 | - | 9 - 10 | 1054 | 84 | |
| AK289963.1 | - | 12 - 13 | 1527 | 84 | ||
| AY324650.1 | - | 13 - 14 | 1786 | 84 | ||
| BC060868.1 | - | 12 - 13 | 1464 | 30345290 | 84 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC07000220.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
ureteric bud development
blood vessel endothelial cell proliferation involved in sprouting angiogenesis
regulation of endothelial cell migration
negative regulation of BMP signaling pathway
endothelial cell activation
endothelial cell differentiation
inner ear development
regulation of pathway-restricted SMAD protein phosphorylation
positive regulation of ERK1 and ERK2 cascade
ureteric bud development
blood vessel endothelial cell proliferation involved in sprouting angiogenesis
regulation of endothelial cell migration
negative regulation of BMP signaling pathway
endothelial cell activation
endothelial cell differentiation
inner ear development
regulation of pathway-restricted SMAD protein phosphorylation
positive regulation of ERK1 and ERK2 cascade
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0700007149.hg.1 | ENST00000297161.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007149.hg.1 | ENST00000297161.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007149.hg.1 | ENST00000297161.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007149.hg.1 | ENST00000297161.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007149.hg.1 | ENST00000297161.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007149.hg.1 | ENST00000297161.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007149.hg.1 | ENST00000297161.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007149.hg.1 | ENST00000297161.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007149.hg.1 | ENST00000297161.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007149.hg.1 | ENST00000297161.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007149.hg.1 | ENST00000297161.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007149.hg.1 | ENST00000297161.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007149.hg.1 | ENST00000297161.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007149.hg.1 | ENST00000297161.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007149.hg.1 | ENST00000297161.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007149.hg.1 | ENST00000297161.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007149.hg.1 | ENST00000297161.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007149.hg.1 | ENST00000297161.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007149.hg.1 | ENST00000297161.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007149.hg.1 | ENST00000297161.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007149.hg.1 | ENST00000297161.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007149.hg.1 | ENST00000297161.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007149.hg.1 | ENST00000436222.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007149.hg.1 | ENST00000436222.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007149.hg.1 | ENST00000436222.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007149.hg.1 | ENST00000436222.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007149.hg.1 | ENST00000436222.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007149.hg.1 | ENST00000436222.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007149.hg.1 | ENST00000444773.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007149.hg.1 | ENST00000444773.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007149.hg.1 | ENST00000444773.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007149.hg.1 | ENST00000444773.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007149.hg.1 | ENST00000444773.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007149.hg.1 | ENST00000444773.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007149.hg.1 | ENST00000448280.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007149.hg.1 | ENST00000448280.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007149.hg.1 | ENST00000448280.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007149.hg.1 | ENST00000448280.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007149.hg.1 | ENST00000448280.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007149.hg.1 | ENST00000448280.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007149.hg.1 | ENST00000448280.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007149.hg.1 | ENST00000476525.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007149.hg.1 | ENST00000476525.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007149.hg.1 | ENST00000476525.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007149.hg.1 | ENST00000494786.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007149.hg.1 | ENST00000494786.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007149.hg.1 | ENST00000494786.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007149.hg.1 | ENST00000494786.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007149.hg.1 | ENST00000496609.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007149.hg.1 | ENST00000496609.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007149.hg.1 | ENST00000496609.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007149.hg.1 | ENST00000496609.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007149.hg.1 | ENST00000496609.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007149.hg.1 | ENST00000496609.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007149.hg.1 | NM_133468.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007149.hg.1 | NM_133468.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007149.hg.1 | NM_133468.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007149.hg.1 | NM_133468.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007149.hg.1 | NM_133468.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007149.hg.1 | NM_133468.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007149.hg.1 | NM_133468.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007149.hg.1 | NM_133468.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007149.hg.1 | NM_133468.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007149.hg.1 | NM_133468.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007149.hg.1 | NM_133468.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007149.hg.1 | NM_133468.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007149.hg.1 | NM_133468.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007149.hg.1 | NM_133468.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007149.hg.1 | NM_133468.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007149.hg.1 | NM_133468.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007149.hg.1 | NM_133468.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007149.hg.1 | NM_133468.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007149.hg.1 | NM_133468.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007149.hg.1 | NM_133468.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007149.hg.1 | NM_133468.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007149.hg.1 | NM_133468.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07003467.hg.1 | TC07000220.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR07003467.hg.1 | ENST00000297161.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07003467.hg.1 | ENST00000426693.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07003467.hg.1 | NM_133468.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700149798.hg.1 | ENST00000297161.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700149798.hg.1 | ENST00000476525.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700149798.hg.1 | NM_133468.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700149798.hg.1 | ENST00000297161.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700149798.hg.1 | ENST00000476525.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700149798.hg.1 | NM_133468.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700149798.hg.1 | TC0700007149.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0700007149.hg.1 | ENST00000297161.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007149.hg.1 | ENST00000297161.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007149.hg.1 | ENST00000297161.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007149.hg.1 | ENST00000297161.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007149.hg.1 | ENST00000297161.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007149.hg.1 | ENST00000297161.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007149.hg.1 | ENST00000297161.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007149.hg.1 | ENST00000297161.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007149.hg.1 | ENST00000297161.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007149.hg.1 | ENST00000297161.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007149.hg.1 | ENST00000297161.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007149.hg.1 | ENST00000297161.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007149.hg.1 | ENST00000297161.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007149.hg.1 | ENST00000297161.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007149.hg.1 | ENST00000297161.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007149.hg.1 | ENST00000297161.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007149.hg.1 | ENST00000297161.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007149.hg.1 | ENST00000297161.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007149.hg.1 | ENST00000297161.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007149.hg.1 | ENST00000297161.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007149.hg.1 | ENST00000297161.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007149.hg.1 | ENST00000297161.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007149.hg.1 | ENST00000436222.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007149.hg.1 | ENST00000436222.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007149.hg.1 | ENST00000436222.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007149.hg.1 | ENST00000436222.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007149.hg.1 | ENST00000436222.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007149.hg.1 | ENST00000436222.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007149.hg.1 | ENST00000444773.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007149.hg.1 | ENST00000444773.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007149.hg.1 | ENST00000444773.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007149.hg.1 | ENST00000444773.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007149.hg.1 | ENST00000444773.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007149.hg.1 | ENST00000444773.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007149.hg.1 | ENST00000448280.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007149.hg.1 | ENST00000448280.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007149.hg.1 | ENST00000448280.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007149.hg.1 | ENST00000448280.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007149.hg.1 | ENST00000448280.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007149.hg.1 | ENST00000448280.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007149.hg.1 | ENST00000448280.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007149.hg.1 | ENST00000476525.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007149.hg.1 | ENST00000476525.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007149.hg.1 | ENST00000476525.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007149.hg.1 | ENST00000494786.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007149.hg.1 | ENST00000494786.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007149.hg.1 | ENST00000494786.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007149.hg.1 | ENST00000494786.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007149.hg.1 | ENST00000496609.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007149.hg.1 | ENST00000496609.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007149.hg.1 | ENST00000496609.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007149.hg.1 | ENST00000496609.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007149.hg.1 | ENST00000496609.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007149.hg.1 | ENST00000496609.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007149.hg.1 | NM_133468.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007149.hg.1 | NM_133468.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007149.hg.1 | NM_133468.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007149.hg.1 | NM_133468.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007149.hg.1 | NM_133468.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007149.hg.1 | NM_133468.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007149.hg.1 | NM_133468.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007149.hg.1 | NM_133468.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007149.hg.1 | NM_133468.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007149.hg.1 | NM_133468.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007149.hg.1 | NM_133468.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007149.hg.1 | NM_133468.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007149.hg.1 | NM_133468.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007149.hg.1 | NM_133468.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007149.hg.1 | NM_133468.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007149.hg.1 | NM_133468.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007149.hg.1 | NM_133468.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007149.hg.1 | NM_133468.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007149.hg.1 | NM_133468.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007149.hg.1 | NM_133468.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007149.hg.1 | NM_133468.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007149.hg.1 | NM_133468.4 | Refseq |

BMPER
Genomic Map