Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015506.hg.1 |
ENST00000403263.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015506.hg.1 |
ENST00000403263.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015506.hg.1 |
ENST00000403263.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015506.hg.1 |
ENST00000403263.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015506.hg.1 |
ENST00000403263.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015506.hg.1 |
ENST00000403263.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015506.hg.1 |
ENST00000403263.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015506.hg.1 |
ENST00000403263.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015506.hg.1 |
ENST00000403263.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015506.hg.1 |
ENST00000403263.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015506.hg.1 |
ENST00000403263.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015506.hg.1 |
ENST00000403263.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015506.hg.1 |
ENST00000403263.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015506.hg.1 |
ENST00000403263.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015506.hg.1 |
ENST00000403263.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015506.hg.1 |
ENST00000403263.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015506.hg.1 |
ENST00000403263.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015506.hg.1 |
ENST00000414320.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015506.hg.1 |
ENST00000414320.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015506.hg.1 |
ENST00000414320.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015506.hg.1 |
ENST00000414320.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015506.hg.1 |
ENST00000414320.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015506.hg.1 |
ENST00000414320.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015506.hg.1 |
ENST00000414320.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015506.hg.1 |
ENST00000424117.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015506.hg.1 |
ENST00000424117.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015506.hg.1 |
ENST00000424117.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015506.hg.1 |
ENST00000424117.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015506.hg.1 |
ENST00000424117.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015506.hg.1 |
ENST00000424117.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015506.hg.1 |
ENST00000424117.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015506.hg.1 |
ENST00000424117.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015506.hg.1 |
ENST00000424117.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015506.hg.1 |
NM_017759.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015506.hg.1 |
NM_017759.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015506.hg.1 |
NM_017759.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015506.hg.1 |
NM_017759.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015506.hg.1 |
NM_017759.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015506.hg.1 |
NM_017759.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015506.hg.1 |
NM_017759.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015506.hg.1 |
NM_017759.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015506.hg.1 |
NM_017759.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015506.hg.1 |
NM_017759.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015506.hg.1 |
NM_017759.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015506.hg.1 |
NM_017759.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015506.hg.1 |
NM_017759.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015506.hg.1 |
NM_017759.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015506.hg.1 |
NM_017759.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015506.hg.1 |
NM_017759.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015506.hg.1 |
NM_017759.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02043436.hg.1 |
TC02002693.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02043436.hg.1 |
ENST00000233270.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02043436.hg.1 |
ENST00000403263.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02043436.hg.1 |
NM_017759.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02043437.hg.1 |
TC02002693.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02043437.hg.1 |
ENST00000233270.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02043437.hg.1 |
ENST00000403263.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02043437.hg.1 |
NM_017759.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200213854.hg.1 |
ENST00000403263.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200213854.hg.1 |
NM_017759.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200213854.hg.1 |
ENST00000403263.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200213854.hg.1 |
NM_017759.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200213854.hg.1 |
TC0200015506.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200213855.hg.1 |
ENST00000403263.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200213855.hg.1 |
ENST00000424117.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200213855.hg.1 |
NM_017759.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200213855.hg.1 |
ENST00000403263.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200213855.hg.1 |
ENST00000424117.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200213855.hg.1 |
NM_017759.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200213855.hg.1 |
TC0200015506.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0200015506.hg.1 |
ENST00000403263.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015506.hg.1 |
ENST00000403263.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015506.hg.1 |
ENST00000403263.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015506.hg.1 |
ENST00000403263.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015506.hg.1 |
ENST00000403263.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015506.hg.1 |
ENST00000403263.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015506.hg.1 |
ENST00000403263.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015506.hg.1 |
ENST00000403263.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015506.hg.1 |
ENST00000403263.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015506.hg.1 |
ENST00000403263.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015506.hg.1 |
ENST00000403263.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015506.hg.1 |
ENST00000403263.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015506.hg.1 |
ENST00000403263.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015506.hg.1 |
ENST00000403263.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015506.hg.1 |
ENST00000403263.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015506.hg.1 |
ENST00000403263.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015506.hg.1 |
ENST00000403263.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015506.hg.1 |
ENST00000414320.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015506.hg.1 |
ENST00000414320.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015506.hg.1 |
ENST00000414320.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015506.hg.1 |
ENST00000414320.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015506.hg.1 |
ENST00000414320.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015506.hg.1 |
ENST00000414320.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015506.hg.1 |
ENST00000414320.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015506.hg.1 |
ENST00000424117.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015506.hg.1 |
ENST00000424117.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015506.hg.1 |
ENST00000424117.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015506.hg.1 |
ENST00000424117.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015506.hg.1 |
ENST00000424117.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015506.hg.1 |
ENST00000424117.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015506.hg.1 |
ENST00000424117.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015506.hg.1 |
ENST00000424117.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015506.hg.1 |
ENST00000424117.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015506.hg.1 |
NM_017759.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015506.hg.1 |
NM_017759.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015506.hg.1 |
NM_017759.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015506.hg.1 |
NM_017759.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015506.hg.1 |
NM_017759.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015506.hg.1 |
NM_017759.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015506.hg.1 |
NM_017759.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015506.hg.1 |
NM_017759.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015506.hg.1 |
NM_017759.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015506.hg.1 |
NM_017759.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015506.hg.1 |
NM_017759.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015506.hg.1 |
NM_017759.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015506.hg.1 |
NM_017759.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015506.hg.1 |
NM_017759.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015506.hg.1 |
NM_017759.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015506.hg.1 |
NM_017759.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015506.hg.1 |
NM_017759.4 |
Refseq |