Assay Details
Gene Symbol: PSMC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_002803.3 | NP_002794.1 | 7 - 8 | 780 | 78 | |
GenBank mRNA | AB075520.1 | - | 7 - 8 | 641 | 78 | |
AK298821.1 | - | 6 - 7 | 541 | 78 | ||
AK312648.1 | - | 7 - 8 | 631 | 78 | ||
BC002589.2 | - | 7 - 8 | 624 | 3161790 | 78 | |
D11094.1 | - | 7 - 8 | 658 | 78 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC07000673.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
MAPK cascade
protein polyubiquitination
osteoblast differentiation
stimulatory C-type lectin receptor signaling pathway
antigen processing and presentation of exogenous peptide antigen via MHC class I, TAP-dependent
ubiquitin-dependent protein catabolic process
regulation of cellular amino acid metabolic process
viral process
ER-associated ubiquitin-dependent protein catabolic process
anaphase-promoting complex-dependent catabolic process
tumor necrosis factor-mediated signaling pathway
NIK/NF-kappaB signaling
Fc-epsilon receptor signaling pathway
proteasome-mediated ubiquitin-dependent protein catabolic process
regulation of mRNA stability
positive regulation of RNA polymerase II transcriptional preinitiation complex assembly
T cell receptor signaling pathway
negative regulation of ubiquitin-protein ligase activity involved in mitotic cell cycle
positive regulation of ubiquitin-protein ligase activity involved in regulation of mitotic cell cycle transition
Wnt signaling pathway, planar cell polarity pathway
negative regulation of canonical Wnt signaling pathway
positive regulation of canonical Wnt signaling pathway
positive regulation of proteasomal protein catabolic process
protein binding
ATP binding
ATPase activity
TBP-class protein binding
proteasome-activating ATPase activity
MAPK cascade
protein polyubiquitination
osteoblast differentiation
stimulatory C-type lectin receptor signaling pathway
antigen processing and presentation of exogenous peptide antigen via MHC class I, TAP-dependent
ubiquitin-dependent protein catabolic process
regulation of cellular amino acid metabolic process
viral process
ER-associated ubiquitin-dependent protein catabolic process
anaphase-promoting complex-dependent catabolic process
tumor necrosis factor-mediated signaling pathway
NIK/NF-kappaB signaling
Fc-epsilon receptor signaling pathway
proteasome-mediated ubiquitin-dependent protein catabolic process
regulation of mRNA stability
positive regulation of RNA polymerase II transcriptional preinitiation complex assembly
T cell receptor signaling pathway
negative regulation of ubiquitin-protein ligase activity involved in mitotic cell cycle
positive regulation of ubiquitin-protein ligase activity involved in regulation of mitotic cell cycle transition
Wnt signaling pathway, planar cell polarity pathway
negative regulation of canonical Wnt signaling pathway
positive regulation of canonical Wnt signaling pathway
positive regulation of proteasomal protein catabolic process
protein binding
ATP binding
ATPase activity
TBP-class protein binding
proteasome-activating ATPase activity
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR07010470.hg.1 | TC07000673.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR07010470.hg.1 | ENST00000292644.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR07010470.hg.1 | ENST00000435765.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR07010470.hg.1 | ENST00000544811.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR07010470.hg.1 | NM_002803.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700008667.hg.1 | ENST00000292644.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008667.hg.1 | ENST00000292644.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008667.hg.1 | ENST00000292644.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008667.hg.1 | ENST00000292644.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008667.hg.1 | ENST00000292644.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008667.hg.1 | ENST00000292644.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008667.hg.1 | ENST00000292644.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008667.hg.1 | ENST00000292644.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008667.hg.1 | ENST00000292644.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008667.hg.1 | ENST00000292644.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008667.hg.1 | ENST00000292644.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008667.hg.1 | ENST00000292644.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008667.hg.1 | ENST00000292644.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008667.hg.1 | ENST00000292644.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008667.hg.1 | ENST00000292644.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008667.hg.1 | ENST00000425206.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008667.hg.1 | ENST00000425206.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008667.hg.1 | ENST00000425206.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008667.hg.1 | ENST00000425206.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008667.hg.1 | ENST00000425206.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008667.hg.1 | ENST00000425206.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008667.hg.1 | ENST00000425206.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008667.hg.1 | ENST00000425206.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008667.hg.1 | ENST00000435765.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008667.hg.1 | ENST00000435765.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008667.hg.1 | ENST00000435765.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008667.hg.1 | ENST00000435765.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008667.hg.1 | ENST00000435765.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008667.hg.1 | ENST00000435765.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008667.hg.1 | ENST00000435765.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008667.hg.1 | ENST00000435765.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008667.hg.1 | ENST00000435765.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008667.hg.1 | ENST00000435765.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008667.hg.1 | ENST00000435765.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008667.hg.1 | ENST00000435765.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008667.hg.1 | ENST00000435765.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008667.hg.1 | ENST00000435765.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008667.hg.1 | ENST00000435765.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008667.hg.1 | ENST00000435765.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008667.hg.1 | ENST00000435765.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008667.hg.1 | ENST00000435765.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008667.hg.1 | ENST00000457587.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008667.hg.1 | ENST00000457587.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008667.hg.1 | ENST00000457587.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008667.hg.1 | ENST00000457587.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008667.hg.1 | ENST00000457587.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008667.hg.1 | ENST00000457587.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008667.hg.1 | ENST00000457587.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008667.hg.1 | ENST00000457587.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008667.hg.1 | ENST00000460021.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008667.hg.1 | ENST00000460021.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008667.hg.1 | ENST00000460021.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008667.hg.1 | ENST00000460021.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008667.hg.1 | ENST00000460021.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008667.hg.1 | ENST00000460021.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008667.hg.1 | NM_001204453.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700008667.hg.1 | NM_001204453.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700008667.hg.1 | NM_001204453.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700008667.hg.1 | NM_001204453.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700008667.hg.1 | NM_001204453.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700008667.hg.1 | NM_001204453.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700008667.hg.1 | NM_001204453.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700008667.hg.1 | NM_001204453.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700008667.hg.1 | NM_001204453.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700008667.hg.1 | NM_002803.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700008667.hg.1 | NM_002803.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700008667.hg.1 | NM_002803.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700008667.hg.1 | NM_002803.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700008667.hg.1 | NM_002803.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700008667.hg.1 | NM_002803.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700008667.hg.1 | NM_002803.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700008667.hg.1 | NM_002803.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700008667.hg.1 | NM_002803.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700008667.hg.1 | NM_002803.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700008667.hg.1 | NM_002803.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700008667.hg.1 | NM_002803.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700008667.hg.1 | NM_002803.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700008667.hg.1 | NM_002803.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700008667.hg.1 | NM_002803.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700008667.hg.1 | NM_002803.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700008667.hg.1 | NM_002803.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700161316.hg.1 | ENST00000292644.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700161316.hg.1 | ENST00000435765.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700161316.hg.1 | NM_002803.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700161316.hg.1 | ENST00000292644.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700161316.hg.1 | ENST00000435765.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700161316.hg.1 | NM_002803.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700161316.hg.1 | TC0700008667.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0700008667.hg.1 | ENST00000292644.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008667.hg.1 | ENST00000292644.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008667.hg.1 | ENST00000292644.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008667.hg.1 | ENST00000292644.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008667.hg.1 | ENST00000292644.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008667.hg.1 | ENST00000292644.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008667.hg.1 | ENST00000292644.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008667.hg.1 | ENST00000292644.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008667.hg.1 | ENST00000292644.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008667.hg.1 | ENST00000292644.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008667.hg.1 | ENST00000292644.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008667.hg.1 | ENST00000292644.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008667.hg.1 | ENST00000292644.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008667.hg.1 | ENST00000292644.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008667.hg.1 | ENST00000292644.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008667.hg.1 | ENST00000425206.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008667.hg.1 | ENST00000425206.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008667.hg.1 | ENST00000425206.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008667.hg.1 | ENST00000425206.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008667.hg.1 | ENST00000425206.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008667.hg.1 | ENST00000425206.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008667.hg.1 | ENST00000425206.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008667.hg.1 | ENST00000425206.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008667.hg.1 | ENST00000435765.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008667.hg.1 | ENST00000435765.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008667.hg.1 | ENST00000435765.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008667.hg.1 | ENST00000435765.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008667.hg.1 | ENST00000435765.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008667.hg.1 | ENST00000435765.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008667.hg.1 | ENST00000435765.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008667.hg.1 | ENST00000435765.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008667.hg.1 | ENST00000435765.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008667.hg.1 | ENST00000435765.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008667.hg.1 | ENST00000435765.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008667.hg.1 | ENST00000435765.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008667.hg.1 | ENST00000435765.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008667.hg.1 | ENST00000435765.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008667.hg.1 | ENST00000435765.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008667.hg.1 | ENST00000435765.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008667.hg.1 | ENST00000435765.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008667.hg.1 | ENST00000435765.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008667.hg.1 | ENST00000457587.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008667.hg.1 | ENST00000457587.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008667.hg.1 | ENST00000457587.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008667.hg.1 | ENST00000457587.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008667.hg.1 | ENST00000457587.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008667.hg.1 | ENST00000457587.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008667.hg.1 | ENST00000457587.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008667.hg.1 | ENST00000457587.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008667.hg.1 | ENST00000460021.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008667.hg.1 | ENST00000460021.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008667.hg.1 | ENST00000460021.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008667.hg.1 | ENST00000460021.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008667.hg.1 | ENST00000460021.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008667.hg.1 | ENST00000460021.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008667.hg.1 | NM_001204453.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700008667.hg.1 | NM_001204453.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700008667.hg.1 | NM_001204453.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700008667.hg.1 | NM_001204453.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700008667.hg.1 | NM_001204453.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700008667.hg.1 | NM_001204453.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700008667.hg.1 | NM_001204453.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700008667.hg.1 | NM_001204453.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700008667.hg.1 | NM_001204453.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700008667.hg.1 | NM_002803.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0700008667.hg.1 | NM_002803.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0700008667.hg.1 | NM_002803.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0700008667.hg.1 | NM_002803.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0700008667.hg.1 | NM_002803.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0700008667.hg.1 | NM_002803.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0700008667.hg.1 | NM_002803.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0700008667.hg.1 | NM_002803.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0700008667.hg.1 | NM_002803.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0700008667.hg.1 | NM_002803.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0700008667.hg.1 | NM_002803.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0700008667.hg.1 | NM_002803.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0700008667.hg.1 | NM_002803.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0700008667.hg.1 | NM_002803.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0700008667.hg.1 | NM_002803.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0700008667.hg.1 | NM_002803.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0700008667.hg.1 | NM_002803.3 | Refseq |
