Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR12025336.hg.1 |
ENST00000266856.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12025336.hg.1 |
ENST00000299162.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12025336.hg.1 |
ENST00000355216.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12025336.hg.1 |
ENST00000468516.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12025336.hg.1 |
NM_213596.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12025336.hg.1 |
TC12001948.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011865.hg.1 |
ENST00000299162.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011865.hg.1 |
ENST00000299162.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011865.hg.1 |
ENST00000299162.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011865.hg.1 |
ENST00000299162.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011865.hg.1 |
ENST00000299162.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011865.hg.1 |
ENST00000299162.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011865.hg.1 |
ENST00000299162.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011865.hg.1 |
ENST00000299162.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011865.hg.1 |
ENST00000299162.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011865.hg.1 |
ENST00000299162.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011865.hg.1 |
ENST00000299162.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011865.hg.1 |
ENST00000299162.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011865.hg.1 |
ENST00000299162.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011865.hg.1 |
ENST00000299162.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011865.hg.1 |
ENST00000299162.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011865.hg.1 |
ENST00000355216.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011865.hg.1 |
ENST00000355216.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011865.hg.1 |
ENST00000355216.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011865.hg.1 |
ENST00000355216.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011865.hg.1 |
ENST00000355216.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011865.hg.1 |
ENST00000355216.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011865.hg.1 |
ENST00000355216.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011865.hg.1 |
ENST00000355216.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011865.hg.1 |
ENST00000355216.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011865.hg.1 |
ENST00000355216.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011865.hg.1 |
ENST00000355216.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011865.hg.1 |
ENST00000423960.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011865.hg.1 |
ENST00000423960.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011865.hg.1 |
ENST00000423960.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011865.hg.1 |
ENST00000423960.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011865.hg.1 |
ENST00000423960.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011865.hg.1 |
ENST00000423960.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011865.hg.1 |
ENST00000423960.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011865.hg.1 |
ENST00000423960.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011865.hg.1 |
ENST00000468516.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011865.hg.1 |
ENST00000468516.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011865.hg.1 |
ENST00000468516.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011865.hg.1 |
NM_213596.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011865.hg.1 |
NM_213596.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011865.hg.1 |
NM_213596.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011865.hg.1 |
NM_213596.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011865.hg.1 |
NM_213596.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011865.hg.1 |
NM_213596.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011865.hg.1 |
NM_213596.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011865.hg.1 |
NM_213596.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011865.hg.1 |
NM_213596.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011865.hg.1 |
NM_213596.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011865.hg.1 |
NM_213596.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011865.hg.1 |
NM_213596.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011865.hg.1 |
NM_213596.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011865.hg.1 |
NM_213596.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011865.hg.1 |
NM_213596.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12025335.hg.1 |
ENST00000266856.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12025335.hg.1 |
ENST00000299162.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12025335.hg.1 |
ENST00000355216.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12025335.hg.1 |
NM_213596.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12025335.hg.1 |
TC12001948.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1200192009.hg.1 |
ENST00000299162.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200192009.hg.1 |
ENST00000355216.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200192009.hg.1 |
ENST00000423960.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200192009.hg.1 |
ENST00000468516.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200192009.hg.1 |
NM_213596.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200192009.hg.1 |
ENST00000299162.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200192009.hg.1 |
ENST00000355216.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200192009.hg.1 |
ENST00000423960.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200192009.hg.1 |
ENST00000468516.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200192009.hg.1 |
NM_213596.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200192009.hg.1 |
TC1200011865.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1200011865.hg.1 |
ENST00000299162.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011865.hg.1 |
ENST00000299162.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011865.hg.1 |
ENST00000299162.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011865.hg.1 |
ENST00000299162.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011865.hg.1 |
ENST00000299162.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011865.hg.1 |
ENST00000299162.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011865.hg.1 |
ENST00000299162.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011865.hg.1 |
ENST00000299162.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011865.hg.1 |
ENST00000299162.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011865.hg.1 |
ENST00000299162.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011865.hg.1 |
ENST00000299162.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011865.hg.1 |
ENST00000299162.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011865.hg.1 |
ENST00000299162.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011865.hg.1 |
ENST00000299162.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011865.hg.1 |
ENST00000299162.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011865.hg.1 |
ENST00000355216.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011865.hg.1 |
ENST00000355216.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011865.hg.1 |
ENST00000355216.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011865.hg.1 |
ENST00000355216.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011865.hg.1 |
ENST00000355216.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011865.hg.1 |
ENST00000355216.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011865.hg.1 |
ENST00000355216.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011865.hg.1 |
ENST00000355216.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011865.hg.1 |
ENST00000355216.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011865.hg.1 |
ENST00000355216.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011865.hg.1 |
ENST00000355216.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011865.hg.1 |
ENST00000423960.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011865.hg.1 |
ENST00000423960.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011865.hg.1 |
ENST00000423960.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011865.hg.1 |
ENST00000423960.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011865.hg.1 |
ENST00000423960.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011865.hg.1 |
ENST00000423960.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011865.hg.1 |
ENST00000423960.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011865.hg.1 |
ENST00000423960.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011865.hg.1 |
ENST00000468516.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011865.hg.1 |
ENST00000468516.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011865.hg.1 |
ENST00000468516.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011865.hg.1 |
NM_213596.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011865.hg.1 |
NM_213596.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011865.hg.1 |
NM_213596.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011865.hg.1 |
NM_213596.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011865.hg.1 |
NM_213596.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011865.hg.1 |
NM_213596.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011865.hg.1 |
NM_213596.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011865.hg.1 |
NM_213596.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011865.hg.1 |
NM_213596.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011865.hg.1 |
NM_213596.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011865.hg.1 |
NM_213596.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011865.hg.1 |
NM_213596.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011865.hg.1 |
NM_213596.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011865.hg.1 |
NM_213596.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011865.hg.1 |
NM_213596.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200192008.hg.1 |
ENST00000299162.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200192008.hg.1 |
ENST00000355216.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200192008.hg.1 |
ENST00000423960.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200192008.hg.1 |
NM_213596.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200192008.hg.1 |
ENST00000299162.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200192008.hg.1 |
ENST00000355216.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200192008.hg.1 |
ENST00000423960.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200192008.hg.1 |
NM_213596.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200192008.hg.1 |
TC1200011865.hg.1 |
Clariom D TC |