Assay Details
Gene Symbol: THAP12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_004705.3 | NP_004696.2 | 2 - 3 | 330 | 146 | |
GenBank mRNA | AF007393.1 | - | 2 - 3 | 201 | 146 | |
AF081567.1 | - | 2 - 3 | 201 | 146 | ||
AK291843.1 | - | 2 - 3 | 202 | 146 | ||
BC110304.1 | - | 2 - 3 | 307 | 5222582 | 146 | |
BC117138.1 | - | 2 - 3 | 259 | 40125689 | 146 | |
BC117140.1 | - | 2 - 3 | 259 | 40125691 | 146 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC11002110.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
regulation of transcription from RNA polymerase II promoter
response to stress
signal transduction
negative regulation of cell proliferation
RNA polymerase II transcription factor activity, sequence-specific DNA binding
DNA binding
protein binding
metal ion binding
protein dimerization activity
regulation of transcription from RNA polymerase II promoter
response to stress
signal transduction
negative regulation of cell proliferation
RNA polymerase II transcription factor activity, sequence-specific DNA binding
DNA binding
protein binding
metal ion binding
protein dimerization activity
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1300006651.hg.1 | ENST00000431978.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006651.hg.1 | ENST00000431978.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006651.hg.1 | ENST00000431978.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11025231.hg.1 | ENST00000260045.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11025231.hg.1 | ENST00000529901.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11025231.hg.1 | ENST00000531878.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11025231.hg.1 | NM_004705.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR11025231.hg.1 | TC11002110.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC0100017934.hg.1 | ENST00000447914.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011684.hg.1 | ENST00000260045.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011684.hg.1 | ENST00000260045.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011684.hg.1 | ENST00000260045.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011684.hg.1 | ENST00000260045.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011684.hg.1 | ENST00000260045.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011684.hg.1 | ENST00000260045.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011684.hg.1 | ENST00000260045.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011684.hg.1 | ENST00000260045.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011684.hg.1 | ENST00000260045.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011684.hg.1 | ENST00000260045.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011684.hg.1 | ENST00000260045.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011684.hg.1 | ENST00000260045.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011684.hg.1 | ENST00000525277.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011684.hg.1 | ENST00000525277.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011684.hg.1 | ENST00000525277.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011684.hg.1 | ENST00000525277.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011684.hg.1 | ENST00000525277.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011684.hg.1 | ENST00000528993.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011684.hg.1 | ENST00000528993.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011684.hg.1 | ENST00000528993.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011684.hg.1 | ENST00000528993.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011684.hg.1 | ENST00000528993.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011684.hg.1 | ENST00000528993.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011684.hg.1 | ENST00000528993.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011684.hg.1 | ENST00000528993.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011684.hg.1 | ENST00000528993.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011684.hg.1 | ENST00000528993.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011684.hg.1 | ENST00000529901.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011684.hg.1 | ENST00000529901.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011684.hg.1 | ENST00000529901.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011684.hg.1 | ENST00000529901.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011684.hg.1 | ENST00000529901.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011684.hg.1 | ENST00000529901.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011684.hg.1 | ENST00000529901.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011684.hg.1 | ENST00000529901.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011684.hg.1 | ENST00000531878.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011684.hg.1 | ENST00000531878.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011684.hg.1 | ENST00000531878.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011684.hg.1 | ENST00000531878.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011684.hg.1 | ENST00000531878.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011684.hg.1 | ENST00000531878.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011684.hg.1 | NM_004705.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100011684.hg.1 | NM_004705.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100011684.hg.1 | NM_004705.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100011684.hg.1 | NM_004705.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100011684.hg.1 | NM_004705.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100011684.hg.1 | NM_004705.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100011684.hg.1 | NM_004705.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100011684.hg.1 | NM_004705.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100011684.hg.1 | NM_004705.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100011684.hg.1 | NM_004705.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100011684.hg.1 | NM_004705.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100011684.hg.1 | NM_004705.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100011684.hg.1 | NR_130898.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100011684.hg.1 | NR_130898.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100011684.hg.1 | NR_130898.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100011684.hg.1 | NR_130898.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100011684.hg.1 | NR_130898.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100011684.hg.1 | NR_130898.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100011684.hg.1 | NR_130898.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100011684.hg.1 | NR_130898.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100011684.hg.1 | NR_130898.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100011684.hg.1 | NR_130898.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100011684.hg.1 | NR_130898.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100011684.hg.1 | NR_130898.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100011684.hg.1 | NR_130898.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR11025232.hg.1 | ENST00000260045.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11025232.hg.1 | ENST00000529901.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11025232.hg.1 | ENST00000531878.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11025232.hg.1 | NM_004705.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR11025232.hg.1 | TC11002110.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1000007645.hg.1 | ENST00000456373.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007645.hg.1 | ENST00000456373.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007084.hg.1 | ENST00000528321.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007084.hg.1 | ENST00000528321.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007984.hg.1 | ENST00000489193.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007984.hg.1 | ENST00000489193.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100190152.hg.1 | ENST00000260045.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100190152.hg.1 | ENST00000525277.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100190152.hg.1 | ENST00000528993.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100190152.hg.1 | ENST00000531878.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100190152.hg.1 | NM_004705.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100190152.hg.1 | NR_130898.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100190152.hg.1 | ENST00000260045.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100190152.hg.1 | ENST00000525277.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100190152.hg.1 | ENST00000528993.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100190152.hg.1 | ENST00000531878.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100190152.hg.1 | NM_004705.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100190152.hg.1 | NR_130898.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100190152.hg.1 | TC1100011684.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1100011684.hg.1 | ENST00000260045.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011684.hg.1 | ENST00000260045.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011684.hg.1 | ENST00000260045.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011684.hg.1 | ENST00000260045.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011684.hg.1 | ENST00000260045.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011684.hg.1 | ENST00000260045.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011684.hg.1 | ENST00000260045.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011684.hg.1 | ENST00000260045.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011684.hg.1 | ENST00000260045.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011684.hg.1 | ENST00000260045.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011684.hg.1 | ENST00000260045.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011684.hg.1 | ENST00000260045.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011684.hg.1 | ENST00000525277.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011684.hg.1 | ENST00000525277.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011684.hg.1 | ENST00000525277.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011684.hg.1 | ENST00000525277.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011684.hg.1 | ENST00000525277.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011684.hg.1 | ENST00000528993.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011684.hg.1 | ENST00000528993.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011684.hg.1 | ENST00000528993.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011684.hg.1 | ENST00000528993.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011684.hg.1 | ENST00000528993.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011684.hg.1 | ENST00000528993.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011684.hg.1 | ENST00000528993.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011684.hg.1 | ENST00000528993.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011684.hg.1 | ENST00000528993.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011684.hg.1 | ENST00000528993.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011684.hg.1 | ENST00000529901.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011684.hg.1 | ENST00000529901.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011684.hg.1 | ENST00000529901.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011684.hg.1 | ENST00000529901.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011684.hg.1 | ENST00000529901.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011684.hg.1 | ENST00000529901.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011684.hg.1 | ENST00000529901.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011684.hg.1 | ENST00000529901.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011684.hg.1 | ENST00000531878.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011684.hg.1 | ENST00000531878.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011684.hg.1 | ENST00000531878.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011684.hg.1 | ENST00000531878.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011684.hg.1 | ENST00000531878.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011684.hg.1 | ENST00000531878.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011684.hg.1 | NM_004705.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100011684.hg.1 | NM_004705.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100011684.hg.1 | NM_004705.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100011684.hg.1 | NM_004705.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100011684.hg.1 | NM_004705.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100011684.hg.1 | NM_004705.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100011684.hg.1 | NM_004705.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100011684.hg.1 | NM_004705.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100011684.hg.1 | NM_004705.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100011684.hg.1 | NM_004705.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100011684.hg.1 | NM_004705.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100011684.hg.1 | NM_004705.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100011684.hg.1 | NR_130898.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100011684.hg.1 | NR_130898.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100011684.hg.1 | NR_130898.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100011684.hg.1 | NR_130898.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100011684.hg.1 | NR_130898.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100011684.hg.1 | NR_130898.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100011684.hg.1 | NR_130898.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100011684.hg.1 | NR_130898.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100011684.hg.1 | NR_130898.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100011684.hg.1 | NR_130898.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100011684.hg.1 | NR_130898.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100011684.hg.1 | NR_130898.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100011684.hg.1 | NR_130898.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300007984.hg.1 | ENST00000489193.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007984.hg.1 | ENST00000489193.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007645.hg.1 | ENST00000456373.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007645.hg.1 | ENST00000456373.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007084.hg.1 | ENST00000528321.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007084.hg.1 | ENST00000528321.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017934.hg.1 | ENST00000447914.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006651.hg.1 | ENST00000431978.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006651.hg.1 | ENST00000431978.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006651.hg.1 | ENST00000431978.1 | Ensembl |
