Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR20006797.hg.1 |
ENST00000370823.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20006797.hg.1 |
ENST00000536470.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20006797.hg.1 |
NM_015666.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20006797.hg.1 |
TC20000484.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007994.hg.1 |
ENST00000370823.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007994.hg.1 |
ENST00000370823.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007994.hg.1 |
ENST00000370823.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007994.hg.1 |
ENST00000370823.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007994.hg.1 |
ENST00000370823.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007994.hg.1 |
ENST00000370823.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007994.hg.1 |
ENST00000370823.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007994.hg.1 |
ENST00000370823.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007994.hg.1 |
ENST00000370823.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007994.hg.1 |
ENST00000370823.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007994.hg.1 |
ENST00000370823.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007994.hg.1 |
ENST00000370823.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007994.hg.1 |
ENST00000448254.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007994.hg.1 |
ENST00000448254.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007994.hg.1 |
ENST00000448254.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007994.hg.1 |
ENST00000448254.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007994.hg.1 |
ENST00000448254.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007994.hg.1 |
ENST00000448254.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007994.hg.1 |
ENST00000448254.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007994.hg.1 |
ENST00000461411.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007994.hg.1 |
ENST00000461411.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007994.hg.1 |
ENST00000461411.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007994.hg.1 |
ENST00000461411.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007994.hg.1 |
ENST00000466099.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007994.hg.1 |
ENST00000466099.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007994.hg.1 |
ENST00000466099.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007994.hg.1 |
ENST00000466099.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007994.hg.1 |
ENST00000466099.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007994.hg.1 |
ENST00000466099.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007994.hg.1 |
ENST00000466099.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007994.hg.1 |
ENST00000466933.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007994.hg.1 |
ENST00000466933.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007994.hg.1 |
ENST00000466933.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007994.hg.1 |
ENST00000466933.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007994.hg.1 |
ENST00000467101.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007994.hg.1 |
ENST00000467101.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007994.hg.1 |
ENST00000467101.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007994.hg.1 |
ENST00000467101.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007994.hg.1 |
ENST00000467101.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007994.hg.1 |
ENST00000467101.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007994.hg.1 |
ENST00000467101.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007994.hg.1 |
ENST00000467101.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007994.hg.1 |
ENST00000467101.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007994.hg.1 |
ENST00000467101.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007994.hg.1 |
ENST00000467101.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007994.hg.1 |
ENST00000471352.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007994.hg.1 |
ENST00000471352.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007994.hg.1 |
ENST00000471352.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007994.hg.1 |
ENST00000471352.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007994.hg.1 |
ENST00000471352.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007994.hg.1 |
ENST00000471352.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007994.hg.1 |
ENST00000471962.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007994.hg.1 |
ENST00000471962.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007994.hg.1 |
ENST00000471962.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007994.hg.1 |
ENST00000471962.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007994.hg.1 |
ENST00000471962.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007994.hg.1 |
ENST00000471962.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007994.hg.1 |
ENST00000472005.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007994.hg.1 |
ENST00000472005.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007994.hg.1 |
ENST00000472005.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007994.hg.1 |
ENST00000472005.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007994.hg.1 |
ENST00000472005.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007994.hg.1 |
ENST00000472005.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007994.hg.1 |
ENST00000488748.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007994.hg.1 |
ENST00000488748.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007994.hg.1 |
NM_015666.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007994.hg.1 |
NM_015666.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007994.hg.1 |
NM_015666.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007994.hg.1 |
NM_015666.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007994.hg.1 |
NM_015666.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007994.hg.1 |
NM_015666.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007994.hg.1 |
NM_015666.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007994.hg.1 |
NM_015666.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007994.hg.1 |
NM_015666.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007994.hg.1 |
NM_015666.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007994.hg.1 |
NM_015666.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007994.hg.1 |
NM_015666.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000155615.hg.1 |
ENST00000370823.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000155615.hg.1 |
ENST00000466099.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000155615.hg.1 |
ENST00000467101.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000155615.hg.1 |
ENST00000471962.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000155615.hg.1 |
ENST00000472005.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000155615.hg.1 |
NM_015666.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000155615.hg.1 |
TC2000007994.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000155615.hg.1 |
ENST00000370823.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000155615.hg.1 |
ENST00000466099.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000155615.hg.1 |
ENST00000467101.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000155615.hg.1 |
ENST00000471962.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000155615.hg.1 |
ENST00000472005.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000155615.hg.1 |
NM_015666.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007994.hg.1 |
ENST00000370823.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007994.hg.1 |
ENST00000370823.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007994.hg.1 |
ENST00000370823.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007994.hg.1 |
ENST00000370823.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007994.hg.1 |
ENST00000370823.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007994.hg.1 |
ENST00000370823.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007994.hg.1 |
ENST00000370823.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007994.hg.1 |
ENST00000370823.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007994.hg.1 |
ENST00000370823.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007994.hg.1 |
ENST00000370823.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007994.hg.1 |
ENST00000370823.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007994.hg.1 |
ENST00000370823.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007994.hg.1 |
ENST00000448254.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007994.hg.1 |
ENST00000448254.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007994.hg.1 |
ENST00000448254.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007994.hg.1 |
ENST00000448254.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007994.hg.1 |
ENST00000448254.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007994.hg.1 |
ENST00000448254.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007994.hg.1 |
ENST00000448254.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007994.hg.1 |
ENST00000461411.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007994.hg.1 |
ENST00000461411.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007994.hg.1 |
ENST00000461411.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007994.hg.1 |
ENST00000461411.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007994.hg.1 |
ENST00000466099.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007994.hg.1 |
ENST00000466099.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007994.hg.1 |
ENST00000466099.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007994.hg.1 |
ENST00000466099.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007994.hg.1 |
ENST00000466099.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007994.hg.1 |
ENST00000466099.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007994.hg.1 |
ENST00000466099.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007994.hg.1 |
ENST00000466933.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007994.hg.1 |
ENST00000466933.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007994.hg.1 |
ENST00000466933.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007994.hg.1 |
ENST00000466933.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007994.hg.1 |
ENST00000467101.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007994.hg.1 |
ENST00000467101.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007994.hg.1 |
ENST00000467101.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007994.hg.1 |
ENST00000467101.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007994.hg.1 |
ENST00000467101.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007994.hg.1 |
ENST00000467101.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007994.hg.1 |
ENST00000467101.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007994.hg.1 |
ENST00000467101.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007994.hg.1 |
ENST00000467101.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007994.hg.1 |
ENST00000467101.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007994.hg.1 |
ENST00000467101.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007994.hg.1 |
ENST00000471352.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007994.hg.1 |
ENST00000471352.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007994.hg.1 |
ENST00000471352.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007994.hg.1 |
ENST00000471352.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007994.hg.1 |
ENST00000471352.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007994.hg.1 |
ENST00000471352.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007994.hg.1 |
ENST00000471962.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007994.hg.1 |
ENST00000471962.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007994.hg.1 |
ENST00000471962.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007994.hg.1 |
ENST00000471962.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007994.hg.1 |
ENST00000471962.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007994.hg.1 |
ENST00000471962.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007994.hg.1 |
ENST00000472005.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007994.hg.1 |
ENST00000472005.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007994.hg.1 |
ENST00000472005.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007994.hg.1 |
ENST00000472005.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007994.hg.1 |
ENST00000472005.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007994.hg.1 |
ENST00000472005.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007994.hg.1 |
ENST00000488748.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007994.hg.1 |
ENST00000488748.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007994.hg.1 |
NM_015666.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007994.hg.1 |
NM_015666.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007994.hg.1 |
NM_015666.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007994.hg.1 |
NM_015666.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007994.hg.1 |
NM_015666.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007994.hg.1 |
NM_015666.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007994.hg.1 |
NM_015666.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007994.hg.1 |
NM_015666.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007994.hg.1 |
NM_015666.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007994.hg.1 |
NM_015666.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007994.hg.1 |
NM_015666.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007994.hg.1 |
NM_015666.3 |
Refseq |