Assay Details
Gene Symbol: RIOK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_003831.3 | NP_003822.2 | 3 - 4 | 949 | 95 | |
XM_011526242.1 | XP_011524544.1 | 3 - 4 | 949 | 95 | ||
XM_011526243.1 | XP_011524545.1 | 3 - 4 | 949 | 95 | ||
GenBank mRNA | AF013591.1 | - | 3 - 4 | 425 | 95 | |
AK057292.1 | - | 3 - 4 | 949 | 95 | ||
AK303938.1 | - | 3 - 4 | 437 | 95 | ||
BC039729.1 | - | 3 - 4 | 464 | 5745005 | 95 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC18000100.hg.1
Additional Information:
This assay is in the 5'UTR region of the XM_011526242.1,XM_011526243.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000339486.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000339486.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000339486.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000339486.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000339486.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000339486.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000339486.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000339486.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000339486.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000339486.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000339486.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000339486.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000339486.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000339486.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000339486.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000339486.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000339486.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000339486.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000339486.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000339486.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000339486.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000339486.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000339486.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000339486.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000577250.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000577250.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000577250.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000577501.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000577501.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000577501.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000577501.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000577501.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000577501.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000577501.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000577501.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000577501.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000577501.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000577501.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000577501.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000577501.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000577501.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000577501.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000577501.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000577501.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000577501.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000577501.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000577501.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000581220.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000581220.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000581220.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000581220.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000581220.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000581220.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000581220.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000581220.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000581302.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000581302.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000581302.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000581302.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000581302.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000581302.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000581302.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000581302.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000581302.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000581302.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000581302.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000581302.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000581302.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000581302.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000581302.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000581302.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000581302.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000581302.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000581302.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000581302.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000581302.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000581302.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000581302.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000581339.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000581339.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000581339.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000581339.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000581339.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000581339.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000581339.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000581339.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000581339.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000581585.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000581585.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000581585.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000581585.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000581585.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000581585.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000581585.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000581585.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000581585.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000581585.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000581585.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000581585.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000581585.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000581585.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000581585.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000581585.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000581585.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000581585.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000581585.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000581585.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000584052.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000584052.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000584052.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000584052.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000584052.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000584052.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000584052.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000584130.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000584130.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000584130.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000584130.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000584130.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000584130.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000584960.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000584960.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000584960.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000584960.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000584992.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000584992.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000584992.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000584992.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | ENST00000584992.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | NM_003831.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | NM_003831.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | NM_003831.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | NM_003831.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | NM_003831.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | NM_003831.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | NM_003831.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | NM_003831.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | NM_003831.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | NM_003831.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | NM_003831.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | NM_003831.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | NM_003831.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | NM_003831.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | NM_003831.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | NM_003831.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | NM_003831.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | NM_003831.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | NM_003831.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | NM_003831.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | NM_003831.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | NM_003831.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | NM_003831.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800006889.hg.1 | NM_003831.4 | Refseq |
