Assay Details
| Gene Symbol: MED12L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053002.5 | NP_443728.3 | 13 - 14 | 2094 | 97 | |
| XM_011512390.2 | XP_011510692.1 | 14 - 15 | 2366 | 97 | ||
| GenBank mRNA | AF388364.1 | - | 13 - 14 | 2498 | 97 | |
| AF399708.1 | - | 13 - 14 | 2003 | 97 | ||
| BC144559.1 | - | 11 - 12 | 1578 | 9053087 | 97 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC03000822.hg.1
Set Membership: |
|

MED12L
Genomic Map