Assay Details
Gene Symbol: CPB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001871.2 | NP_001862.2 | 8 - 9 | 797 | 86 | |
GenBank mRNA | AJ224866.1 | - | 8 - 9 | 779 | 86 | |
AK313940.1 | - | 8 - 9 | 785 | 86 | ||
BC015338.1 | - | 8 - 9 | 778 | 4422620 | 86 | |
BT009910.1 | - | 8 - 9 | 774 | 86 | ||
M81057.1 | - | 8 - 9 | 794 | 86 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC03000799.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0300009120.hg.1 | ENST00000282957.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009120.hg.1 | ENST00000282957.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009120.hg.1 | ENST00000282957.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009120.hg.1 | ENST00000282957.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009120.hg.1 | ENST00000282957.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009120.hg.1 | ENST00000282957.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009120.hg.1 | ENST00000282957.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009120.hg.1 | ENST00000282957.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009120.hg.1 | ENST00000282957.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009120.hg.1 | ENST00000282957.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009120.hg.1 | ENST00000282957.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009120.hg.1 | ENST00000282957.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009120.hg.1 | ENST00000282957.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009120.hg.1 | ENST00000282957.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009120.hg.1 | ENST00000282957.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009120.hg.1 | ENST00000462345.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009120.hg.1 | ENST00000462345.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009120.hg.1 | ENST00000462345.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009120.hg.1 | ENST00000462345.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009120.hg.1 | ENST00000462345.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009120.hg.1 | ENST00000462345.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009120.hg.1 | ENST00000462345.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009120.hg.1 | ENST00000465718.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009120.hg.1 | ENST00000465718.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009120.hg.1 | ENST00000465718.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009120.hg.1 | ENST00000465718.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009120.hg.1 | ENST00000465718.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009120.hg.1 | ENST00000465718.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009120.hg.1 | ENST00000465718.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009120.hg.1 | ENST00000465718.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009120.hg.1 | ENST00000468341.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009120.hg.1 | ENST00000468341.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009120.hg.1 | ENST00000468341.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009120.hg.1 | ENST00000468341.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009120.hg.1 | ENST00000468341.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009120.hg.1 | ENST00000468341.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009120.hg.1 | ENST00000468341.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009120.hg.1 | ENST00000468341.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009120.hg.1 | ENST00000468341.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009120.hg.1 | ENST00000473621.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009120.hg.1 | ENST00000473621.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009120.hg.1 | ENST00000473621.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009120.hg.1 | ENST00000473621.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009120.hg.1 | ENST00000473621.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009120.hg.1 | ENST00000473621.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009120.hg.1 | ENST00000476847.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009120.hg.1 | ENST00000476847.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009120.hg.1 | ENST00000476847.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009120.hg.1 | ENST00000484877.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009120.hg.1 | ENST00000484877.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009120.hg.1 | ENST00000484877.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009120.hg.1 | ENST00000484877.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009120.hg.1 | ENST00000484877.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009120.hg.1 | ENST00000491148.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009120.hg.1 | ENST00000491148.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009120.hg.1 | ENST00000491148.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009120.hg.1 | ENST00000491148.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009120.hg.1 | ENST00000491148.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009120.hg.1 | ENST00000491148.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009120.hg.1 | ENST00000491148.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009120.hg.1 | ENST00000491148.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009120.hg.1 | ENST00000491148.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009120.hg.1 | ENST00000491148.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009120.hg.1 | ENST00000491148.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009120.hg.1 | ENST00000491148.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009120.hg.1 | ENST00000491148.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009120.hg.1 | ENST00000491148.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009120.hg.1 | ENST00000491148.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009120.hg.1 | ENST00000491148.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009120.hg.1 | ENST00000491148.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009120.hg.1 | ENST00000494888.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009120.hg.1 | ENST00000494888.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009120.hg.1 | ENST00000494888.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009120.hg.1 | ENST00000494888.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009120.hg.1 | ENST00000494888.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009120.hg.1 | ENST00000498639.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009120.hg.1 | ENST00000498639.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009120.hg.1 | ENST00000498639.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009120.hg.1 | ENST00000498639.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009120.hg.1 | NM_001871.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009120.hg.1 | NM_001871.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009120.hg.1 | NM_001871.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009120.hg.1 | NM_001871.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009120.hg.1 | NM_001871.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009120.hg.1 | NM_001871.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009120.hg.1 | NM_001871.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009120.hg.1 | NM_001871.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009120.hg.1 | NM_001871.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009120.hg.1 | NM_001871.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009120.hg.1 | NM_001871.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009120.hg.1 | NM_001871.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009120.hg.1 | NM_001871.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009120.hg.1 | NM_001871.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009120.hg.1 | NM_001871.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR03014356.hg.1 | TC03000799.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR03014356.hg.1 | ENST00000282957.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR03014356.hg.1 | ENST00000491148.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR03014356.hg.1 | NM_001871.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300165371.hg.1 | ENST00000282957.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300165371.hg.1 | ENST00000491148.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300165371.hg.1 | NM_001871.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300165371.hg.1 | ENST00000282957.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300165371.hg.1 | ENST00000491148.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300165371.hg.1 | NM_001871.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300165371.hg.1 | TC0300009120.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0300009120.hg.1 | ENST00000282957.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009120.hg.1 | ENST00000282957.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009120.hg.1 | ENST00000282957.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009120.hg.1 | ENST00000282957.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009120.hg.1 | ENST00000282957.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009120.hg.1 | ENST00000282957.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009120.hg.1 | ENST00000282957.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009120.hg.1 | ENST00000282957.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009120.hg.1 | ENST00000282957.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009120.hg.1 | ENST00000282957.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009120.hg.1 | ENST00000282957.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009120.hg.1 | ENST00000282957.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009120.hg.1 | ENST00000282957.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009120.hg.1 | ENST00000282957.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009120.hg.1 | ENST00000282957.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009120.hg.1 | ENST00000462345.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009120.hg.1 | ENST00000462345.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009120.hg.1 | ENST00000462345.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009120.hg.1 | ENST00000462345.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009120.hg.1 | ENST00000462345.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009120.hg.1 | ENST00000462345.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009120.hg.1 | ENST00000462345.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009120.hg.1 | ENST00000465718.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009120.hg.1 | ENST00000465718.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009120.hg.1 | ENST00000465718.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009120.hg.1 | ENST00000465718.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009120.hg.1 | ENST00000465718.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009120.hg.1 | ENST00000465718.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009120.hg.1 | ENST00000465718.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009120.hg.1 | ENST00000465718.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009120.hg.1 | ENST00000468341.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009120.hg.1 | ENST00000468341.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009120.hg.1 | ENST00000468341.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009120.hg.1 | ENST00000468341.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009120.hg.1 | ENST00000468341.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009120.hg.1 | ENST00000468341.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009120.hg.1 | ENST00000468341.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009120.hg.1 | ENST00000468341.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009120.hg.1 | ENST00000468341.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009120.hg.1 | ENST00000473621.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009120.hg.1 | ENST00000473621.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009120.hg.1 | ENST00000473621.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009120.hg.1 | ENST00000473621.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009120.hg.1 | ENST00000473621.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009120.hg.1 | ENST00000473621.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009120.hg.1 | ENST00000476847.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009120.hg.1 | ENST00000476847.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009120.hg.1 | ENST00000476847.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009120.hg.1 | ENST00000484877.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009120.hg.1 | ENST00000484877.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009120.hg.1 | ENST00000484877.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009120.hg.1 | ENST00000484877.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009120.hg.1 | ENST00000484877.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009120.hg.1 | ENST00000491148.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009120.hg.1 | ENST00000491148.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009120.hg.1 | ENST00000491148.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009120.hg.1 | ENST00000491148.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009120.hg.1 | ENST00000491148.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009120.hg.1 | ENST00000491148.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009120.hg.1 | ENST00000491148.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009120.hg.1 | ENST00000491148.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009120.hg.1 | ENST00000491148.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009120.hg.1 | ENST00000491148.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009120.hg.1 | ENST00000491148.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009120.hg.1 | ENST00000491148.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009120.hg.1 | ENST00000491148.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009120.hg.1 | ENST00000491148.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009120.hg.1 | ENST00000491148.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009120.hg.1 | ENST00000491148.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009120.hg.1 | ENST00000491148.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009120.hg.1 | ENST00000494888.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009120.hg.1 | ENST00000494888.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009120.hg.1 | ENST00000494888.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009120.hg.1 | ENST00000494888.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009120.hg.1 | ENST00000494888.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009120.hg.1 | ENST00000498639.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009120.hg.1 | ENST00000498639.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009120.hg.1 | ENST00000498639.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009120.hg.1 | ENST00000498639.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009120.hg.1 | NM_001871.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009120.hg.1 | NM_001871.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009120.hg.1 | NM_001871.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009120.hg.1 | NM_001871.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009120.hg.1 | NM_001871.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009120.hg.1 | NM_001871.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009120.hg.1 | NM_001871.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009120.hg.1 | NM_001871.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009120.hg.1 | NM_001871.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009120.hg.1 | NM_001871.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009120.hg.1 | NM_001871.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009120.hg.1 | NM_001871.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009120.hg.1 | NM_001871.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009120.hg.1 | NM_001871.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009120.hg.1 | NM_001871.2 | Refseq |
