Assay Details
| Gene Symbol: FBXW4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001323541.1 | NP_001310470.1 | 1 - 2 | 56 | 68 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001323541.1 transcripts.
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1000011652.hg.1 | ENST00000331272.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011652.hg.1 | ENST00000331272.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011652.hg.1 | ENST00000331272.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011652.hg.1 | ENST00000331272.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011652.hg.1 | ENST00000331272.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011652.hg.1 | ENST00000331272.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011652.hg.1 | ENST00000331272.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011652.hg.1 | ENST00000331272.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011652.hg.1 | ENST00000331272.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011652.hg.1 | ENST00000331272.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011652.hg.1 | ENST00000331272.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011652.hg.1 | ENST00000331272.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011652.hg.1 | ENST00000331272.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011652.hg.1 | ENST00000331272.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011652.hg.1 | ENST00000331272.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011652.hg.1 | ENST00000331272.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011652.hg.1 | ENST00000457105.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011652.hg.1 | ENST00000457105.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011652.hg.1 | ENST00000457105.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011652.hg.1 | ENST00000457105.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011652.hg.1 | ENST00000457105.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011652.hg.1 | ENST00000457105.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011652.hg.1 | ENST00000457105.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011652.hg.1 | ENST00000470093.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011652.hg.1 | ENST00000470093.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011652.hg.1 | ENST00000470093.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011652.hg.1 | ENST00000470093.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011652.hg.1 | ENST00000470093.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011652.hg.1 | ENST00000470093.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011652.hg.1 | ENST00000470093.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011652.hg.1 | ENST00000482428.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011652.hg.1 | ENST00000482428.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011652.hg.1 | ENST00000482428.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011652.hg.1 | ENST00000489578.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011652.hg.1 | ENST00000489578.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011652.hg.1 | ENST00000489578.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011652.hg.1 | NM_022039.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011652.hg.1 | NM_022039.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011652.hg.1 | NM_022039.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011652.hg.1 | NM_022039.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011652.hg.1 | NM_022039.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011652.hg.1 | NM_022039.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011652.hg.1 | NM_022039.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011652.hg.1 | NM_022039.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011652.hg.1 | NM_022039.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011652.hg.1 | NM_022039.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011652.hg.1 | NM_022039.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011652.hg.1 | NM_022039.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011652.hg.1 | NM_022039.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011652.hg.1 | NM_022039.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011652.hg.1 | NM_022039.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011652.hg.1 | ENST00000331272.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011652.hg.1 | ENST00000331272.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011652.hg.1 | ENST00000331272.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011652.hg.1 | ENST00000331272.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011652.hg.1 | ENST00000331272.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011652.hg.1 | ENST00000331272.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011652.hg.1 | ENST00000331272.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011652.hg.1 | ENST00000331272.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011652.hg.1 | ENST00000331272.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011652.hg.1 | ENST00000331272.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011652.hg.1 | ENST00000331272.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011652.hg.1 | ENST00000331272.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011652.hg.1 | ENST00000331272.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011652.hg.1 | ENST00000331272.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011652.hg.1 | ENST00000331272.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011652.hg.1 | ENST00000331272.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011652.hg.1 | ENST00000457105.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011652.hg.1 | ENST00000457105.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011652.hg.1 | ENST00000457105.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011652.hg.1 | ENST00000457105.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011652.hg.1 | ENST00000457105.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011652.hg.1 | ENST00000457105.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011652.hg.1 | ENST00000457105.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011652.hg.1 | ENST00000470093.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011652.hg.1 | ENST00000470093.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011652.hg.1 | ENST00000470093.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011652.hg.1 | ENST00000470093.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011652.hg.1 | ENST00000470093.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011652.hg.1 | ENST00000470093.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011652.hg.1 | ENST00000470093.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011652.hg.1 | ENST00000482428.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011652.hg.1 | ENST00000482428.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011652.hg.1 | ENST00000482428.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011652.hg.1 | ENST00000489578.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011652.hg.1 | ENST00000489578.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011652.hg.1 | ENST00000489578.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011652.hg.1 | NM_022039.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011652.hg.1 | NM_022039.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011652.hg.1 | NM_022039.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011652.hg.1 | NM_022039.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011652.hg.1 | NM_022039.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011652.hg.1 | NM_022039.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011652.hg.1 | NM_022039.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011652.hg.1 | NM_022039.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011652.hg.1 | NM_022039.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011652.hg.1 | NM_022039.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011652.hg.1 | NM_022039.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011652.hg.1 | NM_022039.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011652.hg.1 | NM_022039.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011652.hg.1 | NM_022039.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011652.hg.1 | NM_022039.3 | Refseq |

FBXW4
Genomic Map