Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Clariom D Assay |
PSR1000159498.hg.1 |
ENST00000278070.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000159498.hg.1 |
ENST00000413464.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000159498.hg.1 |
NM_001288727.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000159498.hg.1 |
NM_001288728.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000159498.hg.1 |
NM_015062.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000159498.hg.1 |
ENST00000278070.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000159498.hg.1 |
ENST00000413464.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000159498.hg.1 |
NM_001288727.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000159498.hg.1 |
NM_001288728.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000159498.hg.1 |
NM_015062.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000159498.hg.1 |
TC1000008712.hg.1 |
Clariom D TC |