Assay Details
Gene Symbol: MRPS16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_016065.3 | NP_057149.1 | 1 - 2 | 223 | 135 | |
XM_017016305.1 | XP_016871794.1 | 1 - 2 | 218 | 135 | ||
GenBank mRNA | AB049948.1 | - | 1 - 2 | 109 | 135 | |
AF151890.1 | - | 1 - 2 | 176 | 133 | ||
AK025715.1 | - | 1 - 2 | 156 | 135 | ||
BC021106.1 | - | 1 - 2 | 118 | 5020046 | 135 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC10002964.hg.1
Additional Information:
This assay is in the 5'UTR region of the AB049948.1,AF151890.1,BC021106.1,NM_016065.3,XM_017016305.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0800008207.hg.1 | ENST00000523462.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10017675.hg.1 | TC10002964.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR10017675.hg.1 | ENST00000372940.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10017675.hg.1 | ENST00000372945.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10017675.hg.1 | ENST00000416782.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10017675.hg.1 | ENST00000479005.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10017675.hg.1 | NM_016065.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR10017670.hg.1 | TC10002964.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR10017670.hg.1 | ENST00000372940.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10017670.hg.1 | ENST00000372945.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10017670.hg.1 | ENST00000416782.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10017670.hg.1 | ENST00000479005.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10017670.hg.1 | NM_016065.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR10017673.hg.1 | TC10002964.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR10017673.hg.1 | ENST00000372940.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10017673.hg.1 | ENST00000372945.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10017673.hg.1 | ENST00000416782.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10017673.hg.1 | ENST00000479005.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10017673.hg.1 | NM_016065.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012566.hg.1 | ENST00000372940.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012566.hg.1 | ENST00000372940.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012566.hg.1 | ENST00000372940.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012566.hg.1 | ENST00000372940.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012566.hg.1 | ENST00000372940.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012566.hg.1 | ENST00000372940.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012566.hg.1 | ENST00000372945.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012566.hg.1 | ENST00000372945.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012566.hg.1 | ENST00000372945.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012566.hg.1 | ENST00000372945.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012566.hg.1 | ENST00000372945.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012566.hg.1 | ENST00000372945.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012566.hg.1 | ENST00000372945.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012566.hg.1 | ENST00000372945.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012566.hg.1 | ENST00000372945.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012566.hg.1 | ENST00000372945.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012566.hg.1 | ENST00000471251.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012566.hg.1 | ENST00000471251.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012566.hg.1 | ENST00000471251.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012566.hg.1 | ENST00000471251.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012566.hg.1 | ENST00000473427.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012566.hg.1 | ENST00000473427.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012566.hg.1 | ENST00000473427.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012566.hg.1 | ENST00000473427.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012566.hg.1 | ENST00000473427.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012566.hg.1 | ENST00000479005.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012566.hg.1 | ENST00000479005.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012566.hg.1 | ENST00000479005.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012566.hg.1 | ENST00000479005.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012566.hg.1 | ENST00000479005.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012566.hg.1 | ENST00000479005.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012566.hg.1 | ENST00000479005.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012566.hg.1 | ENST00000479005.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012566.hg.1 | ENST00000479005.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012566.hg.1 | NM_016065.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012566.hg.1 | NM_016065.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012566.hg.1 | NM_016065.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012566.hg.1 | NM_016065.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012566.hg.1 | NM_016065.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012566.hg.1 | NM_016065.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012566.hg.1 | NM_016065.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012566.hg.1 | NM_016065.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012566.hg.1 | NM_016065.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012566.hg.1 | NM_016065.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR10017678.hg.1 | TC10002964.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR10017678.hg.1 | ENST00000372940.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10017678.hg.1 | ENST00000372945.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10017678.hg.1 | NM_016065.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000007926.hg.1 | ENST00000445106.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10017674.hg.1 | TC10002964.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR10017674.hg.1 | ENST00000372940.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10017674.hg.1 | ENST00000372945.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10017674.hg.1 | ENST00000416782.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10017674.hg.1 | ENST00000479005.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10017674.hg.1 | NM_016065.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008207.hg.1 | ENST00000523462.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000187335.hg.1 | ENST00000372940.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000187335.hg.1 | ENST00000372945.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000187335.hg.1 | ENST00000473427.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000187335.hg.1 | ENST00000479005.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000187335.hg.1 | NM_016065.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000187335.hg.1 | ENST00000372940.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000187335.hg.1 | ENST00000372945.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000187335.hg.1 | ENST00000473427.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000187335.hg.1 | ENST00000479005.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000187335.hg.1 | NM_016065.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000187335.hg.1 | TC1000012566.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1000187338.hg.1 | ENST00000372940.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000187338.hg.1 | ENST00000372945.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000187338.hg.1 | NM_016065.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000187338.hg.1 | ENST00000372940.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000187338.hg.1 | ENST00000372945.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000187338.hg.1 | NM_016065.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000187338.hg.1 | TC1000012566.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1000187334.hg.1 | ENST00000372940.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000187334.hg.1 | ENST00000372945.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000187334.hg.1 | ENST00000479005.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000187334.hg.1 | NM_016065.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000187334.hg.1 | ENST00000372940.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000187334.hg.1 | ENST00000372945.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000187334.hg.1 | ENST00000479005.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000187334.hg.1 | NM_016065.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000187334.hg.1 | TC1000012566.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1000187336.hg.1 | ENST00000372940.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000187336.hg.1 | ENST00000372945.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000187336.hg.1 | ENST00000473427.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000187336.hg.1 | ENST00000479005.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000187336.hg.1 | NM_016065.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000187336.hg.1 | ENST00000372940.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000187336.hg.1 | ENST00000372945.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000187336.hg.1 | ENST00000473427.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000187336.hg.1 | ENST00000479005.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000187336.hg.1 | NM_016065.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000187336.hg.1 | TC1000012566.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1000187330.hg.1 | ENST00000372940.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000187330.hg.1 | ENST00000372945.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000187330.hg.1 | ENST00000471251.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000187330.hg.1 | ENST00000473427.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000187330.hg.1 | ENST00000479005.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000187330.hg.1 | NM_016065.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000187330.hg.1 | ENST00000372940.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000187330.hg.1 | ENST00000372945.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000187330.hg.1 | ENST00000471251.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000187330.hg.1 | ENST00000473427.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000187330.hg.1 | ENST00000479005.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000187330.hg.1 | NM_016065.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000187330.hg.1 | TC1000012566.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC2000007926.hg.1 | ENST00000445106.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012566.hg.1 | ENST00000372940.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012566.hg.1 | ENST00000372940.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012566.hg.1 | ENST00000372940.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012566.hg.1 | ENST00000372940.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012566.hg.1 | ENST00000372940.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012566.hg.1 | ENST00000372940.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012566.hg.1 | ENST00000372945.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012566.hg.1 | ENST00000372945.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012566.hg.1 | ENST00000372945.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012566.hg.1 | ENST00000372945.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012566.hg.1 | ENST00000372945.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012566.hg.1 | ENST00000372945.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012566.hg.1 | ENST00000372945.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012566.hg.1 | ENST00000372945.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012566.hg.1 | ENST00000372945.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012566.hg.1 | ENST00000372945.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012566.hg.1 | ENST00000471251.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012566.hg.1 | ENST00000471251.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012566.hg.1 | ENST00000471251.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012566.hg.1 | ENST00000471251.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012566.hg.1 | ENST00000473427.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012566.hg.1 | ENST00000473427.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012566.hg.1 | ENST00000473427.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012566.hg.1 | ENST00000473427.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012566.hg.1 | ENST00000473427.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012566.hg.1 | ENST00000479005.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012566.hg.1 | ENST00000479005.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012566.hg.1 | ENST00000479005.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012566.hg.1 | ENST00000479005.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012566.hg.1 | ENST00000479005.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012566.hg.1 | ENST00000479005.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012566.hg.1 | ENST00000479005.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012566.hg.1 | ENST00000479005.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012566.hg.1 | ENST00000479005.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012566.hg.1 | NM_016065.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012566.hg.1 | NM_016065.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012566.hg.1 | NM_016065.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012566.hg.1 | NM_016065.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012566.hg.1 | NM_016065.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012566.hg.1 | NM_016065.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012566.hg.1 | NM_016065.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012566.hg.1 | NM_016065.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012566.hg.1 | NM_016065.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012566.hg.1 | NM_016065.3 | Refseq |
