Assay Details
| Gene Symbol: ATP10D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK027598.1 | - | 4 - 5 | 608 | 67 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK027598.1 transcripts.
Set Membership: |
|

ATP10D
Genomic Map