Assay Details
Gene Symbol: NAAA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_017008028.1 | XP_016863517.1 | 10 - 11 | 1134 | 144 | |
XM_017008030.1 | XP_016863519.1 | 9 - 10 | 1105 | 144 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0400012933.hg.1 | ENST00000286733.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012933.hg.1 | ENST00000286733.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012933.hg.1 | ENST00000286733.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012933.hg.1 | ENST00000286733.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012933.hg.1 | ENST00000286733.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012933.hg.1 | ENST00000286733.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012933.hg.1 | ENST00000286733.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012933.hg.1 | ENST00000286733.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012933.hg.1 | ENST00000286733.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012933.hg.1 | ENST00000286733.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012933.hg.1 | ENST00000286733.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012933.hg.1 | ENST00000286733.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012933.hg.1 | ENST00000286733.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012933.hg.1 | ENST00000286733.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012933.hg.1 | ENST00000286733.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012933.hg.1 | ENST00000286733.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012933.hg.1 | ENST00000286733.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012933.hg.1 | ENST00000503636.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012933.hg.1 | ENST00000503636.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012933.hg.1 | ENST00000503636.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012933.hg.1 | ENST00000505594.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012933.hg.1 | ENST00000505594.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012933.hg.1 | ENST00000505594.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012933.hg.1 | ENST00000505594.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012933.hg.1 | ENST00000505594.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012933.hg.1 | ENST00000505594.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012933.hg.1 | ENST00000505594.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012933.hg.1 | ENST00000505594.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012933.hg.1 | ENST00000505594.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012933.hg.1 | ENST00000505594.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012933.hg.1 | ENST00000505594.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012933.hg.1 | ENST00000505594.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012933.hg.1 | ENST00000507187.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012933.hg.1 | ENST00000507187.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012933.hg.1 | ENST00000507187.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012933.hg.1 | ENST00000507187.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012933.hg.1 | ENST00000507187.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012933.hg.1 | ENST00000507940.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012933.hg.1 | ENST00000507940.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012933.hg.1 | ENST00000507940.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012933.hg.1 | ENST00000507940.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012933.hg.1 | ENST00000507940.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012933.hg.1 | ENST00000507956.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012933.hg.1 | ENST00000507956.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012933.hg.1 | ENST00000507956.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012933.hg.1 | ENST00000507956.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012933.hg.1 | ENST00000507956.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012933.hg.1 | ENST00000507956.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012933.hg.1 | ENST00000507956.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012933.hg.1 | ENST00000507956.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012933.hg.1 | ENST00000507956.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012933.hg.1 | ENST00000507956.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012933.hg.1 | ENST00000507956.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012933.hg.1 | ENST00000511606.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012933.hg.1 | ENST00000511606.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012933.hg.1 | ENST00000511606.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012933.hg.1 | ENST00000511606.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012933.hg.1 | ENST00000511606.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012933.hg.1 | ENST00000513045.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012933.hg.1 | ENST00000513045.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012933.hg.1 | ENST00000513045.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012933.hg.1 | ENST00000513045.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012933.hg.1 | ENST00000513045.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012933.hg.1 | ENST00000513045.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012933.hg.1 | ENST00000513045.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012933.hg.1 | ENST00000513045.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012933.hg.1 | ENST00000602782.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012933.hg.1 | ENST00000602782.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012933.hg.1 | ENST00000602782.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012933.hg.1 | ENST00000602782.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012933.hg.1 | ENST00000602782.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012933.hg.1 | ENST00000602782.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012933.hg.1 | ENST00000602782.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012933.hg.1 | ENST00000602782.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012933.hg.1 | ENST00000602782.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012933.hg.1 | ENST00000602782.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012933.hg.1 | NM_001042402.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012933.hg.1 | NM_001042402.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012933.hg.1 | NM_001042402.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012933.hg.1 | NM_001042402.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012933.hg.1 | NM_001042402.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012933.hg.1 | NM_001042402.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012933.hg.1 | NM_001042402.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012933.hg.1 | NM_001042402.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012933.hg.1 | NM_001042402.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012933.hg.1 | NM_001042402.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012933.hg.1 | NM_001042402.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012933.hg.1 | NM_014435.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012933.hg.1 | NM_014435.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012933.hg.1 | NM_014435.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012933.hg.1 | NM_014435.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012933.hg.1 | NM_014435.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012933.hg.1 | NM_014435.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012933.hg.1 | NM_014435.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012933.hg.1 | NM_014435.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012933.hg.1 | NM_014435.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012933.hg.1 | NM_014435.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012933.hg.1 | NM_014435.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012933.hg.1 | NM_014435.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012933.hg.1 | NM_014435.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012933.hg.1 | NM_014435.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012933.hg.1 | NM_014435.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012933.hg.1 | NM_014435.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012933.hg.1 | ENST00000286733.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012933.hg.1 | ENST00000286733.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012933.hg.1 | ENST00000286733.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012933.hg.1 | ENST00000286733.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012933.hg.1 | ENST00000286733.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012933.hg.1 | ENST00000286733.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012933.hg.1 | ENST00000286733.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012933.hg.1 | ENST00000286733.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012933.hg.1 | ENST00000286733.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012933.hg.1 | ENST00000286733.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012933.hg.1 | ENST00000286733.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012933.hg.1 | ENST00000286733.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012933.hg.1 | ENST00000286733.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012933.hg.1 | ENST00000286733.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012933.hg.1 | ENST00000286733.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012933.hg.1 | ENST00000286733.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012933.hg.1 | ENST00000286733.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012933.hg.1 | ENST00000503636.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012933.hg.1 | ENST00000503636.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012933.hg.1 | ENST00000503636.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012933.hg.1 | ENST00000505594.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012933.hg.1 | ENST00000505594.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012933.hg.1 | ENST00000505594.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012933.hg.1 | ENST00000505594.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012933.hg.1 | ENST00000505594.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012933.hg.1 | ENST00000505594.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012933.hg.1 | ENST00000505594.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012933.hg.1 | ENST00000505594.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012933.hg.1 | ENST00000505594.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012933.hg.1 | ENST00000505594.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012933.hg.1 | ENST00000505594.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012933.hg.1 | ENST00000505594.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012933.hg.1 | ENST00000507187.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012933.hg.1 | ENST00000507187.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012933.hg.1 | ENST00000507187.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012933.hg.1 | ENST00000507187.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012933.hg.1 | ENST00000507187.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012933.hg.1 | ENST00000507940.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012933.hg.1 | ENST00000507940.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012933.hg.1 | ENST00000507940.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012933.hg.1 | ENST00000507940.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012933.hg.1 | ENST00000507940.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012933.hg.1 | ENST00000507956.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012933.hg.1 | ENST00000507956.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012933.hg.1 | ENST00000507956.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012933.hg.1 | ENST00000507956.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012933.hg.1 | ENST00000507956.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012933.hg.1 | ENST00000507956.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012933.hg.1 | ENST00000507956.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012933.hg.1 | ENST00000507956.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012933.hg.1 | ENST00000507956.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012933.hg.1 | ENST00000507956.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012933.hg.1 | ENST00000507956.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012933.hg.1 | ENST00000511606.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012933.hg.1 | ENST00000511606.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012933.hg.1 | ENST00000511606.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012933.hg.1 | ENST00000511606.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012933.hg.1 | ENST00000511606.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012933.hg.1 | ENST00000513045.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012933.hg.1 | ENST00000513045.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012933.hg.1 | ENST00000513045.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012933.hg.1 | ENST00000513045.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012933.hg.1 | ENST00000513045.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012933.hg.1 | ENST00000513045.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012933.hg.1 | ENST00000513045.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012933.hg.1 | ENST00000513045.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012933.hg.1 | ENST00000602782.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012933.hg.1 | ENST00000602782.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012933.hg.1 | ENST00000602782.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012933.hg.1 | ENST00000602782.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012933.hg.1 | ENST00000602782.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012933.hg.1 | ENST00000602782.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012933.hg.1 | ENST00000602782.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012933.hg.1 | ENST00000602782.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012933.hg.1 | ENST00000602782.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012933.hg.1 | ENST00000602782.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012933.hg.1 | NM_001042402.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012933.hg.1 | NM_001042402.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012933.hg.1 | NM_001042402.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012933.hg.1 | NM_001042402.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012933.hg.1 | NM_001042402.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012933.hg.1 | NM_001042402.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012933.hg.1 | NM_001042402.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012933.hg.1 | NM_001042402.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012933.hg.1 | NM_001042402.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012933.hg.1 | NM_001042402.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012933.hg.1 | NM_001042402.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012933.hg.1 | NM_014435.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012933.hg.1 | NM_014435.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012933.hg.1 | NM_014435.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012933.hg.1 | NM_014435.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012933.hg.1 | NM_014435.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012933.hg.1 | NM_014435.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012933.hg.1 | NM_014435.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012933.hg.1 | NM_014435.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012933.hg.1 | NM_014435.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012933.hg.1 | NM_014435.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012933.hg.1 | NM_014435.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012933.hg.1 | NM_014435.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012933.hg.1 | NM_014435.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012933.hg.1 | NM_014435.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012933.hg.1 | NM_014435.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012933.hg.1 | NM_014435.3 | Refseq |
