Assay Details
| Gene Symbol: C6orf203 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001142468.2 | NP_001135940.1 | 3 - 4 | 1182 | 104 | |
| NM_001142470.2 | NP_001135942.1 | 4 - 5 | 1265 | 104 | ||
| NM_016487.4 | NP_057571.1 | 3 - 4 | 656 | 104 | ||
| XM_011535876.2 | XP_011534178.1 | 4 - 5 | 760 | 104 | ||
| XM_011535877.2 | XP_011534179.1 | 4 - 5 | 708 | 104 | ||
| XM_017010919.1 | XP_016866408.1 | 3 - 4 | 655 | 104 | ||
| XM_017010920.1 | XP_016866409.1 | 3 - 4 | 1148 | 104 | ||
| GenBank mRNA | AF151064.1 | - | 3 - 4 | 612 | 104 | |
| AI193526.1 | - | 1 - 2 | 104 | |||
| AK091564.1 | - | 3 - 4 | 1141 | 104 | ||
| BC010899.2 | - | 2 - 3 | 558 | 4078296 | 104 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

C6orf203
Genomic Map