Assay Details
| Gene Symbol: LOC114224 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF130079.1 | - | 1 | 1176 | 68 | |
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
|---|---|---|---|---|---|---|---|
| GenBankmRNA | BX095073.1 | - | 1 | Chr. 9 - 34187327 - 34188126 [-] on Build GRCh38 | 68 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | ubiquitin associated protein 1 | UBAP1 |
NAG20, UAP, UBAP, UBAP-1 |
Sense | Chr. 9 - 34179005 - 34252523 on NCBI Build GRCh38 | |
| Downstream | kinesin family member 24 | KIF24 |
C9orf48, bA571F15.4 |
64.4kb | Anti-sense | Chr. 9 - 34252380 - 34332972 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

LOC114224
Genomic Map