Assay Details
| Gene Symbol: FNDC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_032532.2 | NP_115921.2 | 14 - 15 | 4818 | 148 | |
| XM_011536190.2 | XP_011534492.1 | 13 - 14 | 4741 | 148 | ||
| XM_011536191.2 | XP_011534493.1 | 11 - 12 | 4459 | 148 | ||
| GenBank mRNA | AB058769.2 | - | 12 - 13 | 4286 | 148 | |
| AJ586132.1 | - | 4 - 5 | 719 | 148 | ||
| AK127635.1 | - | 4 - 5 | 808 | 148 | ||
| AL832410.1 | - | 4 - 5 | 1017 | 148 | ||
| BC146783.1 | - | 12 - 13 | 4286 | 9007239 | 148 | |
| BC150607.1 | - | 13 - 14 | 4493 | 9056977 | 148 | |
| DQ009660.1 | - | 14 - 15 | 4800 | 148 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC06001142.hg.1
Additional Information:
This assay is in the 5'UTR region of the AJ586132.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | ENST00000297267.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | ENST00000297267.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | ENST00000297267.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | ENST00000297267.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | ENST00000297267.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | ENST00000297267.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | ENST00000297267.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | ENST00000297267.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | ENST00000297267.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | ENST00000297267.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | ENST00000297267.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | ENST00000297267.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | ENST00000297267.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | ENST00000297267.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | ENST00000297267.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | ENST00000297267.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | ENST00000297267.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | ENST00000297267.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | ENST00000297267.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | ENST00000297267.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | ENST00000297267.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | ENST00000297267.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | ENST00000297267.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | ENST00000297267.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | ENST00000297267.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | ENST00000297267.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | ENST00000297267.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | ENST00000329629.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | ENST00000329629.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | ENST00000329629.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | ENST00000329629.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | ENST00000329629.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | ENST00000329629.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | ENST00000329629.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | ENST00000329629.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | ENST00000329629.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | ENST00000329629.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | ENST00000329629.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | ENST00000329629.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | ENST00000329629.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | ENST00000329629.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | ENST00000329629.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | ENST00000329629.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | ENST00000329629.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | ENST00000329629.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | ENST00000329629.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | ENST00000329629.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | ENST00000329629.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | ENST00000329629.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | ENST00000329629.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | ENST00000329629.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | ENST00000480856.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | ENST00000480856.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | ENST00000480856.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | ENST00000480856.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | ENST00000480856.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | NM_032532.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | NM_032532.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | NM_032532.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | NM_032532.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | NM_032532.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | NM_032532.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | NM_032532.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | NM_032532.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | NM_032532.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | NM_032532.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | NM_032532.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | NM_032532.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | NM_032532.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | NM_032532.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | NM_032532.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | NM_032532.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | NM_032532.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | NM_032532.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | NM_032532.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | NM_032532.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | NM_032532.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | NM_032532.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | NM_032532.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | NM_032532.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | NM_032532.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | NM_032532.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | NM_032532.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06013590.hg.1 | TC06001142.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06013590.hg.1 | ENST00000297267.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06013590.hg.1 | ENST00000340366.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06013590.hg.1 | NM_032532.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06013589.hg.1 | TC06001142.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06013589.hg.1 | ENST00000297267.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06013589.hg.1 | ENST00000340366.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06013589.hg.1 | NM_032532.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600168583.hg.1 | ENST00000297267.13 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600168583.hg.1 | ENST00000329629.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600168583.hg.1 | NM_032532.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600168583.hg.1 | ENST00000297267.13 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600168583.hg.1 | ENST00000329629.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600168583.hg.1 | NM_032532.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600168583.hg.1 | TC0600010040.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | ENST00000297267.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | ENST00000297267.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | ENST00000297267.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | ENST00000297267.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | ENST00000297267.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | ENST00000297267.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | ENST00000297267.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | ENST00000297267.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | ENST00000297267.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | ENST00000297267.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | ENST00000297267.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | ENST00000297267.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | ENST00000297267.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | ENST00000297267.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | ENST00000297267.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | ENST00000297267.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | ENST00000297267.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | ENST00000297267.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | ENST00000297267.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | ENST00000297267.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | ENST00000297267.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | ENST00000297267.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | ENST00000297267.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | ENST00000297267.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | ENST00000297267.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | ENST00000297267.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | ENST00000297267.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | ENST00000329629.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | ENST00000329629.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | ENST00000329629.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | ENST00000329629.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | ENST00000329629.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | ENST00000329629.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | ENST00000329629.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | ENST00000329629.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | ENST00000329629.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | ENST00000329629.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | ENST00000329629.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | ENST00000329629.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | ENST00000329629.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | ENST00000329629.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | ENST00000329629.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | ENST00000329629.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | ENST00000329629.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | ENST00000329629.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | ENST00000329629.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | ENST00000329629.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | ENST00000329629.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | ENST00000329629.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | ENST00000329629.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | ENST00000329629.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | ENST00000480856.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | ENST00000480856.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | ENST00000480856.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | ENST00000480856.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | ENST00000480856.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | NM_032532.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | NM_032532.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | NM_032532.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | NM_032532.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | NM_032532.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | NM_032532.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | NM_032532.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | NM_032532.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | NM_032532.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | NM_032532.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | NM_032532.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | NM_032532.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | NM_032532.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | NM_032532.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | NM_032532.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | NM_032532.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | NM_032532.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | NM_032532.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | NM_032532.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | NM_032532.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | NM_032532.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | NM_032532.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | NM_032532.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | NM_032532.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | NM_032532.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | NM_032532.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | NM_032532.2 | Refseq |

FNDC1
Genomic Map