Assay Details
| Gene Symbol: FNDC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_032532.2 | NP_115921.2 | 17 - 18 | 5262 | 115 | |
| XM_011536190.2 | XP_011534492.1 | 16 - 17 | 5185 | 115 | ||
| XM_011536191.2 | XP_011534493.1 | 14 - 15 | 4903 | 115 | ||
| GenBank mRNA | AB058769.2 | - | 15 - 16 | 4730 | 115 | |
| AJ586132.1 | - | 7 - 8 | 115 | |||
| AK127635.1 | - | 7 - 8 | 1252 | 115 | ||
| AL832410.1 | - | 7 - 8 | 1461 | 115 | ||
| BC146783.1 | - | 15 - 16 | 4730 | 9007239 | 115 | |
| BC150607.1 | - | 16 - 17 | 4937 | 9056977 | 115 | |
| DQ009660.1 | - | 17 - 18 | 5244 | 115 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC06001142.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR06013593.hg.1 | TC06001142.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06013593.hg.1 | ENST00000297267.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06013593.hg.1 | ENST00000340366.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06013593.hg.1 | NM_032532.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | ENST00000297267.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | ENST00000297267.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | ENST00000297267.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | ENST00000297267.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | ENST00000297267.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | ENST00000297267.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | ENST00000297267.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | ENST00000297267.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | ENST00000297267.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | ENST00000297267.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | ENST00000297267.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | ENST00000297267.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | ENST00000297267.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | ENST00000297267.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | ENST00000297267.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | ENST00000297267.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | ENST00000297267.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | ENST00000297267.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | ENST00000297267.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | ENST00000297267.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | ENST00000297267.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | ENST00000297267.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | ENST00000297267.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | ENST00000297267.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | ENST00000297267.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | ENST00000297267.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | ENST00000297267.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | ENST00000329629.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | ENST00000329629.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | ENST00000329629.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | ENST00000329629.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | ENST00000329629.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | ENST00000329629.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | ENST00000329629.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | ENST00000329629.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | ENST00000329629.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | ENST00000329629.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | ENST00000329629.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | ENST00000329629.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | ENST00000329629.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | ENST00000329629.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | ENST00000329629.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | ENST00000329629.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | ENST00000329629.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | ENST00000329629.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | ENST00000329629.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | ENST00000329629.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | ENST00000329629.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | ENST00000329629.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | ENST00000329629.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | ENST00000329629.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | ENST00000480856.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | ENST00000480856.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | ENST00000480856.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | ENST00000480856.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | ENST00000480856.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | NM_032532.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | NM_032532.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | NM_032532.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | NM_032532.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | NM_032532.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | NM_032532.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | NM_032532.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | NM_032532.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | NM_032532.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | NM_032532.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | NM_032532.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | NM_032532.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | NM_032532.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | NM_032532.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | NM_032532.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | NM_032532.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | NM_032532.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | NM_032532.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | NM_032532.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | NM_032532.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | NM_032532.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | NM_032532.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | NM_032532.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | NM_032532.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | NM_032532.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | NM_032532.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | NM_032532.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | ENST00000297267.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | ENST00000297267.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | ENST00000297267.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | ENST00000297267.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | ENST00000297267.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | ENST00000297267.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | ENST00000297267.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | ENST00000297267.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | ENST00000297267.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | ENST00000297267.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | ENST00000297267.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | ENST00000297267.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | ENST00000297267.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | ENST00000297267.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | ENST00000297267.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | ENST00000297267.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | ENST00000297267.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | ENST00000297267.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | ENST00000297267.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | ENST00000297267.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | ENST00000297267.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | ENST00000297267.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | ENST00000297267.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | ENST00000297267.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | ENST00000297267.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | ENST00000297267.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | ENST00000297267.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | ENST00000329629.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | ENST00000329629.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | ENST00000329629.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | ENST00000329629.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | ENST00000329629.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | ENST00000329629.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | ENST00000329629.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | ENST00000329629.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | ENST00000329629.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | ENST00000329629.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | ENST00000329629.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | ENST00000329629.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | ENST00000329629.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | ENST00000329629.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | ENST00000329629.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | ENST00000329629.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | ENST00000329629.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | ENST00000329629.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | ENST00000329629.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | ENST00000329629.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | ENST00000329629.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | ENST00000329629.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | ENST00000329629.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | ENST00000329629.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | ENST00000480856.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | ENST00000480856.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | ENST00000480856.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | ENST00000480856.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | ENST00000480856.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | NM_032532.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | NM_032532.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | NM_032532.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | NM_032532.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | NM_032532.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | NM_032532.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | NM_032532.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | NM_032532.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | NM_032532.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | NM_032532.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | NM_032532.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | NM_032532.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | NM_032532.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | NM_032532.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | NM_032532.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | NM_032532.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | NM_032532.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | NM_032532.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | NM_032532.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | NM_032532.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | NM_032532.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | NM_032532.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | NM_032532.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | NM_032532.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | NM_032532.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | NM_032532.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | NM_032532.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600168586.hg.1 | ENST00000297267.13 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600168586.hg.1 | ENST00000329629.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600168586.hg.1 | NM_032532.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600168586.hg.1 | ENST00000297267.13 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600168586.hg.1 | ENST00000329629.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600168586.hg.1 | NM_032532.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600168586.hg.1 | TC0600010040.hg.1 | Clariom D TC |

FNDC1
Genomic Map