Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR13009616.hg.1 |
ENST00000448407.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13009616.hg.1 |
NR_040247.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13009616.hg.1 |
TC13000857.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009701.hg.1 |
ENST00000448407.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009701.hg.1 |
ENST00000448407.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009701.hg.1 |
ENST00000448407.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009701.hg.1 |
ENST00000448407.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009701.hg.1 |
ENST00000448407.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009701.hg.1 |
ENST00000448407.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009701.hg.1 |
ENST00000448407.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009701.hg.1 |
NR_040247.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009701.hg.1 |
NR_040247.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009701.hg.1 |
NR_040247.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009701.hg.1 |
NR_040247.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009701.hg.1 |
NR_040247.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009701.hg.1 |
NR_040247.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009701.hg.1 |
NR_040247.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13009615.hg.1 |
ENST00000448407.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13009615.hg.1 |
NR_040247.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13009615.hg.1 |
TC13000857.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR13009613.hg.1 |
ENST00000448407.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13009613.hg.1 |
NR_040247.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13009613.hg.1 |
TC13000857.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR13009614.hg.1 |
ENST00000448407.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13009614.hg.1 |
NR_040247.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13009614.hg.1 |
TC13000857.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1300164797.hg.1 |
ENST00000448407.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300164797.hg.1 |
NR_040247.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300164797.hg.1 |
ENST00000448407.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300164797.hg.1 |
NR_040247.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300164797.hg.1 |
TC1300009701.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1300164796.hg.1 |
ENST00000448407.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300164796.hg.1 |
NR_040247.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300164796.hg.1 |
ENST00000448407.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300164796.hg.1 |
NR_040247.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300164796.hg.1 |
TC1300009701.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1300164795.hg.1 |
ENST00000448407.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300164795.hg.1 |
NR_040247.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300164795.hg.1 |
ENST00000448407.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300164795.hg.1 |
NR_040247.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300164795.hg.1 |
TC1300009701.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1300009701.hg.1 |
ENST00000448407.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009701.hg.1 |
ENST00000448407.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009701.hg.1 |
ENST00000448407.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009701.hg.1 |
ENST00000448407.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009701.hg.1 |
ENST00000448407.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009701.hg.1 |
ENST00000448407.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009701.hg.1 |
ENST00000448407.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009701.hg.1 |
NR_040247.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009701.hg.1 |
NR_040247.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009701.hg.1 |
NR_040247.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009701.hg.1 |
NR_040247.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009701.hg.1 |
NR_040247.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009701.hg.1 |
NR_040247.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009701.hg.1 |
NR_040247.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300164798.hg.1 |
ENST00000448407.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300164798.hg.1 |
NR_040247.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300164798.hg.1 |
ENST00000448407.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300164798.hg.1 |
NR_040247.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300164798.hg.1 |
TC1300009701.hg.1 |
Clariom D TC |