Assay Details
Gene Symbol: DAOA-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_040247.1 | 6 - 7 | 949 | 79 | ||
GenBank mRNA | AY138548.1 | - | 6 - 7 | 949 | 79 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | long intergenic non-protein coding RNA 1309 | LINC01309 | TCONS_l2_00007459 | -2.0mb | Anti-sense | Chr. 13 - 103425200 - 103427685 on NCBI Build GRCh38 |
Downstream | D-amino acid oxidase activator | DAOA |
LG72, SG72 |
3.4kb | Anti-sense | Chr. 13 - 105465867 - 105491034 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC13000857.hg.1
Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Human Whole Genome ChIP Tiling Array | A_17_P29556164 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P29555777 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09106717 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P29556087 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09107044 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P29556159 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P29556083 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P29556132 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09107120 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09107113 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P29556151 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P29555785 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P29555774 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P29556123 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P29556118 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P29556134 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P29556125 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09106725 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09107027 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P29556139 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09107101 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P29556165 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P29555758 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09107118 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P29556094 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09107028 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09107039 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09107029 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P29556175 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P29556157 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P29556149 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P29555759 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P29556161 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P29556135 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09107111 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P29556143 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09107041 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P29556131 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09107040 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P29556171 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P29555769 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09106732 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P29556140 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09107070 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09107054 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P29556141 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P29556084 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P29556120 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P29556098 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P29555791 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P29556150 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09107024 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P29556117 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09106727 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09107112 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P29556156 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P29556121 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P29555765 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P29555762 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P29555768 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09107025 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09107114 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P29556101 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09106737 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09107071 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P29555760 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P29556102 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P29555790 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P29556177 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P29556160 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09107049 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P29556137 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09107047 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P29556119 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P29555776 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P29556152 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P29556144 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P29555786 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09107051 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P16666996 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P29556162 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P16666998 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P29555780 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09107055 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P29556153 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P29556113 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P29556107 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P29555787 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P29555778 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09107087 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P29556145 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P29556124 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P29556166 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P29556130 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P16666991 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P29556097 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P29555761 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P29555783 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P16666947 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P29556111 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P29556089 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P29556133 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P16666989 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P29556105 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P29556092 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P29555784 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P29556116 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P29556108 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P29556093 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09107033 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P29556146 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P29555782 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P29555756 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P29555766 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P29556138 | - | - |
Agilent SurePrint G3 Human CGH Microarray 4x180K | 136314 | - | - |
Agilent SurePrint G3 Human CGH Microarray 4x180K | 62052 | - | - |
Agilent SurePrint G3 Human CGH Microarray 4x180K | 111422 | AK093130.1 | genbank_mrna_human |
Agilent SurePrint G3 Human CGH Microarray 4x180K | 111422 | AX747943.1 | genbank_mrna_human |
Agilent SurePrint G3 Human CGH Microarray 4x180K | 111422 | BC033501.1 | genbank_mrna_human |
Agilent SurePrint G3 Human CGH Microarray 4x180K | 111422 | BC106951.2 | genbank_mrna_human |
Agilent SurePrint G3 Human CGH Microarray 4x180K | 111422 | BC106952.1 | genbank_mrna_human |
Agilent SurePrint G3 Human CGH Microarray 4x180K | 111422 | NM_001037225.1 | refseq_mrna_human_NM |
Agilent SurePrint G3 Human CGH Microarray 8x60K | 43987 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 898677 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 125172 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 517256 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 961128 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 64287 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 325371 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 707231 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 870668 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 69642 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3801258 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_23_P306240 | - | - |
Agilent Whole Human Genome Oligo Microarray 4x44K | 42057 | - | - |
Agilent Human Genome CGH Microarray 44B | A_14_P127778 | - | - |
Agilent Human Genome CGH Microarray 44A | A_14_P127778 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 10328 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 22780 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3801258 | - | - |
SurePrint G3 Human GE Microarray G4851A | A_33_P3801258 | - | - |
Whole Human Genome Oligo Microarray G4112A | A_23_P306240 | - | - |
Agilent Human Genome CGH Microarray 105A (G4412A) | A_14_P127778 | - | - |
Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_16_P40085029 | - | - |
Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_14_P127778 | - | - |
Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_16_P19945077 | - | - |
Agilent Whole Human Genome Microarray 4x44K G4112F | A_23_P306240 | - | - |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 112492 | - | - |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 52814 | - | - |
Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 | 78649 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_14_P127778 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P19944988 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P19944999 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P40085159 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P19945077 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P19945084 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P40085029 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P19945116 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P40085148 | - | - |
Agilent Human 1B Microarray G4111A | A_32_P436974 | - | - |
Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P19945077 | - | - |
Agilent SurePrint G3 Human CGH Microarray 2x400K | A_14_P127778 | - | - |
Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P40085159 | - | - |
Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P40085029 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Gene 1.0 ST Array | 7972708 | - | - |
Affymetrix Human Gene 1.0 ST Array | 7972704 | - | - |
Affymetrix Human Gene 1.0 ST Array | 7972702 | - | - |
Affymetrix Human Gene 1.0 ST Array | 7969962 | - | - |
Affymetrix Human Gene 1.0 ST Array | 7972705 | - | - |
Affymetrix Human Gene 1.0 ST Array | 7972706 | - | - |
Affymetrix Human Gene 1.0 ST Array | 7972700 | - | - |
Affymetrix Human Gene 1.0 ST Array | 7972707 | - | - |
Affymetrix Human Gene 1.0 ST Array | 7969972 | - | - |
Affymetrix Human Gene 1.0 ST Array | 7972701 | - | - |
Affymetrix Human Gene 1.0 ST Array | 7969971 | - | - |
Affymetrix Human Gene 1.0 ST Array | 7969963 | - | - |
Affymetrix Human Gene 1.0 ST Array | 7972703 | - | - |
Affymetrix Human Gene 1.1 ST Array | 7972699 | - | - |
Affymetrix HT HG-U133+ PM Array Plate | 1553024_PM_at | - | - |
Affymetrix Human Genome U219 Array | 11762502_PM_at | - | - |
Affymetrix Human Genome U219 Array | 11762502_at | - | - |
Affymetrix Human Transcriptome Array 2.0 | PSR13009610.hg.1 | ENST00000448407.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR13009610.hg.1 | NR_040247.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR13009610.hg.1 | TC13000857.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1300009701.hg.1 | ENST00000448407.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300009701.hg.1 | ENST00000448407.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300009701.hg.1 | ENST00000448407.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300009701.hg.1 | ENST00000448407.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300009701.hg.1 | ENST00000448407.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300009701.hg.1 | ENST00000448407.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300009701.hg.1 | ENST00000448407.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300009701.hg.1 | NR_040247.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300009701.hg.1 | NR_040247.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300009701.hg.1 | NR_040247.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300009701.hg.1 | NR_040247.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300009701.hg.1 | NR_040247.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300009701.hg.1 | NR_040247.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300009701.hg.1 | NR_040247.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300007923.hg.1 | ENST00000613021.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300164792.hg.1 | ENST00000448407.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300164792.hg.1 | NR_040247.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300164792.hg.1 | ENST00000448407.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300164792.hg.1 | NR_040247.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300164792.hg.1 | TC1300009701.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1300007923.hg.1 | ENST00000613021.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300009701.hg.1 | ENST00000448407.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300009701.hg.1 | ENST00000448407.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300009701.hg.1 | ENST00000448407.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300009701.hg.1 | ENST00000448407.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300009701.hg.1 | ENST00000448407.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300009701.hg.1 | ENST00000448407.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300009701.hg.1 | ENST00000448407.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300009701.hg.1 | NR_040247.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300009701.hg.1 | NR_040247.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300009701.hg.1 | NR_040247.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300009701.hg.1 | NR_040247.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300009701.hg.1 | NR_040247.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300009701.hg.1 | NR_040247.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300009701.hg.1 | NR_040247.1 | Refseq |
Human Genome U133 Plus 2.0 Array | 1553024_PM_at | - | - |
Human Genome U133 Plus 2.0 Array | 1553024_at | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1553024_at2 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1553024_at4 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1553024_at1 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1553024_at11 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1553024_at5 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1553024_at8 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1553024_at10 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1553024_at3 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1553024_at6 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1553024_at7 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1553024_at9 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A049819 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A049819 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 224591 | - | - |
