Assay Details
Gene Symbol: LGI3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_139278.2 | NP_644807.1 | 1 - 2 | 491 | 64 | |
GenBank mRNA | AF467956.1 | - | 1 - 2 | 412 | 64 | |
AJ487518.1 | - | 1 - 2 | 621 | 64 | ||
AJ487961.1 | - | 1 - 2 | 491 | 64 | ||
AY358116.1 | - | 1 - 2 | 229 | 64 | ||
BC142997.1 | - | 1 - 2 | 411 | 8860454 | 64 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC08001035.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR08013221.hg.1 | TC08001035.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR08013221.hg.1 | ENST00000306317.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR08013221.hg.1 | NM_139278.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | ENST00000306317.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | ENST00000306317.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | ENST00000306317.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | ENST00000306317.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | ENST00000306317.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | ENST00000306317.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | ENST00000306317.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | ENST00000306317.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | ENST00000306317.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | ENST00000306317.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | ENST00000306317.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | ENST00000306317.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | ENST00000306317.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | ENST00000306317.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | ENST00000424267.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | ENST00000424267.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | ENST00000424267.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | ENST00000424267.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | ENST00000424267.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | ENST00000424267.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | ENST00000424267.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | ENST00000424267.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | ENST00000424267.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | ENST00000424267.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | ENST00000424267.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | ENST00000424267.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | ENST00000517694.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | ENST00000517694.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | ENST00000517694.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | ENST00000517694.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | ENST00000517694.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | ENST00000517694.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | ENST00000517694.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | ENST00000517694.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | ENST00000518365.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | ENST00000518365.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | ENST00000518365.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | ENST00000518365.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | ENST00000518365.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | ENST00000518365.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | ENST00000518365.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | ENST00000518365.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | ENST00000520124.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | ENST00000520124.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | ENST00000520124.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | ENST00000520124.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | ENST00000520124.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | ENST00000520124.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | ENST00000520124.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | ENST00000520124.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | ENST00000520124.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | ENST00000520124.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | ENST00000520124.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | ENST00000520124.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | ENST00000520124.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | ENST00000520124.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | ENST00000520124.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | ENST00000520124.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | ENST00000520124.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | NM_139278.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | NM_139278.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | NM_139278.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | NM_139278.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | NM_139278.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | NM_139278.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | NM_139278.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | NM_139278.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | NM_139278.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | NM_139278.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | NM_139278.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | NM_139278.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | NM_139278.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | NM_139278.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR08013219.hg.1 | TC08001035.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR08013219.hg.1 | ENST00000306317.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR08013219.hg.1 | ENST00000424267.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR08013219.hg.1 | NM_139278.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR08013217.hg.1 | TC08001035.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR08013217.hg.1 | ENST00000306317.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR08013217.hg.1 | ENST00000424267.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR08013217.hg.1 | NM_139278.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR08013220.hg.1 | TC08001035.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR08013220.hg.1 | ENST00000306317.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR08013220.hg.1 | ENST00000424267.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR08013220.hg.1 | NM_139278.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800166794.hg.1 | ENST00000306317.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800166794.hg.1 | ENST00000424267.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800166794.hg.1 | ENST00000520124.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800166794.hg.1 | NM_139278.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800166794.hg.1 | ENST00000306317.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800166794.hg.1 | ENST00000424267.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800166794.hg.1 | ENST00000520124.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800166794.hg.1 | NM_139278.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800166794.hg.1 | TC0800009831.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | ENST00000306317.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | ENST00000306317.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | ENST00000306317.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | ENST00000306317.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | ENST00000306317.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | ENST00000306317.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | ENST00000306317.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | ENST00000306317.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | ENST00000306317.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | ENST00000306317.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | ENST00000306317.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | ENST00000306317.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | ENST00000306317.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | ENST00000306317.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | ENST00000424267.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | ENST00000424267.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | ENST00000424267.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | ENST00000424267.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | ENST00000424267.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | ENST00000424267.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | ENST00000424267.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | ENST00000424267.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | ENST00000424267.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | ENST00000424267.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | ENST00000424267.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | ENST00000424267.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | ENST00000517694.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | ENST00000517694.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | ENST00000517694.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | ENST00000517694.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | ENST00000517694.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | ENST00000517694.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | ENST00000517694.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | ENST00000517694.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | ENST00000518365.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | ENST00000518365.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | ENST00000518365.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | ENST00000518365.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | ENST00000518365.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | ENST00000518365.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | ENST00000518365.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | ENST00000518365.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | ENST00000520124.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | ENST00000520124.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | ENST00000520124.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | ENST00000520124.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | ENST00000520124.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | ENST00000520124.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | ENST00000520124.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | ENST00000520124.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | ENST00000520124.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | ENST00000520124.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | ENST00000520124.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | ENST00000520124.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | ENST00000520124.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | ENST00000520124.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | ENST00000520124.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | ENST00000520124.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | ENST00000520124.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | NM_139278.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | NM_139278.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | NM_139278.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | NM_139278.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | NM_139278.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | NM_139278.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | NM_139278.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | NM_139278.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | NM_139278.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | NM_139278.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | NM_139278.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | NM_139278.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | NM_139278.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | NM_139278.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800166792.hg.1 | ENST00000306317.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800166792.hg.1 | ENST00000424267.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800166792.hg.1 | ENST00000518365.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800166792.hg.1 | ENST00000520124.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800166792.hg.1 | NM_139278.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800166792.hg.1 | ENST00000306317.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800166792.hg.1 | ENST00000424267.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800166792.hg.1 | ENST00000518365.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800166792.hg.1 | ENST00000520124.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800166792.hg.1 | NM_139278.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800166792.hg.1 | TC0800009831.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0800166795.hg.1 | ENST00000306317.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800166795.hg.1 | ENST00000520124.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800166795.hg.1 | NM_139278.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800166795.hg.1 | ENST00000306317.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800166795.hg.1 | ENST00000520124.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800166795.hg.1 | NM_139278.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800166795.hg.1 | TC0800009831.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0800166790.hg.1 | ENST00000306317.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800166790.hg.1 | ENST00000424267.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800166790.hg.1 | ENST00000517694.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800166790.hg.1 | ENST00000518365.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800166790.hg.1 | ENST00000520124.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800166790.hg.1 | NM_139278.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800166790.hg.1 | ENST00000306317.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800166790.hg.1 | ENST00000424267.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800166790.hg.1 | ENST00000517694.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800166790.hg.1 | ENST00000518365.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800166790.hg.1 | ENST00000520124.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800166790.hg.1 | NM_139278.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800166790.hg.1 | TC0800009831.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0800166793.hg.1 | ENST00000306317.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800166793.hg.1 | ENST00000424267.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800166793.hg.1 | ENST00000518365.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800166793.hg.1 | ENST00000520124.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800166793.hg.1 | NM_139278.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800166793.hg.1 | ENST00000306317.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800166793.hg.1 | ENST00000424267.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800166793.hg.1 | ENST00000518365.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800166793.hg.1 | ENST00000520124.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800166793.hg.1 | NM_139278.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800166793.hg.1 | TC0800009831.hg.1 | Clariom D TC |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 212262 | - | - |
