Assay Details
Gene Symbol: LGI3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_139278.2 | NP_644807.1 | 2 - 3 | 564 | 99 | |
GenBank mRNA | AF467956.1 | - | 2 - 3 | 485 | 99 | |
AJ487518.1 | - | 2 - 3 | 694 | 99 | ||
AJ487961.1 | - | 2 - 3 | 564 | 99 | ||
AY358116.1 | - | 2 - 3 | 302 | 99 | ||
BC142997.1 | - | 2 - 3 | 484 | 8860454 | 99 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC08001035.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | ENST00000306317.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | ENST00000306317.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | ENST00000306317.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | ENST00000306317.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | ENST00000306317.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | ENST00000306317.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | ENST00000306317.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | ENST00000306317.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | ENST00000306317.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | ENST00000306317.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | ENST00000306317.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | ENST00000306317.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | ENST00000306317.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | ENST00000306317.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | ENST00000424267.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | ENST00000424267.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | ENST00000424267.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | ENST00000424267.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | ENST00000424267.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | ENST00000424267.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | ENST00000424267.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | ENST00000424267.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | ENST00000424267.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | ENST00000424267.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | ENST00000424267.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | ENST00000424267.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | ENST00000517694.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | ENST00000517694.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | ENST00000517694.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | ENST00000517694.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | ENST00000517694.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | ENST00000517694.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | ENST00000517694.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | ENST00000517694.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | ENST00000518365.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | ENST00000518365.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | ENST00000518365.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | ENST00000518365.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | ENST00000518365.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | ENST00000518365.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | ENST00000518365.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | ENST00000518365.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | ENST00000520124.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | ENST00000520124.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | ENST00000520124.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | ENST00000520124.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | ENST00000520124.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | ENST00000520124.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | ENST00000520124.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | ENST00000520124.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | ENST00000520124.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | ENST00000520124.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | ENST00000520124.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | ENST00000520124.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | ENST00000520124.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | ENST00000520124.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | ENST00000520124.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | ENST00000520124.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | ENST00000520124.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | NM_139278.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | NM_139278.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | NM_139278.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | NM_139278.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | NM_139278.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | NM_139278.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | NM_139278.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | NM_139278.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | NM_139278.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | NM_139278.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | NM_139278.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | NM_139278.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | NM_139278.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800009831.hg.1 | NM_139278.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR08013215.hg.1 | TC08001035.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR08013215.hg.1 | ENST00000306317.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR08013215.hg.1 | ENST00000424267.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR08013215.hg.1 | NM_139278.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | ENST00000306317.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | ENST00000306317.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | ENST00000306317.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | ENST00000306317.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | ENST00000306317.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | ENST00000306317.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | ENST00000306317.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | ENST00000306317.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | ENST00000306317.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | ENST00000306317.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | ENST00000306317.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | ENST00000306317.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | ENST00000306317.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | ENST00000306317.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | ENST00000424267.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | ENST00000424267.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | ENST00000424267.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | ENST00000424267.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | ENST00000424267.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | ENST00000424267.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | ENST00000424267.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | ENST00000424267.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | ENST00000424267.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | ENST00000424267.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | ENST00000424267.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | ENST00000424267.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | ENST00000517694.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | ENST00000517694.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | ENST00000517694.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | ENST00000517694.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | ENST00000517694.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | ENST00000517694.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | ENST00000517694.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | ENST00000517694.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | ENST00000518365.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | ENST00000518365.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | ENST00000518365.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | ENST00000518365.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | ENST00000518365.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | ENST00000518365.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | ENST00000518365.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | ENST00000518365.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | ENST00000520124.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | ENST00000520124.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | ENST00000520124.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | ENST00000520124.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | ENST00000520124.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | ENST00000520124.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | ENST00000520124.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | ENST00000520124.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | ENST00000520124.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | ENST00000520124.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | ENST00000520124.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | ENST00000520124.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | ENST00000520124.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | ENST00000520124.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | ENST00000520124.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | ENST00000520124.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | ENST00000520124.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | NM_139278.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | NM_139278.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | NM_139278.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | NM_139278.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | NM_139278.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | NM_139278.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | NM_139278.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | NM_139278.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | NM_139278.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | NM_139278.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | NM_139278.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | NM_139278.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | NM_139278.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800009831.hg.1 | NM_139278.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800166788.hg.1 | ENST00000306317.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800166788.hg.1 | ENST00000424267.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800166788.hg.1 | ENST00000517694.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800166788.hg.1 | ENST00000518365.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800166788.hg.1 | ENST00000520124.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800166788.hg.1 | NM_139278.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800166788.hg.1 | ENST00000306317.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800166788.hg.1 | ENST00000424267.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800166788.hg.1 | ENST00000517694.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800166788.hg.1 | ENST00000518365.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800166788.hg.1 | ENST00000520124.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800166788.hg.1 | NM_139278.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800166788.hg.1 | TC0800009831.hg.1 | Clariom D TC |
