Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR11015272.hg.1 |
ENST00000431683.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11015272.hg.1 |
NM_001080407.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11015272.hg.1 |
TC11001216.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1100172246.hg.1 |
ENST00000410100.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100172246.hg.1 |
ENST00000431683.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100172246.hg.1 |
ENST00000455971.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100172246.hg.1 |
ENST00000467068.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100172246.hg.1 |
ENST00000486034.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100172246.hg.1 |
ENST00000498012.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100172246.hg.1 |
NM_001080407.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100172246.hg.1 |
ENST00000410100.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100172246.hg.1 |
ENST00000431683.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100172246.hg.1 |
ENST00000455971.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100172246.hg.1 |
ENST00000467068.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100172246.hg.1 |
ENST00000486034.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100172246.hg.1 |
ENST00000498012.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100172246.hg.1 |
NM_001080407.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100172246.hg.1 |
TC1100009617.hg.1 |
Clariom D TC |