Assay Details
| Gene Symbol: TMEM51-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_027136.1 | 1 - 2 | 180 | 70 | ||
| GenBank mRNA | AK074283.1 | - | 1 - 2 | 182 | 70 | |
| AK124431.1 | - | 1 - 2 | 118 | 70 | ||
| BC128126.1 | - | 1 - 2 | 162 | 70 | ||
| BC128127.1 | - | 1 - 2 | 162 | 70 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01002240.hg.1
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR01034265.hg.1 | TC01002240.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01034265.hg.1 | ENST00000310916.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01034265.hg.1 | NR_027136.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012990.hg.1 | ENST00000310916.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012990.hg.1 | ENST00000310916.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012990.hg.1 | ENST00000310916.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012990.hg.1 | ENST00000310916.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012990.hg.1 | ENST00000310916.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012990.hg.1 | ENST00000310916.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012990.hg.1 | ENST00000310916.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012990.hg.1 | ENST00000310916.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012990.hg.1 | ENST00000310916.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012990.hg.1 | ENST00000310916.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012990.hg.1 | ENST00000310916.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012990.hg.1 | ENST00000310916.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012990.hg.1 | ENST00000310916.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012990.hg.1 | ENST00000310916.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012990.hg.1 | ENST00000310916.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012990.hg.1 | ENST00000310916.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012990.hg.1 | ENST00000310916.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012990.hg.1 | ENST00000310916.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012990.hg.1 | ENST00000310916.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012990.hg.1 | ENST00000404665.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012990.hg.1 | ENST00000404665.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012990.hg.1 | ENST00000404665.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012990.hg.1 | ENST00000404665.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012990.hg.1 | ENST00000404665.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012990.hg.1 | ENST00000404665.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012990.hg.1 | ENST00000404665.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012990.hg.1 | ENST00000404665.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012990.hg.1 | ENST00000404665.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012990.hg.1 | ENST00000404665.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012990.hg.1 | ENST00000404665.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012990.hg.1 | ENST00000404665.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012990.hg.1 | ENST00000404665.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012990.hg.1 | ENST00000404665.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012990.hg.1 | NR_027136.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012990.hg.1 | NR_027136.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012990.hg.1 | NR_027136.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012990.hg.1 | NR_027136.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012990.hg.1 | NR_027136.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012990.hg.1 | NR_027136.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012990.hg.1 | NR_027136.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012990.hg.1 | NR_027136.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012990.hg.1 | NR_027136.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01034264.hg.1 | TC01002240.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01034264.hg.1 | ENST00000310916.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01034264.hg.1 | NR_027136.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01034263.hg.1 | TC01002240.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01034263.hg.1 | ENST00000310916.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01034263.hg.1 | NR_027136.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100195769.hg.1 | ENST00000310916.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100195769.hg.1 | ENST00000310916.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100195769.hg.1 | ENST00000404665.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100195769.hg.1 | ENST00000404665.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100195769.hg.1 | NR_027136.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100195769.hg.1 | ENST00000310916.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100195769.hg.1 | ENST00000310916.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100195769.hg.1 | ENST00000404665.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100195769.hg.1 | ENST00000404665.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100195769.hg.1 | NR_027136.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100195769.hg.1 | TC0100012990.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100195770.hg.1 | ENST00000310916.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100195770.hg.1 | ENST00000310916.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100195770.hg.1 | NR_027136.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100195770.hg.1 | ENST00000310916.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100195770.hg.1 | ENST00000310916.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100195770.hg.1 | NR_027136.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100195770.hg.1 | TC0100012990.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100195768.hg.1 | ENST00000310916.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100195768.hg.1 | ENST00000310916.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100195768.hg.1 | ENST00000404665.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100195768.hg.1 | ENST00000404665.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100195768.hg.1 | NR_027136.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100195768.hg.1 | ENST00000310916.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100195768.hg.1 | ENST00000310916.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100195768.hg.1 | ENST00000404665.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100195768.hg.1 | ENST00000404665.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100195768.hg.1 | NR_027136.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100195768.hg.1 | TC0100012990.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100012990.hg.1 | ENST00000310916.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012990.hg.1 | ENST00000310916.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012990.hg.1 | ENST00000310916.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012990.hg.1 | ENST00000310916.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012990.hg.1 | ENST00000310916.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012990.hg.1 | ENST00000310916.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012990.hg.1 | ENST00000310916.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012990.hg.1 | ENST00000310916.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012990.hg.1 | ENST00000310916.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012990.hg.1 | ENST00000310916.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012990.hg.1 | ENST00000310916.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012990.hg.1 | ENST00000310916.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012990.hg.1 | ENST00000310916.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012990.hg.1 | ENST00000310916.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012990.hg.1 | ENST00000310916.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012990.hg.1 | ENST00000310916.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012990.hg.1 | ENST00000310916.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012990.hg.1 | ENST00000310916.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012990.hg.1 | ENST00000310916.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012990.hg.1 | ENST00000404665.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012990.hg.1 | ENST00000404665.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012990.hg.1 | ENST00000404665.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012990.hg.1 | ENST00000404665.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012990.hg.1 | ENST00000404665.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012990.hg.1 | ENST00000404665.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012990.hg.1 | ENST00000404665.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012990.hg.1 | ENST00000404665.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012990.hg.1 | ENST00000404665.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012990.hg.1 | ENST00000404665.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012990.hg.1 | ENST00000404665.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012990.hg.1 | ENST00000404665.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012990.hg.1 | ENST00000404665.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012990.hg.1 | ENST00000404665.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012990.hg.1 | NR_027136.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012990.hg.1 | NR_027136.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012990.hg.1 | NR_027136.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012990.hg.1 | NR_027136.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012990.hg.1 | NR_027136.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012990.hg.1 | NR_027136.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012990.hg.1 | NR_027136.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012990.hg.1 | NR_027136.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012990.hg.1 | NR_027136.1 | Refseq |

TMEM51-AS1
Genomic Map