Assay Details
Gene Symbol: TMEM51-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_027136.1 | 2 - 3 | 322 | 80 | ||
GenBank mRNA | AK074283.1 | - | 2 - 3 | 324 | 80 | |
AK124431.1 | - | 2 - 3 | 260 | 80 | ||
BC128126.1 | - | 2 - 3 | 304 | 80 | ||
BC128127.1 | - | 2 - 3 | 304 | 80 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC01002240.hg.1
Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR01034262.hg.1 | TC01002240.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01034262.hg.1 | ENST00000310916.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01034262.hg.1 | NR_027136.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012990.hg.1 | ENST00000310916.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012990.hg.1 | ENST00000310916.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012990.hg.1 | ENST00000310916.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012990.hg.1 | ENST00000310916.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012990.hg.1 | ENST00000310916.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012990.hg.1 | ENST00000310916.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012990.hg.1 | ENST00000310916.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012990.hg.1 | ENST00000310916.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012990.hg.1 | ENST00000310916.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012990.hg.1 | ENST00000310916.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012990.hg.1 | ENST00000310916.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012990.hg.1 | ENST00000310916.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012990.hg.1 | ENST00000310916.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012990.hg.1 | ENST00000310916.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012990.hg.1 | ENST00000310916.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012990.hg.1 | ENST00000310916.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012990.hg.1 | ENST00000310916.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012990.hg.1 | ENST00000310916.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012990.hg.1 | ENST00000310916.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012990.hg.1 | ENST00000404665.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012990.hg.1 | ENST00000404665.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012990.hg.1 | ENST00000404665.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012990.hg.1 | ENST00000404665.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012990.hg.1 | ENST00000404665.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012990.hg.1 | ENST00000404665.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012990.hg.1 | ENST00000404665.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012990.hg.1 | ENST00000404665.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012990.hg.1 | ENST00000404665.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012990.hg.1 | ENST00000404665.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012990.hg.1 | ENST00000404665.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012990.hg.1 | ENST00000404665.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012990.hg.1 | ENST00000404665.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012990.hg.1 | ENST00000404665.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012990.hg.1 | NR_027136.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012990.hg.1 | NR_027136.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012990.hg.1 | NR_027136.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012990.hg.1 | NR_027136.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012990.hg.1 | NR_027136.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012990.hg.1 | NR_027136.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012990.hg.1 | NR_027136.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012990.hg.1 | NR_027136.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012990.hg.1 | NR_027136.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100195766.hg.1 | ENST00000310916.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100195766.hg.1 | ENST00000310916.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100195766.hg.1 | ENST00000404665.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100195766.hg.1 | ENST00000404665.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100195766.hg.1 | NR_027136.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100195766.hg.1 | ENST00000310916.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100195766.hg.1 | ENST00000310916.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100195766.hg.1 | ENST00000404665.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100195766.hg.1 | ENST00000404665.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100195766.hg.1 | NR_027136.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100195766.hg.1 | TC0100012990.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100195767.hg.1 | ENST00000310916.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100195767.hg.1 | ENST00000310916.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100195767.hg.1 | ENST00000404665.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100195767.hg.1 | ENST00000404665.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100195767.hg.1 | NR_027136.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100195767.hg.1 | ENST00000310916.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100195767.hg.1 | ENST00000310916.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100195767.hg.1 | ENST00000404665.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100195767.hg.1 | ENST00000404665.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100195767.hg.1 | NR_027136.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100195767.hg.1 | TC0100012990.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0100012990.hg.1 | ENST00000310916.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012990.hg.1 | ENST00000310916.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012990.hg.1 | ENST00000310916.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012990.hg.1 | ENST00000310916.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012990.hg.1 | ENST00000310916.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012990.hg.1 | ENST00000310916.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012990.hg.1 | ENST00000310916.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012990.hg.1 | ENST00000310916.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012990.hg.1 | ENST00000310916.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012990.hg.1 | ENST00000310916.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012990.hg.1 | ENST00000310916.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012990.hg.1 | ENST00000310916.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012990.hg.1 | ENST00000310916.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012990.hg.1 | ENST00000310916.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012990.hg.1 | ENST00000310916.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012990.hg.1 | ENST00000310916.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012990.hg.1 | ENST00000310916.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012990.hg.1 | ENST00000310916.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012990.hg.1 | ENST00000310916.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012990.hg.1 | ENST00000404665.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012990.hg.1 | ENST00000404665.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012990.hg.1 | ENST00000404665.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012990.hg.1 | ENST00000404665.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012990.hg.1 | ENST00000404665.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012990.hg.1 | ENST00000404665.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012990.hg.1 | ENST00000404665.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012990.hg.1 | ENST00000404665.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012990.hg.1 | ENST00000404665.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012990.hg.1 | ENST00000404665.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012990.hg.1 | ENST00000404665.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012990.hg.1 | ENST00000404665.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012990.hg.1 | ENST00000404665.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012990.hg.1 | ENST00000404665.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012990.hg.1 | NR_027136.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012990.hg.1 | NR_027136.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012990.hg.1 | NR_027136.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012990.hg.1 | NR_027136.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012990.hg.1 | NR_027136.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012990.hg.1 | NR_027136.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012990.hg.1 | NR_027136.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012990.hg.1 | NR_027136.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012990.hg.1 | NR_027136.1 | Refseq |
