Assay Details
Gene Symbol: TMEM51-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK074283.1 | - | 3 - 4 | 411 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0100012990.hg.1 | ENST00000310916.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012990.hg.1 | ENST00000310916.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012990.hg.1 | ENST00000310916.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012990.hg.1 | ENST00000310916.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012990.hg.1 | ENST00000310916.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012990.hg.1 | ENST00000310916.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012990.hg.1 | ENST00000310916.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012990.hg.1 | ENST00000310916.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012990.hg.1 | ENST00000310916.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012990.hg.1 | ENST00000310916.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012990.hg.1 | ENST00000310916.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012990.hg.1 | ENST00000310916.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012990.hg.1 | ENST00000310916.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012990.hg.1 | ENST00000310916.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012990.hg.1 | ENST00000310916.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012990.hg.1 | ENST00000310916.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012990.hg.1 | ENST00000310916.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012990.hg.1 | ENST00000310916.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012990.hg.1 | ENST00000310916.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012990.hg.1 | ENST00000404665.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012990.hg.1 | ENST00000404665.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012990.hg.1 | ENST00000404665.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012990.hg.1 | ENST00000404665.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012990.hg.1 | ENST00000404665.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012990.hg.1 | ENST00000404665.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012990.hg.1 | ENST00000404665.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012990.hg.1 | ENST00000404665.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012990.hg.1 | ENST00000404665.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012990.hg.1 | ENST00000404665.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012990.hg.1 | ENST00000404665.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012990.hg.1 | ENST00000404665.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012990.hg.1 | ENST00000404665.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012990.hg.1 | ENST00000404665.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012990.hg.1 | NR_027136.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012990.hg.1 | NR_027136.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012990.hg.1 | NR_027136.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012990.hg.1 | NR_027136.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012990.hg.1 | NR_027136.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012990.hg.1 | NR_027136.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012990.hg.1 | NR_027136.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012990.hg.1 | NR_027136.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012990.hg.1 | NR_027136.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012990.hg.1 | ENST00000310916.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012990.hg.1 | ENST00000310916.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012990.hg.1 | ENST00000310916.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012990.hg.1 | ENST00000310916.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012990.hg.1 | ENST00000310916.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012990.hg.1 | ENST00000310916.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012990.hg.1 | ENST00000310916.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012990.hg.1 | ENST00000310916.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012990.hg.1 | ENST00000310916.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012990.hg.1 | ENST00000310916.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012990.hg.1 | ENST00000310916.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012990.hg.1 | ENST00000310916.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012990.hg.1 | ENST00000310916.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012990.hg.1 | ENST00000310916.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012990.hg.1 | ENST00000310916.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012990.hg.1 | ENST00000310916.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012990.hg.1 | ENST00000310916.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012990.hg.1 | ENST00000310916.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012990.hg.1 | ENST00000310916.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012990.hg.1 | ENST00000404665.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012990.hg.1 | ENST00000404665.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012990.hg.1 | ENST00000404665.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012990.hg.1 | ENST00000404665.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012990.hg.1 | ENST00000404665.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012990.hg.1 | ENST00000404665.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012990.hg.1 | ENST00000404665.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012990.hg.1 | ENST00000404665.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012990.hg.1 | ENST00000404665.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012990.hg.1 | ENST00000404665.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012990.hg.1 | ENST00000404665.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012990.hg.1 | ENST00000404665.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012990.hg.1 | ENST00000404665.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012990.hg.1 | ENST00000404665.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012990.hg.1 | NR_027136.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012990.hg.1 | NR_027136.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012990.hg.1 | NR_027136.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012990.hg.1 | NR_027136.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012990.hg.1 | NR_027136.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012990.hg.1 | NR_027136.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012990.hg.1 | NR_027136.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012990.hg.1 | NR_027136.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012990.hg.1 | NR_027136.1 | Refseq |
