Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Clariom D Assay |
PSR0X00168944.hg.1 |
ENST00000253577.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00168944.hg.1 |
ENST00000339447.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00168944.hg.1 |
ENST00000373394.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00168944.hg.1 |
ENST00000526404.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00168944.hg.1 |
ENST00000529949.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00168944.hg.1 |
ENST00000620875.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00168944.hg.1 |
NM_001271696.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00168944.hg.1 |
NM_001271697.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00168944.hg.1 |
NM_001271698.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00168944.hg.1 |
NM_001271699.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00168944.hg.1 |
NM_004299.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00168944.hg.1 |
ENST00000253577.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00168944.hg.1 |
ENST00000339447.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00168944.hg.1 |
ENST00000373394.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00168944.hg.1 |
ENST00000526404.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00168944.hg.1 |
ENST00000529949.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00168944.hg.1 |
ENST00000620875.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00168944.hg.1 |
NM_001271696.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00168944.hg.1 |
NM_001271697.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00168944.hg.1 |
NM_001271698.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00168944.hg.1 |
NM_001271699.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00168944.hg.1 |
NM_004299.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00168944.hg.1 |
TC0X00010099.hg.1 |
Clariom D TC |