Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009425.hg.1 |
ENST00000376678.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009425.hg.1 |
ENST00000376678.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009425.hg.1 |
ENST00000376678.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009425.hg.1 |
ENST00000376678.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009425.hg.1 |
ENST00000376678.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009425.hg.1 |
ENST00000376678.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009425.hg.1 |
ENST00000376678.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009425.hg.1 |
ENST00000376678.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009425.hg.1 |
ENST00000376678.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009425.hg.1 |
ENST00000376678.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009425.hg.1 |
ENST00000376678.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009425.hg.1 |
ENST00000376678.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009425.hg.1 |
ENST00000376678.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009425.hg.1 |
ENST00000376678.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009425.hg.1 |
ENST00000376678.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009425.hg.1 |
ENST00000376678.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009425.hg.1 |
ENST00000536191.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009425.hg.1 |
ENST00000536191.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009425.hg.1 |
ENST00000536191.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009425.hg.1 |
ENST00000536191.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009425.hg.1 |
NR_026670.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009425.hg.1 |
NR_026670.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009425.hg.1 |
NR_026670.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009425.hg.1 |
NR_026670.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009425.hg.1 |
NR_026670.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009425.hg.1 |
NR_026670.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009425.hg.1 |
NR_026670.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009425.hg.1 |
NR_026670.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12013700.hg.1 |
ENST00000376678.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12013700.hg.1 |
ENST00000536191.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12013700.hg.1 |
NR_026670.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12013700.hg.1 |
TC12001042.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR12013701.hg.1 |
ENST00000376678.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12013701.hg.1 |
NR_026670.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12013701.hg.1 |
TC12001042.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1200170187.hg.1 |
ENST00000376678.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200170187.hg.1 |
ENST00000376678.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200170187.hg.1 |
ENST00000536191.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200170187.hg.1 |
NR_026670.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200170187.hg.1 |
ENST00000376678.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200170187.hg.1 |
ENST00000376678.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200170187.hg.1 |
ENST00000536191.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200170187.hg.1 |
NR_026670.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200170187.hg.1 |
TC1200009425.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1200170188.hg.1 |
ENST00000376678.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200170188.hg.1 |
ENST00000376678.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200170188.hg.1 |
ENST00000536191.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200170188.hg.1 |
NR_026670.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200170188.hg.1 |
ENST00000376678.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200170188.hg.1 |
ENST00000376678.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200170188.hg.1 |
ENST00000536191.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200170188.hg.1 |
NR_026670.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200170188.hg.1 |
TC1200009425.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1200170189.hg.1 |
ENST00000376678.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200170189.hg.1 |
ENST00000376678.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200170189.hg.1 |
NR_026670.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200170189.hg.1 |
ENST00000376678.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200170189.hg.1 |
ENST00000376678.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200170189.hg.1 |
NR_026670.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200170189.hg.1 |
TC1200009425.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1200009425.hg.1 |
ENST00000376678.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009425.hg.1 |
ENST00000376678.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009425.hg.1 |
ENST00000376678.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009425.hg.1 |
ENST00000376678.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009425.hg.1 |
ENST00000376678.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009425.hg.1 |
ENST00000376678.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009425.hg.1 |
ENST00000376678.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009425.hg.1 |
ENST00000376678.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009425.hg.1 |
ENST00000376678.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009425.hg.1 |
ENST00000376678.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009425.hg.1 |
ENST00000376678.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009425.hg.1 |
ENST00000376678.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009425.hg.1 |
ENST00000376678.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009425.hg.1 |
ENST00000376678.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009425.hg.1 |
ENST00000376678.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009425.hg.1 |
ENST00000376678.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009425.hg.1 |
ENST00000536191.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009425.hg.1 |
ENST00000536191.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009425.hg.1 |
ENST00000536191.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009425.hg.1 |
ENST00000536191.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009425.hg.1 |
NR_026670.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009425.hg.1 |
NR_026670.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009425.hg.1 |
NR_026670.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009425.hg.1 |
NR_026670.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009425.hg.1 |
NR_026670.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009425.hg.1 |
NR_026670.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009425.hg.1 |
NR_026670.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009425.hg.1 |
NR_026670.2 |
Refseq |