Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR06004856.hg.1 |
TC06000413.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06004856.hg.1 |
ENST00000374662.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06004856.hg.1 |
ENST00000469186.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06004856.hg.1 |
NM_014234.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_qbl_hap6001883.hg.1 |
TC6_qbl_hap6000094.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_qbl_hap6001883.hg.1 |
NM_014234.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_dbb_hap3001902.hg.1 |
TC6_dbb_hap3000095.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_dbb_hap3001902.hg.1 |
NM_014234.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007685.hg.1 |
ENST00000374662.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007685.hg.1 |
ENST00000374662.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007685.hg.1 |
ENST00000374662.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007685.hg.1 |
ENST00000374662.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007685.hg.1 |
ENST00000374662.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007685.hg.1 |
ENST00000374662.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007685.hg.1 |
ENST00000374662.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007685.hg.1 |
ENST00000374662.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007685.hg.1 |
ENST00000374662.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007685.hg.1 |
ENST00000374662.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007685.hg.1 |
ENST00000469186.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007685.hg.1 |
ENST00000469186.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007685.hg.1 |
ENST00000469186.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007685.hg.1 |
ENST00000469186.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007685.hg.1 |
ENST00000469186.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007685.hg.1 |
ENST00000469186.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007685.hg.1 |
ENST00000469186.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007685.hg.1 |
ENST00000469186.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007685.hg.1 |
ENST00000469186.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mcf_hap5001697.hg.1 |
TC6_mcf_hap5000088.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mcf_hap5001697.hg.1 |
NM_014234.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mann_hap4001530.hg.1 |
TC6_mann_hap4000087.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mann_hap4001530.hg.1 |
NM_014234.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_cox_hap2001919.hg.1 |
TC6_cox_hap2000103.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_cox_hap2001919.hg.1 |
NM_014234.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600153122.hg.1 |
ENST00000374662.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600153122.hg.1 |
ENST00000469186.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600153122.hg.1 |
ENST00000374662.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600153122.hg.1 |
ENST00000469186.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600153122.hg.1 |
TC0600007685.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0600007685.hg.1 |
ENST00000374662.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007685.hg.1 |
ENST00000374662.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007685.hg.1 |
ENST00000374662.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007685.hg.1 |
ENST00000374662.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007685.hg.1 |
ENST00000374662.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007685.hg.1 |
ENST00000374662.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007685.hg.1 |
ENST00000374662.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007685.hg.1 |
ENST00000374662.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007685.hg.1 |
ENST00000374662.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007685.hg.1 |
ENST00000374662.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007685.hg.1 |
ENST00000469186.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007685.hg.1 |
ENST00000469186.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007685.hg.1 |
ENST00000469186.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007685.hg.1 |
ENST00000469186.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007685.hg.1 |
ENST00000469186.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007685.hg.1 |
ENST00000469186.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007685.hg.1 |
ENST00000469186.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007685.hg.1 |
ENST00000469186.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007685.hg.1 |
ENST00000469186.1 |
Ensembl |