Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_cox_hap2001920.hg.1 |
TC6_cox_hap2000103.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_cox_hap2001920.hg.1 |
NM_014234.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_cox_hap2001921.hg.1 |
TC6_cox_hap2000103.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_cox_hap2001921.hg.1 |
NM_014234.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mann_hap4001531.hg.1 |
TC6_mann_hap4000087.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mann_hap4001531.hg.1 |
NM_014234.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_qbl_hap6001884.hg.1 |
TC6_qbl_hap6000094.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_qbl_hap6001884.hg.1 |
NM_014234.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mann_hap4001532.hg.1 |
TC6_mann_hap4000087.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mann_hap4001532.hg.1 |
NM_014234.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_qbl_hap6001885.hg.1 |
TC6_qbl_hap6000094.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_qbl_hap6001885.hg.1 |
NM_014234.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mcf_hap5001699.hg.1 |
TC6_mcf_hap5000088.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mcf_hap5001699.hg.1 |
NM_014234.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06004857.hg.1 |
TC06000413.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06004857.hg.1 |
ENST00000374662.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06004857.hg.1 |
ENST00000469186.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06004857.hg.1 |
NM_014234.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06004858.hg.1 |
TC06000413.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06004858.hg.1 |
ENST00000374662.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06004858.hg.1 |
ENST00000469186.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06004858.hg.1 |
NM_014234.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007685.hg.1 |
ENST00000374662.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007685.hg.1 |
ENST00000374662.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007685.hg.1 |
ENST00000374662.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007685.hg.1 |
ENST00000374662.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007685.hg.1 |
ENST00000374662.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007685.hg.1 |
ENST00000374662.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007685.hg.1 |
ENST00000374662.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007685.hg.1 |
ENST00000374662.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007685.hg.1 |
ENST00000374662.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007685.hg.1 |
ENST00000374662.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007685.hg.1 |
ENST00000469186.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007685.hg.1 |
ENST00000469186.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007685.hg.1 |
ENST00000469186.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007685.hg.1 |
ENST00000469186.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007685.hg.1 |
ENST00000469186.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007685.hg.1 |
ENST00000469186.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007685.hg.1 |
ENST00000469186.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007685.hg.1 |
ENST00000469186.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007685.hg.1 |
ENST00000469186.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_dbb_hap3001903.hg.1 |
TC6_dbb_hap3000095.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_dbb_hap3001903.hg.1 |
NM_014234.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_dbb_hap3001904.hg.1 |
TC6_dbb_hap3000095.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_dbb_hap3001904.hg.1 |
NM_014234.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mcf_hap5001698.hg.1 |
TC6_mcf_hap5000088.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mcf_hap5001698.hg.1 |
NM_014234.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007685.hg.1 |
ENST00000374662.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007685.hg.1 |
ENST00000374662.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007685.hg.1 |
ENST00000374662.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007685.hg.1 |
ENST00000374662.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007685.hg.1 |
ENST00000374662.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007685.hg.1 |
ENST00000374662.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007685.hg.1 |
ENST00000374662.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007685.hg.1 |
ENST00000374662.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007685.hg.1 |
ENST00000374662.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007685.hg.1 |
ENST00000374662.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007685.hg.1 |
ENST00000469186.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007685.hg.1 |
ENST00000469186.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007685.hg.1 |
ENST00000469186.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007685.hg.1 |
ENST00000469186.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007685.hg.1 |
ENST00000469186.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007685.hg.1 |
ENST00000469186.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007685.hg.1 |
ENST00000469186.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007685.hg.1 |
ENST00000469186.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007685.hg.1 |
ENST00000469186.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600153123.hg.1 |
ENST00000374662.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600153123.hg.1 |
ENST00000469186.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600153123.hg.1 |
ENST00000374662.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600153123.hg.1 |
ENST00000469186.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600153123.hg.1 |
TC0600007685.hg.1 |
Clariom D TC |