Assay Details
| Gene Symbol: LINC00508 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_126452.2 | 4 | 1274 | 74 | ||
| GenBank mRNA | BC032019.1 | - | 1 | 963 | 74 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC105370068 | LOC105370068 | -232.6kb | Sense | Chr. 12 - 127648896 - 127655588 on NCBI Build GRCh38 | |
| Downstream | long intergenic non-protein coding RNA 507 | LINC00507 | 30.3kb | Anti-sense | Chr. 12 - 127915410 - 127951552 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

LINC00508
Genomic Map